Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155134663-155135172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNA1
Entrez Gene ID
1942
Gene Name
ephrin A1
Gene Aliases
B61, ECKLG, EFL1, EPLG1, LERK-1, LERK1, TNFAIP4
UniGene
Hs.516664
Gene Chromosome Location
Chr.1: 155127873-155134910 on Build GRCh38
Transcripts
NM_004428.2 NM_182685.1

SNP Detail

SNP ID Chromosome Location
rs550458654 Chr.1: 155134972-155134973 on Build GRCh38
rs940565922 Chr.1: 155135145-155135145 on Build GRCh38
rs79648291 Chr.1: 155135097-155135097 on Build GRCh38
rs774117591 Chr.1: 155134748-155134748 on Build GRCh38
rs901494573 Chr.1: 155134818-155134818 on Build GRCh38
rs1005207619 Chr.1: 155134806-155134806 on Build GRCh38
rs907212363 Chr.1: 155134697-155134697 on Build GRCh38
rs989864014 Chr.1: 155134893-155134893 on Build GRCh38
rs543632739 Chr.1: 155134810-155134810 on Build GRCh38
rs925062922 Chr.1: 155134877-155134877 on Build GRCh38
rs576609812 Chr.1: 155134707-155134707 on Build GRCh38
rs974239896 Chr.1: 155134746-155134746 on Build GRCh38
rs1028928225 Chr.1: 155134763-155134763 on Build GRCh38
rs544095797 Chr.1: 155134972-155134972 on Build GRCh38
rs1015708848 Chr.1: 155134705-155134705 on Build GRCh38
rs936440564 Chr.1: 155134989-155134989 on Build GRCh38
rs200484385 Chr.1: 155134978-155134979 on Build GRCh38
rs967611682 Chr.1: 155134943-155134943 on Build GRCh38
rs11545252 Chr.1: 155134784-155134784 on Build GRCh38
rs760796408 Chr.1: 155134766-155134766 on Build GRCh38
rs575134804 Chr.1: 155134867-155134867 on Build GRCh38
rs541481987 Chr.1: 155134990-155134990 on Build GRCh38
rs1037045753 Chr.1: 155135109-155135109 on Build GRCh38
rs141515891 Chr.1: 155134699-155134699 on Build GRCh38
rs987466022 Chr.1: 155134800-155134800 on Build GRCh38
rs931347718 Chr.1: 155135105-155135105 on Build GRCh38
rs552089828 Chr.1: 155135101-155135101 on Build GRCh38
rs937319982 Chr.1: 155135057-155135057 on Build GRCh38
rs113958160 Chr.1: 155134831-155134831 on Build GRCh38
rs188263523 Chr.1: 155134945-155134945 on Build GRCh38
rs928930436 Chr.1: 155135013-155135013 on Build GRCh38
rs945084884 Chr.1: 155134833-155134833 on Build GRCh38
rs977792741 Chr.1: 155134848-155134848 on Build GRCh38
rs916404521 Chr.1: 155135034-155135034 on Build GRCh38
rs957087207 Chr.1: 155134862-155134862 on Build GRCh38
rs78066920 Chr.1: 155135003-155135003 on Build GRCh38
rs1004242313 Chr.1: 155135106-155135106 on Build GRCh38
rs559718333 Chr.1: 155134991-155134991 on Build GRCh38
rs1020368184 Chr.1: 155134924-155134924 on Build GRCh38
rs150889956 Chr.1: 155134853-155134853 on Build GRCh38
rs1054945281 Chr.1: 155135018-155135018 on Build GRCh38
rs1024271920 Chr.1: 155134689-155134689 on Build GRCh38
rs560800506 Chr.1: 155135114-155135114 on Build GRCh38
rs995881508 Chr.1: 155134823-155134823 on Build GRCh38
rs761357675 Chr.1: 155134749-155134749 on Build GRCh38
rs886267955 Chr.1: 155135123-155135123 on Build GRCh38
rs949299673 Chr.1: 155135086-155135086 on Build GRCh38
rs111787654 Chr.1: 155134858-155134858 on Build GRCh38
rs768372089 Chr.1: 155134746-155134747 on Build GRCh38

View Full Product Details