Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 47994233-47994737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUN3
Entrez Gene ID
256979
Gene Name
Sad1 and UNC84 domain containing 3
Gene Aliases
SUNC1
UniGene
Hs.406741
Gene Chromosome Location
Chr.7: 47987148-48035899 on Build GRCh38
Transcripts
NM_001030019.1 NM_001284350.1 NM_152782.3 XM_017011930.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6005964 c.825+1G>T p.? Chr.7: 47994314-47994314 on Build GRCh38
6793197 c.820T>C p.S274P Chr.7: 47994356-47994356 on Build GRCh38
7392466 c.491G>T p.W164L Chr.7: 47994456-47994456 on Build GRCh38
5939760 c.782C>T p.P261L Chr.7: 47994358-47994358 on Build GRCh38
6793194 c.717C>A p.C239* Chr.7: 47994459-47994459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775871269 Chr.7: 47994403-47994403 on Build GRCh38
rs1011441113 Chr.7: 47994661-47994661 on Build GRCh38
rs769223567 Chr.7: 47994415-47994415 on Build GRCh38
rs376459386 Chr.7: 47994275-47994275 on Build GRCh38
rs770500360 Chr.7: 47994517-47994517 on Build GRCh38
rs769913226 Chr.7: 47994266-47994266 on Build GRCh38
rs144089181 Chr.7: 47994458-47994458 on Build GRCh38
rs761199827 Chr.7: 47994413-47994413 on Build GRCh38
rs772352527 Chr.7: 47994400-47994400 on Build GRCh38
rs1021536577 Chr.7: 47994667-47994667 on Build GRCh38
rs755259561 Chr.7: 47994278-47994278 on Build GRCh38
rs762630332 Chr.7: 47994341-47994341 on Build GRCh38
rs776205338 Chr.7: 47994460-47994460 on Build GRCh38
rs878966546 Chr.7: 47994511-47994511 on Build GRCh38
rs752606115 Chr.7: 47994430-47994430 on Build GRCh38
rs188328639 Chr.7: 47994481-47994481 on Build GRCh38
rs201760001 Chr.7: 47994270-47994270 on Build GRCh38
rs759409600 Chr.7: 47994427-47994427 on Build GRCh38
rs759217141 Chr.7: 47994522-47994522 on Build GRCh38
rs767046234 Chr.7: 47994525-47994525 on Build GRCh38
rs967051992 Chr.7: 47994713-47994713 on Build GRCh38
rs779091892 Chr.7: 47994380-47994380 on Build GRCh38
rs1038718854 Chr.7: 47994340-47994340 on Build GRCh38
rs958582849 Chr.7: 47994658-47994658 on Build GRCh38
rs372204872 Chr.7: 47994497-47994497 on Build GRCh38
rs771230887 Chr.7: 47994277-47994277 on Build GRCh38
rs138815958 Chr.7: 47994357-47994357 on Build GRCh38
rs779108714 Chr.7: 47994467-47994467 on Build GRCh38
rs769257831 Chr.7: 47994490-47994490 on Build GRCh38
rs554434941 Chr.7: 47994566-47994566 on Build GRCh38
rs780429517 Chr.7: 47994480-47994480 on Build GRCh38
rs777869710 Chr.7: 47994360-47994360 on Build GRCh38
rs763093543 Chr.7: 47994702-47994702 on Build GRCh38
rs374898891 Chr.7: 47994521-47994521 on Build GRCh38
rs912323756 Chr.7: 47994446-47994446 on Build GRCh38
rs914247591 Chr.7: 47994373-47994373 on Build GRCh38
rs751452483 Chr.7: 47994348-47994348 on Build GRCh38
rs1048389505 Chr.7: 47994533-47994533 on Build GRCh38
rs756116158 Chr.7: 47994435-47994435 on Build GRCh38
rs763913442 Chr.7: 47994449-47994449 on Build GRCh38
rs543506520 Chr.7: 47994641-47994641 on Build GRCh38
rs775968599 Chr.7: 47994323-47994323 on Build GRCh38
rs758781242 Chr.7: 47994476-47994476 on Build GRCh38
rs906625809 Chr.7: 47994624-47994624 on Build GRCh38
rs368580888 Chr.7: 47994347-47994347 on Build GRCh38
rs368900181 Chr.7: 47994362-47994362 on Build GRCh38
rs1034150695 Chr.7: 47994643-47994643 on Build GRCh38
rs757155390 Chr.7: 47994458-47994458 on Build GRCh38
rs757321860 Chr.7: 47994465-47994465 on Build GRCh38
rs748874845 Chr.7: 47994513-47994513 on Build GRCh38
rs910762006 Chr.7: 47994372-47994373 on Build GRCh38
rs751425966 Chr.7: 47994450-47994450 on Build GRCh38
rs115353261 Chr.7: 47994421-47994421 on Build GRCh38
rs887097560 Chr.7: 47994550-47994550 on Build GRCh38
rs983565610 Chr.7: 47994620-47994620 on Build GRCh38
rs774145397 Chr.7: 47994423-47994423 on Build GRCh38
rs957801371 Chr.7: 47994466-47994466 on Build GRCh38
rs369258450 Chr.7: 47994329-47994329 on Build GRCh38
rs574585569 Chr.7: 47994583-47994583 on Build GRCh38
rs771191332 Chr.7: 47994372-47994372 on Build GRCh38
rs775402617 Chr.7: 47994529-47994529 on Build GRCh38
rs73111110 Chr.7: 47994330-47994330 on Build GRCh38
rs746027846 Chr.7: 47994469-47994469 on Build GRCh38
rs759870735 Chr.7: 47994300-47994300 on Build GRCh38
rs754808750 Chr.7: 47994354-47994354 on Build GRCh38
rs774735476 Chr.7: 47994283-47994283 on Build GRCh38
rs767364136 Chr.7: 47994429-47994429 on Build GRCh38
rs923268307 Chr.7: 47994567-47994567 on Build GRCh38
rs1031352916 Chr.7: 47994436-47994436 on Build GRCh38
rs772821222 Chr.7: 47994416-47994416 on Build GRCh38
rs147110748 Chr.7: 47994396-47994396 on Build GRCh38
rs567424793 Chr.7: 47994314-47994314 on Build GRCh38
rs779240969 Chr.7: 47994295-47994295 on Build GRCh38

View Full Product Details