Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 78688933-78689433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL7
Entrez Gene ID
3574
Gene Name
interleukin 7
Gene Aliases
IL-7
UniGene
Hs.591873
Gene Chromosome Location
Chr.8: 78675870-78805523 on Build GRCh38
Transcripts
XM_011517523.2 NM_016010.2 XM_005251256.1 XM_006716454.2 XM_011517539.2

Gene Symbol
ZC2HC1A
Entrez Gene ID
51101
Gene Name
zinc finger C2HC-type containing 1A
Gene Aliases
C8orf70, CGI-62, FAM164A
UniGene
Hs.271876
Gene Chromosome Location
Chr.8: 78666047-78719765 on Build GRCh38
Transcripts
XM_011517523.2 NM_016010.2 XM_005251256.1 XM_006716454.2 XM_011517539.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4777943 c.443T>C p.I148T Chr.8: 78689312-78689312 on Build GRCh38
1101877 c.441T>C p.R147R Chr.8: 78689310-78689310 on Build GRCh38
7425379 c.442A>G p.I148V Chr.8: 78689311-78689311 on Build GRCh38
6825543 c.493C>T p.R165W Chr.8: 78689362-78689362 on Build GRCh38
5492398 c.440G>A p.R147H Chr.8: 78689309-78689309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397793683 Chr.8: 78689047-78689048 on Build GRCh38
rs768463430 Chr.8: 78689389-78689389 on Build GRCh38
rs146184464 Chr.8: 78689336-78689336 on Build GRCh38
rs368612042 Chr.8: 78689211-78689211 on Build GRCh38
rs756139685 Chr.8: 78689232-78689232 on Build GRCh38
rs201769469 Chr.8: 78689328-78689328 on Build GRCh38
rs770704036 Chr.8: 78689177-78689177 on Build GRCh38
rs142420326 Chr.8: 78689258-78689258 on Build GRCh38
rs1006870134 Chr.8: 78689129-78689129 on Build GRCh38
rs746865537 Chr.8: 78689377-78689377 on Build GRCh38
rs1029741379 Chr.8: 78689068-78689068 on Build GRCh38
rs779257641 Chr.8: 78689245-78689245 on Build GRCh38
rs914791489 Chr.8: 78688972-78688972 on Build GRCh38
rs990557761 Chr.8: 78688971-78688973 on Build GRCh38
rs377254636 Chr.8: 78689214-78689214 on Build GRCh38
rs200872378 Chr.8: 78689363-78689363 on Build GRCh38
rs764732047 Chr.8: 78689342-78689342 on Build GRCh38
rs1031253180 Chr.8: 78689184-78689184 on Build GRCh38
rs17850447 Chr.8: 78689335-78689335 on Build GRCh38
rs756586522 Chr.8: 78689340-78689340 on Build GRCh38
rs769902001 Chr.8: 78689171-78689171 on Build GRCh38
rs5892662 Chr.8: 78689034-78689035 on Build GRCh38
rs148270750 Chr.8: 78689356-78689356 on Build GRCh38
rs775604140 Chr.8: 78689186-78689186 on Build GRCh38
rs564507665 Chr.8: 78689139-78689139 on Build GRCh38
rs938859960 Chr.8: 78689145-78689145 on Build GRCh38
rs1872072 Chr.8: 78689185-78689185 on Build GRCh38
rs749496429 Chr.8: 78689175-78689175 on Build GRCh38
rs751907524 Chr.8: 78689213-78689213 on Build GRCh38
rs146492124 Chr.8: 78689273-78689273 on Build GRCh38
rs778017845 Chr.8: 78689173-78689173 on Build GRCh38
rs753658944 Chr.8: 78689369-78689370 on Build GRCh38
rs547186068 Chr.8: 78689080-78689080 on Build GRCh38
rs772891311 Chr.8: 78689314-78689314 on Build GRCh38
rs759474753 Chr.8: 78689179-78689179 on Build GRCh38
rs749463362 Chr.8: 78689236-78689236 on Build GRCh38
rs928769144 Chr.8: 78689130-78689130 on Build GRCh38
rs373470064 Chr.8: 78689367-78689367 on Build GRCh38
rs761443209 Chr.8: 78689309-78689309 on Build GRCh38
rs761376001 Chr.8: 78689205-78689205 on Build GRCh38
rs141497872 Chr.8: 78689315-78689315 on Build GRCh38
rs775998190 Chr.8: 78689307-78689307 on Build GRCh38
rs200077086 Chr.8: 78689404-78689404 on Build GRCh38
rs776287698 Chr.8: 78689204-78689204 on Build GRCh38
rs753236294 Chr.8: 78689230-78689230 on Build GRCh38
rs777730540 Chr.8: 78689233-78689233 on Build GRCh38
rs763531852 Chr.8: 78689203-78689203 on Build GRCh38
rs574401853 Chr.8: 78689371-78689371 on Build GRCh38
rs373208347 Chr.8: 78689362-78689362 on Build GRCh38
rs529082146 Chr.8: 78689079-78689079 on Build GRCh38
rs760219335 Chr.8: 78689189-78689189 on Build GRCh38
rs746953463 Chr.8: 78689289-78689289 on Build GRCh38
rs994038469 Chr.8: 78689012-78689012 on Build GRCh38
rs375969699 Chr.8: 78689394-78689394 on Build GRCh38
rs775516268 Chr.8: 78689283-78689283 on Build GRCh38
rs757419307 Chr.8: 78689238-78689238 on Build GRCh38
rs528487547 Chr.8: 78689149-78689149 on Build GRCh38
rs769378447 Chr.8: 78689398-78689398 on Build GRCh38
rs891241653 Chr.8: 78689127-78689127 on Build GRCh38
rs78933754 Chr.8: 78689403-78689403 on Build GRCh38
rs200110247 Chr.8: 78689357-78689357 on Build GRCh38
rs147883140 Chr.8: 78689311-78689311 on Build GRCh38
rs59498384 Chr.8: 78689047-78689047 on Build GRCh38
rs539358457 Chr.8: 78689178-78689178 on Build GRCh38
rs768143827 Chr.8: 78689303-78689303 on Build GRCh38
rs776678836 Chr.8: 78689390-78689390 on Build GRCh38
rs569657473 Chr.8: 78689219-78689219 on Build GRCh38

View Full Product Details