Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36338125-36338671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF38
Entrez Gene ID
152006
Gene Name
ring finger protein 38
Gene Aliases
-
UniGene
Hs.333503
Gene Chromosome Location
Chr.9: 36336398-36487384 on Build GRCh38
Transcripts
NM_022781.4 NM_194328.2 NM_194329.2 NM_194330.2 NM_194332.2 XM_005251366.3 XM_005251367.3 XM_005251368.3 XM_006716721.3 XM_011517712.2 XM_011517713.2 XM_017014294.1 XM_017014295.1 XM_017014296.1 XM_017014297.1

SNP Detail

SNP ID Chromosome Location
rs564370136 Chr.9: 36338343-36338343 on Build GRCh38
rs962171652 Chr.9: 36338434-36338434 on Build GRCh38
rs977491152 Chr.9: 36338520-36338520 on Build GRCh38
rs756119692 Chr.9: 36338401-36338405 on Build GRCh38
rs894352070 Chr.9: 36338617-36338617 on Build GRCh38
rs779310327 Chr.9: 36338605-36338606 on Build GRCh38
rs551678910 Chr.9: 36338538-36338538 on Build GRCh38
rs199773884 Chr.9: 36338570-36338571 on Build GRCh38
rs145075232 Chr.9: 36338512-36338512 on Build GRCh38
rs944802540 Chr.9: 36338251-36338251 on Build GRCh38
rs931945738 Chr.9: 36338225-36338225 on Build GRCh38
rs559553045 Chr.9: 36338338-36338338 on Build GRCh38
rs577803338 Chr.9: 36338385-36338385 on Build GRCh38
rs914575904 Chr.9: 36338243-36338244 on Build GRCh38
rs966047703 Chr.9: 36338501-36338501 on Build GRCh38
rs1023980104 Chr.9: 36338372-36338372 on Build GRCh38
rs948654748 Chr.9: 36338644-36338644 on Build GRCh38
rs912551103 Chr.9: 36338489-36338489 on Build GRCh38
rs953657784 Chr.9: 36338476-36338476 on Build GRCh38
rs1003961498 Chr.9: 36338417-36338417 on Build GRCh38
rs956155191 Chr.9: 36338197-36338197 on Build GRCh38
rs1012959071 Chr.9: 36338347-36338347 on Build GRCh38
rs117301940 Chr.9: 36338287-36338287 on Build GRCh38
rs530864871 Chr.9: 36338508-36338511 on Build GRCh38
rs1048134941 Chr.9: 36338346-36338346 on Build GRCh38
rs903001941 Chr.9: 36338311-36338311 on Build GRCh38
rs1054294980 Chr.9: 36338591-36338591 on Build GRCh38
rs1015035525 Chr.9: 36338175-36338175 on Build GRCh38
rs41277103 Chr.9: 36338482-36338482 on Build GRCh38
rs906895631 Chr.9: 36338409-36338409 on Build GRCh38
rs780697341 Chr.9: 36338231-36338232 on Build GRCh38
rs931436039 Chr.9: 36338529-36338529 on Build GRCh38
rs775614317 Chr.9: 36338256-36338256 on Build GRCh38
rs563262081 Chr.9: 36338423-36338423 on Build GRCh38
rs577121838 Chr.9: 36338165-36338165 on Build GRCh38
rs778844316 Chr.9: 36338500-36338500 on Build GRCh38
rs1041863495 Chr.9: 36338252-36338252 on Build GRCh38
rs765570241 Chr.9: 36338513-36338513 on Build GRCh38
rs759509426 Chr.9: 36338229-36338229 on Build GRCh38
rs947813912 Chr.9: 36338571-36338571 on Build GRCh38
rs962222556 Chr.9: 36338179-36338179 on Build GRCh38
rs954088063 Chr.9: 36338460-36338460 on Build GRCh38
rs767588777 Chr.9: 36338232-36338232 on Build GRCh38
rs985740945 Chr.9: 36338250-36338250 on Build GRCh38
rs550451315 Chr.9: 36338615-36338615 on Build GRCh38
rs973554166 Chr.9: 36338435-36338435 on Build GRCh38
rs529666876 Chr.9: 36338422-36338422 on Build GRCh38
rs954010530 Chr.9: 36338244-36338244 on Build GRCh38
rs957825805 Chr.9: 36338428-36338428 on Build GRCh38
rs894821665 Chr.9: 36338383-36338386 on Build GRCh38
rs185921172 Chr.9: 36338589-36338589 on Build GRCh38
rs754074103 Chr.9: 36338459-36338459 on Build GRCh38
rs1054346778 Chr.9: 36338323-36338323 on Build GRCh38
rs1030775688 Chr.9: 36338190-36338190 on Build GRCh38
rs537122494 Chr.9: 36338535-36338535 on Build GRCh38
rs973605067 Chr.9: 36338185-36338185 on Build GRCh38
rs181493270 Chr.9: 36338203-36338203 on Build GRCh38
rs906468838 Chr.9: 36338422-36338426 on Build GRCh38
rs1046331457 Chr.9: 36338404-36338404 on Build GRCh38
rs548137674 Chr.9: 36338453-36338454 on Build GRCh38
rs935850482 Chr.9: 36338318-36338318 on Build GRCh38
rs78254954 Chr.9: 36338312-36338312 on Build GRCh38
rs528936483 Chr.9: 36338457-36338457 on Build GRCh38
rs762039644 Chr.9: 36338168-36338168 on Build GRCh38

View Full Product Details