Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48814583-48815087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC6
Entrez Gene ID
10013
Gene Name
histone deacetylase 6
Gene Aliases
CPBHM, HD6, JM21, PPP1R90
UniGene
Hs.6764
Gene Chromosome Location
Chr.X: 48801377-48824982 on Build GRCh38
Transcripts
NM_001321225.1 NM_001321226.1 NM_001321227.1 NM_001321228.1 NM_001321229.1 NM_006044.3 NR_135591.1 NR_135592.1 XM_017029187.1

SNP Detail

SNP ID Chromosome Location
rs782509715 Chr.X: 48814915-48814915 on Build GRCh38
rs782701334 Chr.X: 48815013-48815013 on Build GRCh38
rs782105383 Chr.X: 48814800-48814800 on Build GRCh38
rs782586891 Chr.X: 48814632-48814632 on Build GRCh38
rs782341889 Chr.X: 48814929-48814929 on Build GRCh38
rs1035017784 Chr.X: 48814617-48814617 on Build GRCh38
rs370628443 Chr.X: 48814952-48814952 on Build GRCh38
rs781836327 Chr.X: 48814717-48814717 on Build GRCh38
rs782735977 Chr.X: 48814811-48814811 on Build GRCh38
rs782637538 Chr.X: 48814917-48814917 on Build GRCh38
rs182847687 Chr.X: 48814699-48814699 on Build GRCh38
rs781914689 Chr.X: 48814732-48814732 on Build GRCh38
rs369700028 Chr.X: 48814716-48814716 on Build GRCh38
rs782037409 Chr.X: 48815019-48815019 on Build GRCh38
rs782438091 Chr.X: 48814652-48814652 on Build GRCh38
rs782176475 Chr.X: 48814636-48814636 on Build GRCh38
rs781847242 Chr.X: 48814627-48814627 on Build GRCh38
rs782240476 Chr.X: 48814607-48814607 on Build GRCh38
rs782116929 Chr.X: 48814603-48814603 on Build GRCh38
rs782431015 Chr.X: 48814824-48814824 on Build GRCh38
rs147652236 Chr.X: 48814749-48814749 on Build GRCh38
rs782795837 Chr.X: 48815033-48815033 on Build GRCh38
rs781996305 Chr.X: 48814602-48814602 on Build GRCh38
rs943020305 Chr.X: 48814991-48814991 on Build GRCh38
rs782119212 Chr.X: 48814987-48814987 on Build GRCh38
rs146634571 Chr.X: 48814698-48814698 on Build GRCh38
rs782365359 Chr.X: 48814728-48814728 on Build GRCh38
rs782358466 Chr.X: 48814988-48814988 on Build GRCh38
rs202088206 Chr.X: 48815036-48815036 on Build GRCh38
rs141467577 Chr.X: 48814982-48814982 on Build GRCh38
rs781858924 Chr.X: 48814799-48814799 on Build GRCh38
rs782745280 Chr.X: 48814605-48814605 on Build GRCh38
rs782660849 Chr.X: 48814653-48814653 on Build GRCh38
rs377605184 Chr.X: 48814899-48814899 on Build GRCh38
rs782638509 Chr.X: 48814624-48814624 on Build GRCh38
rs782479985 Chr.X: 48814630-48814630 on Build GRCh38
rs145223784 Chr.X: 48815023-48815023 on Build GRCh38
rs374583816 Chr.X: 48814992-48814992 on Build GRCh38
rs781958232 Chr.X: 48814764-48814765 on Build GRCh38
rs782798063 Chr.X: 48814794-48814794 on Build GRCh38
rs782685934 Chr.X: 48814611-48814611 on Build GRCh38
rs191082746 Chr.X: 48814609-48814609 on Build GRCh38
rs782760207 Chr.X: 48814927-48814927 on Build GRCh38
rs782274867 Chr.X: 48815016-48815016 on Build GRCh38
rs781897644 Chr.X: 48814614-48814614 on Build GRCh38
rs782318981 Chr.X: 48814731-48814731 on Build GRCh38
rs782466350 Chr.X: 48814914-48814914 on Build GRCh38
rs782439377 Chr.X: 48814860-48814860 on Build GRCh38
rs782415213 Chr.X: 48814973-48814973 on Build GRCh38
rs782175804 Chr.X: 48814969-48814969 on Build GRCh38
rs782074452 Chr.X: 48815007-48815007 on Build GRCh38

View Full Product Details