Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63759953-63760455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf95
Entrez Gene ID
65998
Gene Name
chromosome 11 open reading frame 95
Gene Aliases
-
UniGene
Hs.191073
Gene Chromosome Location
Chr.11: 63759892-63768641 on Build GRCh38
Transcripts
NM_001144936.1

SNP Detail

SNP ID Chromosome Location
rs756025147 Chr.11: 63760312-63760312 on Build GRCh38
rs919976616 Chr.11: 63760235-63760235 on Build GRCh38
rs975810495 Chr.11: 63760408-63760408 on Build GRCh38
rs1056497933 Chr.11: 63760308-63760308 on Build GRCh38
rs973250012 Chr.11: 63760219-63760219 on Build GRCh38
rs537314236 Chr.11: 63760155-63760155 on Build GRCh38
rs905397880 Chr.11: 63760291-63760291 on Build GRCh38
rs537728509 Chr.11: 63760241-63760241 on Build GRCh38
rs774776509 Chr.11: 63760141-63760141 on Build GRCh38
rs767009856 Chr.11: 63760258-63760258 on Build GRCh38
rs1012998138 Chr.11: 63760424-63760424 on Build GRCh38
rs947529208 Chr.11: 63760353-63760353 on Build GRCh38
rs138682928 Chr.11: 63760068-63760069 on Build GRCh38
rs1043193972 Chr.11: 63760356-63760356 on Build GRCh38
rs903387396 Chr.11: 63760370-63760370 on Build GRCh38
rs1017431321 Chr.11: 63760386-63760386 on Build GRCh38
rs866169524 Chr.11: 63760363-63760363 on Build GRCh38
rs998560536 Chr.11: 63760376-63760376 on Build GRCh38
rs773995014 Chr.11: 63760247-63760247 on Build GRCh38
rs879763232 Chr.11: 63760161-63760161 on Build GRCh38
rs552639603 Chr.11: 63760042-63760042 on Build GRCh38
rs896466886 Chr.11: 63760309-63760309 on Build GRCh38
rs548450035 Chr.11: 63760108-63760108 on Build GRCh38
rs963201948 Chr.11: 63760207-63760207 on Build GRCh38
rs185282982 Chr.11: 63760249-63760249 on Build GRCh38
rs554417143 Chr.11: 63760332-63760334 on Build GRCh38
rs188412065 Chr.11: 63760421-63760421 on Build GRCh38
rs1020889671 Chr.11: 63760071-63760071 on Build GRCh38
rs1035885219 Chr.11: 63760387-63760387 on Build GRCh38
rs528635684 Chr.11: 63760100-63760100 on Build GRCh38
rs749993667 Chr.11: 63760283-63760283 on Build GRCh38
rs1001231904 Chr.11: 63760371-63760371 on Build GRCh38
rs568649690 Chr.11: 63760134-63760134 on Build GRCh38
rs566169221 Chr.11: 63760069-63760069 on Build GRCh38
rs1047319650 Chr.11: 63760245-63760245 on Build GRCh38
rs900102691 Chr.11: 63760334-63760334 on Build GRCh38
rs181433357 Chr.11: 63760021-63760021 on Build GRCh38
rs547493938 Chr.11: 63760305-63760305 on Build GRCh38
rs557653746 Chr.11: 63759989-63759996 on Build GRCh38
rs964456661 Chr.11: 63760198-63760198 on Build GRCh38
rs539035732 Chr.11: 63760257-63760257 on Build GRCh38
rs964567192 Chr.11: 63760388-63760388 on Build GRCh38
rs149647103 Chr.11: 63760025-63760025 on Build GRCh38
rs960724913 Chr.11: 63760044-63760044 on Build GRCh38
rs941491673 Chr.11: 63760265-63760265 on Build GRCh38
rs552593472 Chr.11: 63760295-63760295 on Build GRCh38
rs960016998 Chr.11: 63760154-63760154 on Build GRCh38
rs557207853 Chr.11: 63760201-63760201 on Build GRCh38

View Full Product Details