Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75381031-75381607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A3BP
Entrez Gene ID
10087
Gene Name
collagen type IV alpha 3 binding protein
Gene Aliases
CERT, CERTL, GPBP, MRD34, STARD11
UniGene
Hs.270437
Gene Chromosome Location
Chr.5: 75368486-75511981 on Build GRCh38
Transcripts
NM_001130105.1 NM_005713.2 NM_031361.2 XM_006714513.2 XM_011543090.2 XM_011543091.2 XM_017008919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6633246 c.1720C>A p.L574I Chr.5: 75381099-75381099 on Build GRCh38
4654004 c.1657A>G p.M553V Chr.5: 75381162-75381162 on Build GRCh38
3828426 c.1628G>A p.R543Q Chr.5: 75381191-75381191 on Build GRCh38
739239 c.1735T>A p.Y579N Chr.5: 75381084-75381084 on Build GRCh38
3828425 c.2012G>A p.R671Q Chr.5: 75381191-75381191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977373842 Chr.5: 75381381-75381381 on Build GRCh38
rs967621054 Chr.5: 75381371-75381371 on Build GRCh38
rs566822672 Chr.5: 75381142-75381142 on Build GRCh38
rs756579529 Chr.5: 75381226-75381226 on Build GRCh38
rs968549617 Chr.5: 75381561-75381561 on Build GRCh38
rs781623182 Chr.5: 75381068-75381068 on Build GRCh38
rs997036918 Chr.5: 75381267-75381267 on Build GRCh38
rs143076351 Chr.5: 75381178-75381178 on Build GRCh38
rs773229774 Chr.5: 75381123-75381123 on Build GRCh38
rs762397975 Chr.5: 75381182-75381182 on Build GRCh38
rs955867882 Chr.5: 75381461-75381461 on Build GRCh38
rs369711101 Chr.5: 75381236-75381236 on Build GRCh38
rs764836042 Chr.5: 75381169-75381169 on Build GRCh38
rs1046761564 Chr.5: 75381359-75381359 on Build GRCh38
rs1056322049 Chr.5: 75381443-75381443 on Build GRCh38
rs898205438 Chr.5: 75381578-75381578 on Build GRCh38
rs956657577 Chr.5: 75381516-75381516 on Build GRCh38
rs749537709 Chr.5: 75381230-75381230 on Build GRCh38
rs936110598 Chr.5: 75381131-75381131 on Build GRCh38
rs759295755 Chr.5: 75381148-75381148 on Build GRCh38
rs766212483 Chr.5: 75381328-75381328 on Build GRCh38
rs35875083 Chr.5: 75381112-75381112 on Build GRCh38
rs936092257 Chr.5: 75381423-75381423 on Build GRCh38
rs139588122 Chr.5: 75381238-75381238 on Build GRCh38
rs756017060 Chr.5: 75381285-75381285 on Build GRCh38
rs200370086 Chr.5: 75381139-75381139 on Build GRCh38
rs767958451 Chr.5: 75381218-75381218 on Build GRCh38
rs780377096 Chr.5: 75381229-75381229 on Build GRCh38
rs1005357254 Chr.5: 75381378-75381378 on Build GRCh38
rs1018150225 Chr.5: 75381061-75381061 on Build GRCh38
rs989077749 Chr.5: 75381464-75381464 on Build GRCh38
rs963935705 Chr.5: 75381400-75381400 on Build GRCh38
rs755044895 Chr.5: 75381232-75381232 on Build GRCh38
rs746276341 Chr.5: 75381076-75381076 on Build GRCh38
rs751943626 Chr.5: 75381191-75381191 on Build GRCh38
rs749342048 Chr.5: 75381110-75381110 on Build GRCh38
rs926864147 Chr.5: 75381254-75381254 on Build GRCh38
rs544659844 Chr.5: 75381540-75381540 on Build GRCh38
rs118092017 Chr.5: 75381399-75381399 on Build GRCh38
rs1056663478 Chr.5: 75381289-75381289 on Build GRCh38
rs528126200 Chr.5: 75381514-75381514 on Build GRCh38
rs745633445 Chr.5: 75381249-75381249 on Build GRCh38
rs763605468 Chr.5: 75381187-75381187 on Build GRCh38
rs776469839 Chr.5: 75381245-75381245 on Build GRCh38
rs750431443 Chr.5: 75381270-75381270 on Build GRCh38
rs1045610259 Chr.5: 75381484-75381484 on Build GRCh38
rs915304446 Chr.5: 75381541-75381541 on Build GRCh38
rs559103921 Chr.5: 75381468-75381468 on Build GRCh38
rs750770297 Chr.5: 75381225-75381225 on Build GRCh38
rs998100584 Chr.5: 75381467-75381467 on Build GRCh38
rs544444733 Chr.5: 75381303-75381303 on Build GRCh38
rs939590784 Chr.5: 75381269-75381269 on Build GRCh38
rs78587954 Chr.5: 75381345-75381345 on Build GRCh38
rs912115806 Chr.5: 75381365-75381365 on Build GRCh38
rs757777770 Chr.5: 75381199-75381199 on Build GRCh38
rs962443116 Chr.5: 75381260-75381260 on Build GRCh38
rs888212352 Chr.5: 75381369-75381369 on Build GRCh38
rs780465149 Chr.5: 75381099-75381099 on Build GRCh38
rs138391738 Chr.5: 75381133-75381133 on Build GRCh38
rs141459538 Chr.5: 75381097-75381097 on Build GRCh38
rs1046026155 Chr.5: 75381227-75381227 on Build GRCh38
rs753200250 Chr.5: 75381235-75381235 on Build GRCh38
rs761229265 Chr.5: 75381113-75381113 on Build GRCh38
rs1035455924 Chr.5: 75381453-75381453 on Build GRCh38
rs779062129 Chr.5: 75381525-75381525 on Build GRCh38
rs769597641 Chr.5: 75381250-75381250 on Build GRCh38
rs376008922 Chr.5: 75381233-75381233 on Build GRCh38
rs190893261 Chr.5: 75381170-75381170 on Build GRCh38
rs762493715 Chr.5: 75381218-75381219 on Build GRCh38
rs1029618830 Chr.5: 75381501-75381501 on Build GRCh38
rs183271652 Chr.5: 75381322-75381322 on Build GRCh38
rs767045597 Chr.5: 75381149-75381149 on Build GRCh38

View Full Product Details