Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48759213-48759718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL4
Entrez Gene ID
84871
Gene Name
ATP/GTP binding protein like 4
Gene Aliases
CCP6
UniGene
Hs.551844
Gene Chromosome Location
Chr.1: 48520157-50024011 on Build GRCh38
Transcripts
NM_001323573.1 NM_001323574.1 NM_001323575.1 NM_032785.3 NR_136623.1 XM_011542308.2 XM_017002595.1 XM_017002596.1 XM_017002597.1 NM_001302082.1 NM_024603.3 XM_011542141.2 XM_011542142.2 XM_017002331.1 XM_017002332.1 XM_017002333.1 XM_017002334.1

Gene Symbol
BEND5
Entrez Gene ID
79656
Gene Name
BEN domain containing 5
Gene Aliases
C1orf165
UniGene
Hs.475348
Gene Chromosome Location
Chr.1: 48727523-48778603 on Build GRCh38
Transcripts
NM_001323573.1 NM_001323574.1 NM_001323575.1 NM_032785.3 NR_136623.1 XM_011542308.2 XM_017002595.1 XM_017002596.1 XM_017002597.1 NM_001302082.1 NM_024603.3 XM_011542141.2 XM_011542142.2 XM_017002331.1 XM_017002332.1 XM_017002333.1 XM_017002334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6200258 c.386A>C p.E129A Chr.1: 48759259-48759259 on Build GRCh38
6841198 c.374G>A p.R125Q Chr.1: 48759271-48759271 on Build GRCh38
5629643 c.364C>A p.P122T Chr.1: 48759281-48759281 on Build GRCh38
6191816 c.402C>T p.S134S Chr.1: 48759243-48759243 on Build GRCh38
4655028 c.399G>C p.K133N Chr.1: 48759246-48759246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569270988 Chr.1: 48759547-48759547 on Build GRCh38
rs772557186 Chr.1: 48759311-48759311 on Build GRCh38
rs896765241 Chr.1: 48759578-48759578 on Build GRCh38
rs951790778 Chr.1: 48759622-48759622 on Build GRCh38
rs747457186 Chr.1: 48759424-48759424 on Build GRCh38
rs72681063 Chr.1: 48759625-48759625 on Build GRCh38
rs1012446845 Chr.1: 48759602-48759602 on Build GRCh38
rs531139897 Chr.1: 48759298-48759298 on Build GRCh38
rs1027816419 Chr.1: 48759607-48759607 on Build GRCh38
rs748438575 Chr.1: 48759265-48759265 on Build GRCh38
rs773286114 Chr.1: 48759299-48759299 on Build GRCh38
rs549650665 Chr.1: 48759329-48759329 on Build GRCh38
rs995601818 Chr.1: 48759662-48759662 on Build GRCh38
rs749096188 Chr.1: 48759264-48759264 on Build GRCh38
rs56982079 Chr.1: 48759314-48759314 on Build GRCh38
rs912205418 Chr.1: 48759696-48759696 on Build GRCh38
rs536986032 Chr.1: 48759621-48759621 on Build GRCh38
rs76381601 Chr.1: 48759416-48759416 on Build GRCh38
rs377250325 Chr.1: 48759591-48759591 on Build GRCh38
rs761016220 Chr.1: 48759307-48759307 on Build GRCh38
rs944562541 Chr.1: 48759653-48759653 on Build GRCh38
rs1890727 Chr.1: 48759689-48759689 on Build GRCh38
rs546783573 Chr.1: 48759422-48759422 on Build GRCh38
rs371832189 Chr.1: 48759407-48759407 on Build GRCh38
rs539051610 Chr.1: 48759494-48759494 on Build GRCh38
rs999551768 Chr.1: 48759539-48759539 on Build GRCh38
rs867289331 Chr.1: 48759665-48759665 on Build GRCh38
rs138475470 Chr.1: 48759663-48759663 on Build GRCh38
rs750979533 Chr.1: 48759239-48759239 on Build GRCh38
rs755957106 Chr.1: 48759255-48759255 on Build GRCh38
rs182601888 Chr.1: 48759632-48759632 on Build GRCh38
rs780671877 Chr.1: 48759246-48759246 on Build GRCh38
rs962471023 Chr.1: 48759253-48759253 on Build GRCh38
rs902567406 Chr.1: 48759531-48759531 on Build GRCh38
rs541190696 Chr.1: 48759643-48759643 on Build GRCh38
rs748136013 Chr.1: 48759271-48759271 on Build GRCh38
rs557411763 Chr.1: 48759252-48759252 on Build GRCh38
rs1023747124 Chr.1: 48759247-48759247 on Build GRCh38
rs951186189 Chr.1: 48759664-48759664 on Build GRCh38
rs776914684 Chr.1: 48759315-48759315 on Build GRCh38
rs912503662 Chr.1: 48759349-48759349 on Build GRCh38
rs759911695 Chr.1: 48759318-48759318 on Build GRCh38
rs780057963 Chr.1: 48759261-48759261 on Build GRCh38
rs56960839 Chr.1: 48759313-48759313 on Build GRCh38
rs942599703 Chr.1: 48759406-48759406 on Build GRCh38
rs771432898 Chr.1: 48759530-48759530 on Build GRCh38
rs557426335 Chr.1: 48759527-48759527 on Build GRCh38
rs1023802545 Chr.1: 48759605-48759605 on Build GRCh38
rs571743140 Chr.1: 48759481-48759481 on Build GRCh38
rs910358602 Chr.1: 48759647-48759647 on Build GRCh38
rs1043842770 Chr.1: 48759482-48759482 on Build GRCh38

View Full Product Details