Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20933701-20934212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRKL
Entrez Gene ID
1399
Gene Name
CRK like proto-oncogene, adaptor protein
Gene Aliases
-
UniGene
Hs.5613
Gene Chromosome Location
Chr.22: 20917398-20953749 on Build GRCh38
Transcripts
NM_005207.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6977060 c.433delA p.K146fs*5 Chr.22: 20933900-20933900 on Build GRCh38
5858976 c.375delG p.E126fs*25 Chr.22: 20933842-20933842 on Build GRCh38
3552347 c.467C>T p.P156L Chr.22: 20933934-20933934 on Build GRCh38
210955 c.440G>T p.G147V Chr.22: 20933907-20933907 on Build GRCh38
6951600 c.590G>A p.S197N Chr.22: 20934057-20934057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752340785 Chr.22: 20934118-20934118 on Build GRCh38
rs764823179 Chr.22: 20934116-20934116 on Build GRCh38
rs771313570 Chr.22: 20933755-20933755 on Build GRCh38
rs775637595 Chr.22: 20933784-20933784 on Build GRCh38
rs757836194 Chr.22: 20934026-20934026 on Build GRCh38
rs925122172 Chr.22: 20933811-20933811 on Build GRCh38
rs755863490 Chr.22: 20933789-20933789 on Build GRCh38
rs781570771 Chr.22: 20933894-20933894 on Build GRCh38
rs145813322 Chr.22: 20934128-20934128 on Build GRCh38
rs780153751 Chr.22: 20934164-20934164 on Build GRCh38
rs756451346 Chr.22: 20934144-20934144 on Build GRCh38
rs771933873 Chr.22: 20933921-20933921 on Build GRCh38
rs376691633 Chr.22: 20933813-20933813 on Build GRCh38
rs369055917 Chr.22: 20933959-20933959 on Build GRCh38
rs765164920 Chr.22: 20933775-20933775 on Build GRCh38
rs773135257 Chr.22: 20934070-20934070 on Build GRCh38
rs761713297 Chr.22: 20933786-20933786 on Build GRCh38
rs550457678 Chr.22: 20933920-20933920 on Build GRCh38
rs762375550 Chr.22: 20934120-20934120 on Build GRCh38
rs758872714 Chr.22: 20933847-20933847 on Build GRCh38
rs776452921 Chr.22: 20934088-20934088 on Build GRCh38
rs778350864 Chr.22: 20933818-20933818 on Build GRCh38
rs1042313936 Chr.22: 20933751-20933751 on Build GRCh38
rs767466740 Chr.22: 20933788-20933788 on Build GRCh38
rs535421669 Chr.22: 20933999-20933999 on Build GRCh38
rs200712124 Chr.22: 20933958-20933958 on Build GRCh38
rs752182830 Chr.22: 20934001-20934001 on Build GRCh38
rs147576993 Chr.22: 20934023-20934023 on Build GRCh38
rs778388431 Chr.22: 20933877-20933877 on Build GRCh38
rs775479004 Chr.22: 20933905-20933905 on Build GRCh38
rs760083004 Chr.22: 20934108-20934110 on Build GRCh38
rs748247058 Chr.22: 20934054-20934054 on Build GRCh38
rs749349384 Chr.22: 20934056-20934056 on Build GRCh38
rs771364672 Chr.22: 20933890-20933890 on Build GRCh38
rs952900119 Chr.22: 20934009-20934009 on Build GRCh38
rs200629140 Chr.22: 20934074-20934074 on Build GRCh38
rs770045446 Chr.22: 20933900-20933900 on Build GRCh38
rs199907840 Chr.22: 20934036-20934036 on Build GRCh38
rs776944535 Chr.22: 20933757-20933757 on Build GRCh38
rs377625090 Chr.22: 20934028-20934028 on Build GRCh38
rs976691655 Chr.22: 20934081-20934081 on Build GRCh38
rs897384727 Chr.22: 20933962-20933962 on Build GRCh38
rs148491827 Chr.22: 20933831-20933831 on Build GRCh38
rs780282215 Chr.22: 20934055-20934055 on Build GRCh38
rs141971963 Chr.22: 20934061-20934061 on Build GRCh38
rs768104683 Chr.22: 20934132-20934132 on Build GRCh38
rs894895904 Chr.22: 20933893-20933893 on Build GRCh38
rs183244835 Chr.22: 20933956-20933956 on Build GRCh38
rs746144085 Chr.22: 20933896-20933896 on Build GRCh38
rs757880216 Chr.22: 20934013-20934013 on Build GRCh38
rs772975472 Chr.22: 20933742-20933743 on Build GRCh38
rs936540793 Chr.22: 20933817-20933817 on Build GRCh38
rs377262969 Chr.22: 20933731-20933731 on Build GRCh38
rs200265850 Chr.22: 20933795-20933795 on Build GRCh38
rs770839945 Chr.22: 20934077-20934077 on Build GRCh38
rs759338529 Chr.22: 20934091-20934091 on Build GRCh38
rs373785511 Chr.22: 20933973-20933973 on Build GRCh38
rs753427288 Chr.22: 20933808-20933808 on Build GRCh38
rs759703632 Chr.22: 20933762-20933762 on Build GRCh38
rs747350029 Chr.22: 20933743-20933743 on Build GRCh38
rs936484318 Chr.22: 20934166-20934166 on Build GRCh38
rs763765173 Chr.22: 20934127-20934127 on Build GRCh38
rs750783216 Chr.22: 20934140-20934140 on Build GRCh38
rs757890975 Chr.22: 20934129-20934129 on Build GRCh38
rs746219908 Chr.22: 20934029-20934029 on Build GRCh38
rs759166693 Chr.22: 20933990-20933990 on Build GRCh38
rs747404847 Chr.22: 20933883-20933883 on Build GRCh38
rs769948686 Chr.22: 20934046-20934046 on Build GRCh38
rs749456958 Chr.22: 20934186-20934186 on Build GRCh38
rs760471921 Chr.22: 20933957-20933957 on Build GRCh38
rs945307538 Chr.22: 20933732-20933732 on Build GRCh38
rs755185083 Chr.22: 20934187-20934187 on Build GRCh38
rs3180410 Chr.22: 20933816-20933816 on Build GRCh38
rs373069327 Chr.22: 20933739-20933739 on Build GRCh38
rs555439236 Chr.22: 20934019-20934019 on Build GRCh38
rs145156356 Chr.22: 20934022-20934022 on Build GRCh38
rs752232310 Chr.22: 20933832-20933832 on Build GRCh38

View Full Product Details