Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50217434-50217944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SELO
Entrez Gene ID
83642
Gene Name
selenoprotein O
Gene Aliases
-
UniGene
Hs.365405
Gene Chromosome Location
Chr.22: 50200979-50217616 on Build GRCh38
Transcripts
NM_031454.1 NM_020461.3

Gene Symbol
TUBGCP6
Entrez Gene ID
85378
Gene Name
tubulin gamma complex associated protein 6
Gene Aliases
GCP-6, GCP6, MCCRP, MCCRP1, MCPHCR
UniGene
Hs.336431
Gene Chromosome Location
Chr.22: 50217689-50245024 on Build GRCh38
Transcripts
NM_031454.1 NM_020461.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4755022 c.5425C>T p.R1809C Chr.22: 50217771-50217771 on Build GRCh38
3390298 c.5351C>T p.S1784F Chr.22: 50217935-50217935 on Build GRCh38
4105053 c.5454C>T p.D1818D Chr.22: 50217742-50217742 on Build GRCh38
4882673 c.5422C>A p.L1808M Chr.22: 50217774-50217774 on Build GRCh38
1751937 c.5398C>T p.Q1800* Chr.22: 50217798-50217798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775181111 Chr.22: 50217704-50217704 on Build GRCh38
rs115016508 Chr.22: 50217614-50217614 on Build GRCh38
rs915728440 Chr.22: 50217610-50217610 on Build GRCh38
rs925320821 Chr.22: 50217627-50217627 on Build GRCh38
rs900169100 Chr.22: 50217607-50217607 on Build GRCh38
rs150137904 Chr.22: 50217804-50217804 on Build GRCh38
rs751685809 Chr.22: 50217769-50217769 on Build GRCh38
rs754731247 Chr.22: 50217881-50217882 on Build GRCh38
rs556653451 Chr.22: 50217696-50217696 on Build GRCh38
rs761399858 Chr.22: 50217826-50217826 on Build GRCh38
rs765823423 Chr.22: 50217761-50217761 on Build GRCh38
rs534644764 Chr.22: 50217666-50217666 on Build GRCh38
rs758336111 Chr.22: 50217865-50217865 on Build GRCh38
rs746804508 Chr.22: 50217871-50217871 on Build GRCh38
rs770493065 Chr.22: 50217593-50217593 on Build GRCh38
rs907614304 Chr.22: 50217824-50217824 on Build GRCh38
rs754776705 Chr.22: 50217906-50217906 on Build GRCh38
rs995659853 Chr.22: 50217569-50217569 on Build GRCh38
rs992844769 Chr.22: 50217665-50217665 on Build GRCh38
rs533244082 Chr.22: 50217644-50217644 on Build GRCh38
rs781068597 Chr.22: 50217772-50217772 on Build GRCh38
rs529266243 Chr.22: 50217619-50217619 on Build GRCh38
rs763708813 Chr.22: 50217706-50217706 on Build GRCh38
rs759122979 Chr.22: 50217699-50217699 on Build GRCh38
rs931491100 Chr.22: 50217849-50217849 on Build GRCh38
rs552589056 Chr.22: 50217504-50217504 on Build GRCh38
rs370024869 Chr.22: 50217740-50217740 on Build GRCh38
rs387907019 Chr.22: 50217738-50217738 on Build GRCh38
rs768399908 Chr.22: 50217874-50217874 on Build GRCh38
rs141707083 Chr.22: 50217745-50217745 on Build GRCh38
rs750319436 Chr.22: 50217861-50217861 on Build GRCh38
rs540920396 Chr.22: 50217608-50217608 on Build GRCh38
rs1039909311 Chr.22: 50217650-50217650 on Build GRCh38
rs991257148 Chr.22: 50217488-50217488 on Build GRCh38
rs748711979 Chr.22: 50217816-50217816 on Build GRCh38
rs766814239 Chr.22: 50217848-50217848 on Build GRCh38
rs373541807 Chr.22: 50217794-50217794 on Build GRCh38
rs1052834 Chr.22: 50217468-50217468 on Build GRCh38
rs946253122 Chr.22: 50217475-50217475 on Build GRCh38
rs960040324 Chr.22: 50217661-50217662 on Build GRCh38
rs371551733 Chr.22: 50217889-50217889 on Build GRCh38
rs1004741727 Chr.22: 50217639-50217639 on Build GRCh38
rs148008115 Chr.22: 50217741-50217741 on Build GRCh38
rs983742489 Chr.22: 50217517-50217517 on Build GRCh38
rs374034625 Chr.22: 50217851-50217851 on Build GRCh38
rs549275810 Chr.22: 50217485-50217485 on Build GRCh38
rs534027251 Chr.22: 50217732-50217732 on Build GRCh38
rs201070582 Chr.22: 50217888-50217888 on Build GRCh38
rs540957245 Chr.22: 50217770-50217770 on Build GRCh38
rs1000556465 Chr.22: 50217487-50217487 on Build GRCh38
rs371919513 Chr.22: 50217691-50217691 on Build GRCh38
rs373083386 Chr.22: 50217883-50217883 on Build GRCh38
rs55955211 Chr.22: 50217680-50217680 on Build GRCh38
rs371217966 Chr.22: 50217720-50217720 on Build GRCh38
rs759423934 Chr.22: 50217642-50217642 on Build GRCh38
rs747299890 Chr.22: 50217721-50217721 on Build GRCh38
rs956764514 Chr.22: 50217748-50217748 on Build GRCh38
rs767171403 Chr.22: 50217700-50217700 on Build GRCh38
rs766902364 Chr.22: 50217768-50217768 on Build GRCh38
rs747848123 Chr.22: 50217882-50217882 on Build GRCh38
rs752717976 Chr.22: 50217653-50217653 on Build GRCh38
rs776201082 Chr.22: 50217823-50217823 on Build GRCh38
rs766334237 Chr.22: 50217904-50217904 on Build GRCh38
rs762167154 Chr.22: 50217756-50217756 on Build GRCh38
rs760315093 Chr.22: 50217548-50217548 on Build GRCh38
rs375852691 Chr.22: 50217692-50217692 on Build GRCh38
rs766213817 Chr.22: 50217833-50217834 on Build GRCh38
rs143516483 Chr.22: 50217551-50217551 on Build GRCh38
rs113121446 Chr.22: 50217605-50217605 on Build GRCh38
rs376446485 Chr.22: 50217857-50217857 on Build GRCh38
rs374811995 Chr.22: 50217725-50217725 on Build GRCh38
rs773235113 Chr.22: 50217746-50217748 on Build GRCh38
rs913357304 Chr.22: 50217576-50217576 on Build GRCh38
rs760483595 Chr.22: 50217753-50217754 on Build GRCh38
rs1052692424 Chr.22: 50217533-50217533 on Build GRCh38
rs2281117 Chr.22: 50217643-50217643 on Build GRCh38
rs982655920 Chr.22: 50217471-50217471 on Build GRCh38
rs779389082 Chr.22: 50217806-50217806 on Build GRCh38
rs576751322 Chr.22: 50217489-50217489 on Build GRCh38
rs1040261734 Chr.22: 50217561-50217561 on Build GRCh38
rs574088170 Chr.22: 50217505-50217505 on Build GRCh38
rs567956645 Chr.22: 50217679-50217679 on Build GRCh38
rs202182732 Chr.22: 50217840-50217840 on Build GRCh38
rs1026682683 Chr.22: 50217573-50217573 on Build GRCh38
rs753624847 Chr.22: 50217880-50217880 on Build GRCh38
rs749300601 Chr.22: 50217688-50217688 on Build GRCh38
rs925373533 Chr.22: 50217731-50217731 on Build GRCh38
rs767767513 Chr.22: 50217853-50217853 on Build GRCh38
rs944252084 Chr.22: 50217647-50217647 on Build GRCh38
rs904798452 Chr.22: 50217594-50217594 on Build GRCh38
rs753927243 Chr.22: 50217897-50217897 on Build GRCh38
rs756245644 Chr.22: 50217790-50217790 on Build GRCh38
rs562694971 Chr.22: 50217839-50217839 on Build GRCh38
rs760396776 Chr.22: 50217742-50217742 on Build GRCh38
rs760917530 Chr.22: 50217890-50217890 on Build GRCh38
rs9628305 Chr.22: 50217697-50217697 on Build GRCh38
rs755376931 Chr.22: 50217786-50217786 on Build GRCh38
rs752837357 Chr.22: 50217778-50217778 on Build GRCh38
rs199976800 Chr.22: 50217818-50217818 on Build GRCh38
rs772726137 Chr.22: 50217547-50217547 on Build GRCh38
rs368966472 Chr.22: 50217846-50217846 on Build GRCh38
rs189757808 Chr.22: 50217763-50217763 on Build GRCh38
rs569779421 Chr.22: 50217456-50217456 on Build GRCh38
rs12165978 Chr.22: 50217789-50217789 on Build GRCh38
rs951059488 Chr.22: 50217588-50217588 on Build GRCh38
rs959988313 Chr.22: 50217681-50217681 on Build GRCh38
rs552354730 Chr.22: 50217661-50217661 on Build GRCh38
rs771804706 Chr.22: 50217805-50217805 on Build GRCh38
rs1048620967 Chr.22: 50217673-50217673 on Build GRCh38
rs537237422 Chr.22: 50217465-50217465 on Build GRCh38
rs757812432 Chr.22: 50217714-50217714 on Build GRCh38
rs570301459 Chr.22: 50217662-50217662 on Build GRCh38
rs747766416 Chr.22: 50217913-50217913 on Build GRCh38
rs138609686 Chr.22: 50217807-50217807 on Build GRCh38
rs761975484 Chr.22: 50217664-50217664 on Build GRCh38
rs1051474238 Chr.22: 50217821-50217821 on Build GRCh38
rs768366546 Chr.22: 50217808-50217808 on Build GRCh38
rs772468910 Chr.22: 50217875-50217875 on Build GRCh38
rs1016199370 Chr.22: 50217531-50217531 on Build GRCh38
rs2038050 Chr.22: 50217667-50217667 on Build GRCh38
rs2281116 Chr.22: 50217624-50217624 on Build GRCh38
rs534562303 Chr.22: 50217490-50217490 on Build GRCh38
rs765629153 Chr.22: 50217717-50217717 on Build GRCh38
rs886247158 Chr.22: 50217630-50217630 on Build GRCh38
rs754995187 Chr.22: 50217724-50217724 on Build GRCh38
rs200537421 Chr.22: 50217893-50217893 on Build GRCh38
rs528804463 Chr.22: 50217606-50217606 on Build GRCh38
rs777852464 Chr.22: 50217860-50217860 on Build GRCh38
rs899602863 Chr.22: 50217564-50217564 on Build GRCh38
rs530316963 Chr.22: 50217685-50217685 on Build GRCh38
rs750475834 Chr.22: 50217674-50217674 on Build GRCh38
rs779654243 Chr.22: 50217866-50217866 on Build GRCh38
rs776207916 Chr.22: 50217750-50217750 on Build GRCh38
rs556883073 Chr.22: 50217697-50217697 on Build GRCh38
rs200702993 Chr.22: 50217718-50217718 on Build GRCh38
rs544619554 Chr.22: 50217833-50217833 on Build GRCh38
rs938807649 Chr.22: 50217572-50217572 on Build GRCh38
rs774075764 Chr.22: 50217730-50217730 on Build GRCh38
rs759621081 Chr.22: 50217852-50217852 on Build GRCh38
rs768009656 Chr.22: 50217744-50217744 on Build GRCh38
rs936310779 Chr.22: 50217604-50217604 on Build GRCh38
rs1016582580 Chr.22: 50217641-50217641 on Build GRCh38
rs780874726 Chr.22: 50217909-50217909 on Build GRCh38
rs756155879 Chr.22: 50217858-50217858 on Build GRCh38
rs928520135 Chr.22: 50217481-50217481 on Build GRCh38
rs754248493 Chr.22: 50217710-50217710 on Build GRCh38
rs1027046348 Chr.22: 50217457-50217457 on Build GRCh38
rs373569549 Chr.22: 50217532-50217532 on Build GRCh38
rs754181880 Chr.22: 50217762-50217762 on Build GRCh38
rs886834775 Chr.22: 50217678-50217678 on Build GRCh38
rs541585456 Chr.22: 50217523-50217523 on Build GRCh38
rs749749382 Chr.22: 50217884-50217884 on Build GRCh38
rs757383120 Chr.22: 50217903-50217903 on Build GRCh38
rs1030128313 Chr.22: 50217509-50217509 on Build GRCh38
rs563380111 Chr.22: 50217524-50217524 on Build GRCh38
rs751487093 Chr.22: 50217905-50217905 on Build GRCh38
rs950586734 Chr.22: 50217511-50217511 on Build GRCh38
rs760072668 Chr.22: 50217705-50217705 on Build GRCh38

View Full Product Details