Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76310465-76310977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM47E-STBD1
Entrez Gene ID
100631383
Gene Name
FAM47E-STBD1 readthrough
Gene Aliases
-
UniGene
Hs.720509
Gene Chromosome Location
Chr.4: 76251700-76311130 on Build GRCh38
Transcripts
NM_001242939.1 NM_003943.4

Gene Symbol
STBD1
Entrez Gene ID
8987
Gene Name
starch binding domain 1
Gene Aliases
GENEX3414, GENX-3414
UniGene
Hs.109590
Gene Chromosome Location
Chr.4: 76306026-76311130 on Build GRCh38
Transcripts
NM_001242939.1 NM_003943.4

SNP Detail

SNP ID Chromosome Location
rs949789273 Chr.4: 76310906-76310908 on Build GRCh38
rs144849069 Chr.4: 76310768-76310768 on Build GRCh38
rs993721129 Chr.4: 76310580-76310580 on Build GRCh38
rs181717881 Chr.4: 76310525-76310525 on Build GRCh38
rs976863519 Chr.4: 76310730-76310730 on Build GRCh38
rs914397435 Chr.4: 76310675-76310675 on Build GRCh38
rs13680 Chr.4: 76310890-76310890 on Build GRCh38
rs1019388912 Chr.4: 76310620-76310620 on Build GRCh38
rs138652084 Chr.4: 76310885-76310885 on Build GRCh38
rs778994439 Chr.4: 76310760-76310760 on Build GRCh38
rs577118529 Chr.4: 76310524-76310524 on Build GRCh38
rs948815795 Chr.4: 76310804-76310804 on Build GRCh38
rs7671037 Chr.4: 76310506-76310506 on Build GRCh38
rs548293824 Chr.4: 76310925-76310925 on Build GRCh38
rs1018305528 Chr.4: 76310695-76310695 on Build GRCh38
rs777688704 Chr.4: 76310808-76310808 on Build GRCh38
rs541939055 Chr.4: 76310629-76310629 on Build GRCh38
rs533747185 Chr.4: 76310927-76310927 on Build GRCh38
rs751170882 Chr.4: 76310621-76310621 on Build GRCh38
rs770738020 Chr.4: 76310916-76310916 on Build GRCh38
rs754819803 Chr.4: 76310753-76310753 on Build GRCh38
rs1037759198 Chr.4: 76310841-76310841 on Build GRCh38
rs975581875 Chr.4: 76310951-76310951 on Build GRCh38
rs568204620 Chr.4: 76310926-76310926 on Build GRCh38
rs77487079 Chr.4: 76310935-76310935 on Build GRCh38
rs1026496983 Chr.4: 76310581-76310581 on Build GRCh38
rs750705231 Chr.4: 76310566-76310566 on Build GRCh38
rs1048675615 Chr.4: 76310494-76310494 on Build GRCh38
rs576092074 Chr.4: 76310592-76310592 on Build GRCh38
rs527862088 Chr.4: 76310735-76310735 on Build GRCh38
rs965435476 Chr.4: 76310721-76310721 on Build GRCh38
rs533688978 Chr.4: 76310838-76310838 on Build GRCh38
rs758515763 Chr.4: 76310638-76310638 on Build GRCh38
rs558044061 Chr.4: 76310953-76310953 on Build GRCh38
rs1019697388 Chr.4: 76310933-76310933 on Build GRCh38
rs951071120 Chr.4: 76310594-76310594 on Build GRCh38
rs1057439243 Chr.4: 76310738-76310738 on Build GRCh38
rs148864645 Chr.4: 76310909-76310909 on Build GRCh38
rs1055878417 Chr.4: 76310535-76310535 on Build GRCh38
rs140532969 Chr.4: 76310650-76310650 on Build GRCh38
rs556527746 Chr.4: 76310557-76310557 on Build GRCh38
rs1032968740 Chr.4: 76310676-76310676 on Build GRCh38
rs780245242 Chr.4: 76310658-76310658 on Build GRCh38
rs547570218 Chr.4: 76310934-76310934 on Build GRCh38
rs899355413 Chr.4: 76310871-76310871 on Build GRCh38
rs1006858568 Chr.4: 76310686-76310686 on Build GRCh38
rs986854587 Chr.4: 76310615-76310615 on Build GRCh38
rs910103106 Chr.4: 76310800-76310800 on Build GRCh38
rs768660731 Chr.4: 76310731-76310731 on Build GRCh38
rs541650284 Chr.4: 76310736-76310736 on Build GRCh38
rs369660250 Chr.4: 76310701-76310701 on Build GRCh38
rs747042936 Chr.4: 76310678-76310678 on Build GRCh38
rs767430894 Chr.4: 76310620-76310621 on Build GRCh38

View Full Product Details