Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83463095-83463612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175A
Entrez Gene ID
84142
Gene Name
family with sequence similarity 175 member A
Gene Aliases
ABRA1, CCDC98
UniGene
Hs.334772
Gene Chromosome Location
Chr.4: 83459515-83485181 on Build GRCh38
Transcripts
NM_139076.2 XM_005263280.4 XM_017008694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1495898 c.796+2T>A p.? Chr.4: 83463492-83463492 on Build GRCh38
6960600 c.698T>C p.V233A Chr.4: 83463592-83463592 on Build GRCh38
4138530 c.694A>C p.K232Q Chr.4: 83463596-83463596 on Build GRCh38
6388695 c.767A>G p.K256R Chr.4: 83463523-83463523 on Build GRCh38
5344769 c.707G>T p.S236I Chr.4: 83463583-83463583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947123468 Chr.4: 83463275-83463276 on Build GRCh38
rs756092226 Chr.4: 83463446-83463446 on Build GRCh38
rs749472899 Chr.4: 83463456-83463456 on Build GRCh38
rs747765363 Chr.4: 83463518-83463521 on Build GRCh38
rs907367049 Chr.4: 83463156-83463156 on Build GRCh38
rs758296447 Chr.4: 83463450-83463451 on Build GRCh38
rs931382192 Chr.4: 83463151-83463151 on Build GRCh38
rs549320751 Chr.4: 83463451-83463451 on Build GRCh38
rs777651096 Chr.4: 83463584-83463584 on Build GRCh38
rs544149809 Chr.4: 83463241-83463241 on Build GRCh38
rs995589963 Chr.4: 83463435-83463438 on Build GRCh38
rs1045896572 Chr.4: 83463419-83463419 on Build GRCh38
rs753968871 Chr.4: 83463558-83463558 on Build GRCh38
rs780887812 Chr.4: 83463491-83463492 on Build GRCh38
rs190760959 Chr.4: 83463457-83463457 on Build GRCh38
rs376811916 Chr.4: 83463516-83463516 on Build GRCh38
rs922428012 Chr.4: 83463307-83463307 on Build GRCh38
rs200727513 Chr.4: 83463512-83463512 on Build GRCh38
rs779038861 Chr.4: 83463570-83463570 on Build GRCh38
rs907339627 Chr.4: 83463427-83463427 on Build GRCh38
rs758079543 Chr.4: 83463581-83463581 on Build GRCh38
rs1026995604 Chr.4: 83463201-83463201 on Build GRCh38
rs962770577 Chr.4: 83463194-83463194 on Build GRCh38
rs755187051 Chr.4: 83463563-83463563 on Build GRCh38
rs748002786 Chr.4: 83463464-83463464 on Build GRCh38
rs768140721 Chr.4: 83463488-83463488 on Build GRCh38
rs778658785 Chr.4: 83463458-83463458 on Build GRCh38
rs573603631 Chr.4: 83463237-83463237 on Build GRCh38
rs898534185 Chr.4: 83463434-83463435 on Build GRCh38
rs540071037 Chr.4: 83463141-83463141 on Build GRCh38
rs186517125 Chr.4: 83463181-83463181 on Build GRCh38
rs200414461 Chr.4: 83463521-83463521 on Build GRCh38
rs372204900 Chr.4: 83463375-83463375 on Build GRCh38
rs1049954771 Chr.4: 83463204-83463204 on Build GRCh38
rs878855208 Chr.4: 83463525-83463525 on Build GRCh38
rs545362940 Chr.4: 83463350-83463350 on Build GRCh38
rs201648921 Chr.4: 83463149-83463149 on Build GRCh38
rs994088508 Chr.4: 83463454-83463454 on Build GRCh38
rs186204843 Chr.4: 83463428-83463428 on Build GRCh38
rs889965782 Chr.4: 83463218-83463218 on Build GRCh38
rs28696794 Chr.4: 83463200-83463200 on Build GRCh38
rs370034758 Chr.4: 83463477-83463477 on Build GRCh38
rs199541299 Chr.4: 83463503-83463503 on Build GRCh38
rs11099602 Chr.4: 83463259-83463259 on Build GRCh38
rs977345020 Chr.4: 83463340-83463340 on Build GRCh38
rs779972353 Chr.4: 83463468-83463468 on Build GRCh38
rs373605348 Chr.4: 83463509-83463509 on Build GRCh38
rs191027464 Chr.4: 83463345-83463345 on Build GRCh38
rs896745771 Chr.4: 83463354-83463355 on Build GRCh38
rs746885355 Chr.4: 83463585-83463585 on Build GRCh38
rs1026546660 Chr.4: 83463455-83463455 on Build GRCh38
rs746513523 Chr.4: 83463473-83463474 on Build GRCh38
rs180953179 Chr.4: 83463362-83463362 on Build GRCh38
rs984777512 Chr.4: 83463246-83463246 on Build GRCh38
rs902678476 Chr.4: 83463320-83463320 on Build GRCh38
rs894228113 Chr.4: 83463383-83463383 on Build GRCh38
rs114513239 Chr.4: 83463535-83463535 on Build GRCh38
rs770303098 Chr.4: 83463481-83463481 on Build GRCh38
rs1025894241 Chr.4: 83463203-83463203 on Build GRCh38
rs759239699 Chr.4: 83463491-83463491 on Build GRCh38
rs752929794 Chr.4: 83463575-83463575 on Build GRCh38
rs1017628547 Chr.4: 83463256-83463256 on Build GRCh38
rs1013082855 Chr.4: 83463147-83463147 on Build GRCh38
rs771903826 Chr.4: 83463476-83463476 on Build GRCh38
rs771336457 Chr.4: 83463578-83463578 on Build GRCh38
rs1006168284 Chr.4: 83463551-83463551 on Build GRCh38
rs201475497 Chr.4: 83463514-83463514 on Build GRCh38
rs924546780 Chr.4: 83463347-83463347 on Build GRCh38
rs767941812 Chr.4: 83463519-83463519 on Build GRCh38
rs776317671 Chr.4: 83463486-83463486 on Build GRCh38
rs555501219 Chr.4: 83463192-83463192 on Build GRCh38
rs948411627 Chr.4: 83463411-83463411 on Build GRCh38
rs964797236 Chr.4: 83463257-83463257 on Build GRCh38
rs369385726 Chr.4: 83463529-83463529 on Build GRCh38
rs750850603 Chr.4: 83463520-83463520 on Build GRCh38
rs371035914 Chr.4: 83463236-83463236 on Build GRCh38
rs896436402 Chr.4: 83463126-83463126 on Build GRCh38
rs933833852 Chr.4: 83463332-83463332 on Build GRCh38
rs1035315699 Chr.4: 83463121-83463121 on Build GRCh38
rs770131496 Chr.4: 83463268-83463269 on Build GRCh38

View Full Product Details