Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42897088-42897589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOOK3
Entrez Gene ID
84376
Gene Name
hook microtubule-tethering protein 3
Gene Aliases
HK3
UniGene
Hs.162852
Gene Chromosome Location
Chr.8: 42896890-43030539 on Build GRCh38
Transcripts
NM_032410.3 XM_011544666.2

Gene Symbol
RNF170
Entrez Gene ID
81790
Gene Name
ring finger protein 170
Gene Aliases
ADSA, SNAX1
UniGene
Hs.491626
Gene Chromosome Location
Chr.8: 42849637-42897288 on Build GRCh38
Transcripts
NM_032410.3 XM_011544666.2

SNP Detail

SNP ID Chromosome Location
rs901065612 Chr.8: 42897387-42897387 on Build GRCh38
rs562561846 Chr.8: 42897126-42897126 on Build GRCh38
rs1053824519 Chr.8: 42897342-42897342 on Build GRCh38
rs1019966338 Chr.8: 42897408-42897408 on Build GRCh38
rs982883875 Chr.8: 42897526-42897526 on Build GRCh38
rs554397385 Chr.8: 42897560-42897560 on Build GRCh38
rs936774909 Chr.8: 42897559-42897559 on Build GRCh38
rs952435641 Chr.8: 42897240-42897240 on Build GRCh38
rs958345233 Chr.8: 42897495-42897495 on Build GRCh38
rs978670775 Chr.8: 42897553-42897553 on Build GRCh38
rs975810975 Chr.8: 42897260-42897260 on Build GRCh38
rs575469119 Chr.8: 42897377-42897377 on Build GRCh38
rs113969303 Chr.8: 42897263-42897263 on Build GRCh38
rs983827038 Chr.8: 42897451-42897451 on Build GRCh38
rs967488336 Chr.8: 42897162-42897162 on Build GRCh38
rs535804364 Chr.8: 42897370-42897370 on Build GRCh38
rs557914624 Chr.8: 42897533-42897533 on Build GRCh38
rs975462330 Chr.8: 42897473-42897473 on Build GRCh38
rs937239880 Chr.8: 42897557-42897557 on Build GRCh38
rs952502968 Chr.8: 42897445-42897445 on Build GRCh38
rs922582385 Chr.8: 42897487-42897487 on Build GRCh38
rs924106006 Chr.8: 42897558-42897558 on Build GRCh38
rs983335983 Chr.8: 42897224-42897224 on Build GRCh38
rs367697887 Chr.8: 42897244-42897244 on Build GRCh38
rs918116868 Chr.8: 42897286-42897286 on Build GRCh38
rs779657271 Chr.8: 42897118-42897118 on Build GRCh38
rs573565675 Chr.8: 42897564-42897564 on Build GRCh38
rs1034005056 Chr.8: 42897460-42897460 on Build GRCh38
rs1006835851 Chr.8: 42897375-42897375 on Build GRCh38
rs769975228 Chr.8: 42897165-42897165 on Build GRCh38
rs951524286 Chr.8: 42897129-42897129 on Build GRCh38
rs902610034 Chr.8: 42897330-42897330 on Build GRCh38
rs551581089 Chr.8: 42897171-42897171 on Build GRCh38
rs1042533998 Chr.8: 42897320-42897320 on Build GRCh38
rs531446046 Chr.8: 42897131-42897131 on Build GRCh38
rs895593051 Chr.8: 42897357-42897357 on Build GRCh38
rs527599889 Chr.8: 42897250-42897250 on Build GRCh38
rs940515408 Chr.8: 42897552-42897552 on Build GRCh38
rs1001244462 Chr.8: 42897337-42897337 on Build GRCh38
rs949603857 Chr.8: 42897311-42897311 on Build GRCh38
rs938300933 Chr.8: 42897261-42897261 on Build GRCh38
rs991467644 Chr.8: 42897264-42897264 on Build GRCh38
rs1025833702 Chr.8: 42897223-42897223 on Build GRCh38
rs918174839 Chr.8: 42897530-42897530 on Build GRCh38
rs889781381 Chr.8: 42897355-42897355 on Build GRCh38
rs749452734 Chr.8: 42897179-42897179 on Build GRCh38
rs936697987 Chr.8: 42897336-42897336 on Build GRCh38
rs781455902 Chr.8: 42897154-42897154 on Build GRCh38
rs771001701 Chr.8: 42897218-42897218 on Build GRCh38
rs774374152 Chr.8: 42897225-42897225 on Build GRCh38
rs907722827 Chr.8: 42897284-42897284 on Build GRCh38
rs752769156 Chr.8: 42897123-42897124 on Build GRCh38
rs926166805 Chr.8: 42897228-42897228 on Build GRCh38
rs755460322 Chr.8: 42897145-42897145 on Build GRCh38
rs1033256919 Chr.8: 42897435-42897435 on Build GRCh38
rs112219235 Chr.8: 42897419-42897419 on Build GRCh38
rs972531738 Chr.8: 42897159-42897159 on Build GRCh38
rs994419750 Chr.8: 42897400-42897400 on Build GRCh38
rs571604061 Chr.8: 42897234-42897234 on Build GRCh38
rs566757417 Chr.8: 42897291-42897291 on Build GRCh38
rs577778489 Chr.8: 42897550-42897550 on Build GRCh38
rs141707234 Chr.8: 42897219-42897219 on Build GRCh38
rs964083982 Chr.8: 42897230-42897230 on Build GRCh38
rs983754915 Chr.8: 42897246-42897246 on Build GRCh38
rs992481605 Chr.8: 42897509-42897509 on Build GRCh38
rs908589833 Chr.8: 42897546-42897547 on Build GRCh38
rs748511367 Chr.8: 42897157-42897157 on Build GRCh38
rs372059901 Chr.8: 42897117-42897117 on Build GRCh38
rs960052442 Chr.8: 42897120-42897120 on Build GRCh38
rs978998961 Chr.8: 42897201-42897201 on Build GRCh38
rs534241285 Chr.8: 42897554-42897554 on Build GRCh38

View Full Product Details