Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58147744-58148251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM110B
Entrez Gene ID
90362
Gene Name
family with sequence similarity 110 member B
Gene Aliases
C8orf72
UniGene
Hs.154652
Gene Chromosome Location
Chr.8: 57994444-58149718 on Build GRCh38
Transcripts
NM_147189.2 XM_005251324.2 XM_005251325.3 XM_005251326.2 XM_017013948.1

SNP Detail

SNP ID Chromosome Location
rs150522357 Chr.8: 58148177-58148178 on Build GRCh38
rs748106392 Chr.8: 58147907-58147907 on Build GRCh38
rs752156469 Chr.8: 58148167-58148177 on Build GRCh38
rs1039165545 Chr.8: 58147862-58147862 on Build GRCh38
rs796431380 Chr.8: 58148152-58148152 on Build GRCh38
rs141893293 Chr.8: 58148166-58148166 on Build GRCh38
rs549296290 Chr.8: 58148174-58148174 on Build GRCh38
rs754811204 Chr.8: 58147824-58147824 on Build GRCh38
rs749812815 Chr.8: 58148109-58148109 on Build GRCh38
rs938229339 Chr.8: 58148100-58148100 on Build GRCh38
rs1013350269 Chr.8: 58148118-58148118 on Build GRCh38
rs561758511 Chr.8: 58147798-58147799 on Build GRCh38
rs767416124 Chr.8: 58148055-58148057 on Build GRCh38
rs567022372 Chr.8: 58148147-58148147 on Build GRCh38
rs58388044 Chr.8: 58148140-58148140 on Build GRCh38
rs1013078968 Chr.8: 58147957-58147957 on Build GRCh38
rs200430917 Chr.8: 58148171-58148172 on Build GRCh38
rs147919130 Chr.8: 58147853-58147853 on Build GRCh38
rs867948434 Chr.8: 58148172-58148172 on Build GRCh38
rs558186897 Chr.8: 58148191-58148191 on Build GRCh38
rs920784461 Chr.8: 58148173-58148173 on Build GRCh38
rs895527428 Chr.8: 58147948-58147948 on Build GRCh38
rs534980952 Chr.8: 58148178-58148178 on Build GRCh38
rs77218799 Chr.8: 58148090-58148090 on Build GRCh38
rs574206150 Chr.8: 58147956-58147956 on Build GRCh38
rs188380125 Chr.8: 58148066-58148066 on Build GRCh38
rs755153114 Chr.8: 58148112-58148112 on Build GRCh38
rs911954326 Chr.8: 58147833-58147833 on Build GRCh38
rs911968667 Chr.8: 58148098-58148098 on Build GRCh38
rs116233890 Chr.8: 58147813-58147813 on Build GRCh38
rs771839548 Chr.8: 58147793-58147793 on Build GRCh38
rs796737138 Chr.8: 58148173-58148176 on Build GRCh38
rs1003853416 Chr.8: 58147929-58147929 on Build GRCh38
rs868104643 Chr.8: 58148028-58148028 on Build GRCh38
rs559624843 Chr.8: 58147893-58147893 on Build GRCh38
rs1047979546 Chr.8: 58148214-58148214 on Build GRCh38
rs370583114 Chr.8: 58147949-58147949 on Build GRCh38
rs1047947097 Chr.8: 58147899-58147899 on Build GRCh38
rs896891199 Chr.8: 58148222-58148222 on Build GRCh38
rs896623764 Chr.8: 58148111-58148111 on Build GRCh38
rs562306555 Chr.8: 58147910-58147910 on Build GRCh38
rs866024628 Chr.8: 58147871-58147871 on Build GRCh38
rs1036770100 Chr.8: 58148181-58148181 on Build GRCh38
rs778643041 Chr.8: 58147869-58147869 on Build GRCh38
rs995121031 Chr.8: 58147883-58147883 on Build GRCh38
rs1035855822 Chr.8: 58147933-58147933 on Build GRCh38
rs781297675 Chr.8: 58148101-58148101 on Build GRCh38
rs978699329 Chr.8: 58147995-58147995 on Build GRCh38
rs896245449 Chr.8: 58147827-58147827 on Build GRCh38
rs945385476 Chr.8: 58148057-58148057 on Build GRCh38
rs1045369305 Chr.8: 58148157-58148157 on Build GRCh38
rs904088845 Chr.8: 58147791-58147791 on Build GRCh38
rs978668408 Chr.8: 58147788-58147788 on Build GRCh38
rs545001931 Chr.8: 58147940-58147940 on Build GRCh38
rs968527131 Chr.8: 58147980-58147980 on Build GRCh38
rs77096367 Chr.8: 58148177-58148177 on Build GRCh38
rs913977505 Chr.8: 58148037-58148037 on Build GRCh38
rs912349682 Chr.8: 58147961-58147961 on Build GRCh38
rs377258015 Chr.8: 58148186-58148187 on Build GRCh38
rs953960691 Chr.8: 58147904-58147904 on Build GRCh38
rs564509296 Chr.8: 58147943-58147943 on Build GRCh38
rs1031571266 Chr.8: 58148043-58148043 on Build GRCh38
rs778035121 Chr.8: 58147996-58147996 on Build GRCh38
rs966155343 Chr.8: 58147962-58147962 on Build GRCh38

View Full Product Details