Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58149579-58150090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM110B
Entrez Gene ID
90362
Gene Name
family with sequence similarity 110 member B
Gene Aliases
C8orf72
UniGene
Hs.154652
Gene Chromosome Location
Chr.8: 57994444-58149718 on Build GRCh38
Transcripts
NM_147189.2

SNP Detail

SNP ID Chromosome Location
rs888702690 Chr.8: 58149809-58149809 on Build GRCh38
rs537701825 Chr.8: 58150040-58150040 on Build GRCh38
rs749497341 Chr.8: 58149803-58149803 on Build GRCh38
rs796188618 Chr.8: 58149738-58149739 on Build GRCh38
rs778879408 Chr.8: 58149842-58149842 on Build GRCh38
rs368594588 Chr.8: 58149782-58149782 on Build GRCh38
rs753663078 Chr.8: 58149720-58149721 on Build GRCh38
rs879641103 Chr.8: 58149741-58149741 on Build GRCh38
rs979696349 Chr.8: 58149727-58149727 on Build GRCh38
rs145477914 Chr.8: 58149740-58149741 on Build GRCh38
rs879347152 Chr.8: 58150035-58150035 on Build GRCh38
rs1013670507 Chr.8: 58149676-58149676 on Build GRCh38
rs5891670 Chr.8: 58149719-58149719 on Build GRCh38
rs967111194 Chr.8: 58150000-58150000 on Build GRCh38
rs1048980468 Chr.8: 58149886-58149886 on Build GRCh38
rs894283208 Chr.8: 58149671-58149671 on Build GRCh38
rs957033015 Chr.8: 58149738-58149741 on Build GRCh38
rs754424093 Chr.8: 58149697-58149697 on Build GRCh38
rs1036811871 Chr.8: 58149947-58149947 on Build GRCh38
rs568833627 Chr.8: 58150009-58150009 on Build GRCh38
rs536145320 Chr.8: 58149987-58149987 on Build GRCh38
rs988842128 Chr.8: 58149759-58149760 on Build GRCh38
rs774739696 Chr.8: 58149988-58149988 on Build GRCh38
rs975844017 Chr.8: 58149830-58149830 on Build GRCh38
rs1354815 Chr.8: 58149717-58149717 on Build GRCh38
rs772573629 Chr.8: 58150034-58150034 on Build GRCh38
rs554560472 Chr.8: 58150056-58150056 on Build GRCh38
rs1032563953 Chr.8: 58149728-58149728 on Build GRCh38
rs887662524 Chr.8: 58149888-58149888 on Build GRCh38
rs189947244 Chr.8: 58149704-58149704 on Build GRCh38
rs944486865 Chr.8: 58149784-58149784 on Build GRCh38
rs546828556 Chr.8: 58149824-58149824 on Build GRCh38
rs921791239 Chr.8: 58149869-58149869 on Build GRCh38
rs1004811759 Chr.8: 58149623-58149623 on Build GRCh38
rs896525142 Chr.8: 58149675-58149675 on Build GRCh38
rs759339022 Chr.8: 58149719-58149721 on Build GRCh38
rs76704284 Chr.8: 58149998-58149998 on Build GRCh38
rs912894088 Chr.8: 58149762-58149762 on Build GRCh38
rs1046501833 Chr.8: 58149703-58149703 on Build GRCh38
rs879543370 Chr.8: 58149740-58149740 on Build GRCh38
rs931792635 Chr.8: 58149871-58149871 on Build GRCh38
rs1056037168 Chr.8: 58149604-58149604 on Build GRCh38
rs940699420 Chr.8: 58149944-58149944 on Build GRCh38
rs566937114 Chr.8: 58149862-58149862 on Build GRCh38
rs778639483 Chr.8: 58149721-58149721 on Build GRCh38
rs138662624 Chr.8: 58149980-58149980 on Build GRCh38
rs1024259296 Chr.8: 58149677-58149677 on Build GRCh38
rs1018872585 Chr.8: 58149852-58149852 on Build GRCh38
rs963859870 Chr.8: 58149709-58149709 on Build GRCh38
rs1025568993 Chr.8: 58149807-58149807 on Build GRCh38
rs567300713 Chr.8: 58149673-58149673 on Build GRCh38
rs906606942 Chr.8: 58149798-58149798 on Build GRCh38

View Full Product Details