Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 84280718-84281228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC28A3
Entrez Gene ID
64078
Gene Name
solute carrier family 28 member 3
Gene Aliases
CNT3
UniGene
Hs.535966
Gene Chromosome Location
Chr.9: 84273123-84368634 on Build GRCh38
Transcripts
NM_001199633.1 NM_022127.2 NR_037638.2 XM_011518905.2 XM_011518906.2 XM_011518907.2 XM_011518908.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7093073 c.1719C>T p.I573I Chr.9: 84280811-84280811 on Build GRCh38
5639715 c.1674C>T p.Y558Y Chr.9: 84280856-84280856 on Build GRCh38
1265834 c.1729A>G p.T577A Chr.9: 84280801-84280801 on Build GRCh38
6299546 c.1648-9T>A p.? Chr.9: 84280891-84280891 on Build GRCh38
7300392 c.1675G>A p.A559T Chr.9: 84280855-84280855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370998873 Chr.9: 84281077-84281077 on Build GRCh38
rs546259080 Chr.9: 84281105-84281105 on Build GRCh38
rs1029476248 Chr.9: 84280855-84280855 on Build GRCh38
rs985318211 Chr.9: 84280899-84280899 on Build GRCh38
rs1004332798 Chr.9: 84280743-84280743 on Build GRCh38
rs534360209 Chr.9: 84280832-84280832 on Build GRCh38
rs556329524 Chr.9: 84280893-84280893 on Build GRCh38
rs746679335 Chr.9: 84280864-84280864 on Build GRCh38
rs777580587 Chr.9: 84280811-84280811 on Build GRCh38
rs1036742938 Chr.9: 84281145-84281145 on Build GRCh38
rs1048417521 Chr.9: 84280991-84280991 on Build GRCh38
rs201060612 Chr.9: 84280879-84280879 on Build GRCh38
rs773727942 Chr.9: 84280865-84280865 on Build GRCh38
rs990191785 Chr.9: 84280975-84280975 on Build GRCh38
rs960115492 Chr.9: 84281057-84281057 on Build GRCh38
rs925589717 Chr.9: 84281172-84281172 on Build GRCh38
rs1041261681 Chr.9: 84280904-84280904 on Build GRCh38
rs767711336 Chr.9: 84280920-84280920 on Build GRCh38
rs554012823 Chr.9: 84281023-84281023 on Build GRCh38
rs773797100 Chr.9: 84280751-84280751 on Build GRCh38
rs981641122 Chr.9: 84280998-84280998 on Build GRCh38
rs757957901 Chr.9: 84280807-84280807 on Build GRCh38
rs66851409 Chr.9: 84281195-84281195 on Build GRCh38
rs937503742 Chr.9: 84281016-84281016 on Build GRCh38
rs762396296 Chr.9: 84280856-84280857 on Build GRCh38
rs912912178 Chr.9: 84280978-84280978 on Build GRCh38
rs201219440 Chr.9: 84280802-84280802 on Build GRCh38
rs942134604 Chr.9: 84281142-84281142 on Build GRCh38
rs766494782 Chr.9: 84280884-84280884 on Build GRCh38
rs771663064 Chr.9: 84280840-84280840 on Build GRCh38
rs897843983 Chr.9: 84281194-84281194 on Build GRCh38
rs376409630 Chr.9: 84280990-84280990 on Build GRCh38
rs777195572 Chr.9: 84280853-84280853 on Build GRCh38
rs545561834 Chr.9: 84281139-84281139 on Build GRCh38
rs146066088 Chr.9: 84280843-84280843 on Build GRCh38
rs1056006019 Chr.9: 84281075-84281075 on Build GRCh38
rs10868135 Chr.9: 84280938-84280938 on Build GRCh38
rs371756879 Chr.9: 84280780-84280780 on Build GRCh38
rs184620681 Chr.9: 84280873-84280873 on Build GRCh38
rs369935460 Chr.9: 84280890-84280890 on Build GRCh38
rs969947987 Chr.9: 84281113-84281113 on Build GRCh38
rs778850020 Chr.9: 84280830-84280830 on Build GRCh38
rs571932883 Chr.9: 84281080-84281080 on Build GRCh38
rs777105688 Chr.9: 84280775-84280775 on Build GRCh38
rs757197192 Chr.9: 84280829-84280829 on Build GRCh38
rs765762697 Chr.9: 84280789-84280789 on Build GRCh38
rs200468466 Chr.9: 84280821-84280821 on Build GRCh38
rs751607100 Chr.9: 84280898-84280898 on Build GRCh38
rs914031512 Chr.9: 84280895-84280895 on Build GRCh38
rs11568392 Chr.9: 84280856-84280856 on Build GRCh38
rs117481772 Chr.9: 84281004-84281004 on Build GRCh38
rs755258760 Chr.9: 84280908-84280908 on Build GRCh38
rs368332766 Chr.9: 84280771-84280771 on Build GRCh38
rs112950285 Chr.9: 84280863-84280863 on Build GRCh38
rs768419417 Chr.9: 84280852-84280852 on Build GRCh38
rs1018440247 Chr.9: 84280910-84280910 on Build GRCh38
rs767266970 Chr.9: 84280947-84280947 on Build GRCh38
rs749101126 Chr.9: 84280816-84280816 on Build GRCh38
rs374149371 Chr.9: 84280764-84280764 on Build GRCh38
rs757038926 Chr.9: 84280926-84280926 on Build GRCh38
rs747607766 Chr.9: 84280878-84280878 on Build GRCh38
rs755170584 Chr.9: 84280810-84280810 on Build GRCh38
rs1056270349 Chr.9: 84280752-84280752 on Build GRCh38
rs777123804 Chr.9: 84280831-84280831 on Build GRCh38
rs1004665280 Chr.9: 84281019-84281019 on Build GRCh38
rs753587453 Chr.9: 84280925-84280925 on Build GRCh38
rs747756033 Chr.9: 84280800-84280800 on Build GRCh38
rs745368018 Chr.9: 84280755-84280755 on Build GRCh38
rs746703852 Chr.9: 84280848-84280848 on Build GRCh38

View Full Product Details