Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129012147-129012647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3GLB2
Entrez Gene ID
56904
Gene Name
SH3 domain containing GRB2 like endophilin B2
Gene Aliases
PP6569, PP9455, RRIG1
UniGene
Hs.460238
Gene Chromosome Location
Chr.9: 129007792-129028353 on Build GRCh38
Transcripts
NM_001287045.1 NM_001287046.1 NM_020145.3 XM_005252098.2 XM_005252100.2 XM_005252101.3 XM_006717188.2 XM_006717189.2 XM_006717190.1 XM_006717191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5410507 c.562-1G>A p.? Chr.9: 129012299-129012299 on Build GRCh38
5410508 c.574-1G>A p.? Chr.9: 129012299-129012299 on Build GRCh38
4640897 c.628G>A p.A210T Chr.9: 129012244-129012244 on Build GRCh38
6362946 c.613A>G p.S205G Chr.9: 129012247-129012247 on Build GRCh38
1105802 c.602T>G p.I201S Chr.9: 129012258-129012258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771321511 Chr.9: 129012303-129012303 on Build GRCh38
rs750347190 Chr.9: 129012257-129012257 on Build GRCh38
rs945052346 Chr.9: 129012548-129012548 on Build GRCh38
rs1021320821 Chr.9: 129012278-129012278 on Build GRCh38
rs759957235 Chr.9: 129012318-129012318 on Build GRCh38
rs1049625721 Chr.9: 129012577-129012577 on Build GRCh38
rs1023910184 Chr.9: 129012331-129012331 on Build GRCh38
rs111853332 Chr.9: 129012313-129012313 on Build GRCh38
rs12006160 Chr.9: 129012249-129012249 on Build GRCh38
rs17433059 Chr.9: 129012236-129012236 on Build GRCh38
rs921867854 Chr.9: 129012429-129012429 on Build GRCh38
rs961698760 Chr.9: 129012367-129012367 on Build GRCh38
rs146310812 Chr.9: 129012409-129012409 on Build GRCh38
rs536062326 Chr.9: 129012581-129012581 on Build GRCh38
rs147238711 Chr.9: 129012562-129012562 on Build GRCh38
rs376650558 Chr.9: 129012322-129012322 on Build GRCh38
rs1000673438 Chr.9: 129012296-129012296 on Build GRCh38
rs999066967 Chr.9: 129012211-129012211 on Build GRCh38
rs775177312 Chr.9: 129012229-129012229 on Build GRCh38
rs768353956 Chr.9: 129012319-129012319 on Build GRCh38
rs774483984 Chr.9: 129012186-129012186 on Build GRCh38
rs927410401 Chr.9: 129012453-129012453 on Build GRCh38
rs1045118806 Chr.9: 129012588-129012588 on Build GRCh38
rs112830714 Chr.9: 129012170-129012170 on Build GRCh38
rs17433066 Chr.9: 129012563-129012563 on Build GRCh38
rs369139369 Chr.9: 129012330-129012330 on Build GRCh38
rs145492886 Chr.9: 129012239-129012239 on Build GRCh38
rs1032534961 Chr.9: 129012372-129012372 on Build GRCh38
rs192518933 Chr.9: 129012540-129012540 on Build GRCh38
rs778204978 Chr.9: 129012293-129012293 on Build GRCh38
rs944369809 Chr.9: 129012230-129012230 on Build GRCh38
rs370122853 Chr.9: 129012250-129012250 on Build GRCh38
rs952915175 Chr.9: 129012419-129012419 on Build GRCh38
rs931948535 Chr.9: 129012446-129012446 on Build GRCh38
rs767519525 Chr.9: 129012252-129012252 on Build GRCh38
rs772512808 Chr.9: 129012201-129012201 on Build GRCh38
rs892643408 Chr.9: 129012605-129012605 on Build GRCh38
rs1042014386 Chr.9: 129012557-129012557 on Build GRCh38
rs751054150 Chr.9: 129012244-129012244 on Build GRCh38
rs202071696 Chr.9: 129012297-129012297 on Build GRCh38
rs755216231 Chr.9: 129012260-129012260 on Build GRCh38
rs368272967 Chr.9: 129012210-129012210 on Build GRCh38
rs373557709 Chr.9: 129012344-129012344 on Build GRCh38
rs755838515 Chr.9: 129012304-129012304 on Build GRCh38
rs71499435 Chr.9: 129012205-129012205 on Build GRCh38
rs17452555 Chr.9: 129012407-129012407 on Build GRCh38
rs774533474 Chr.9: 129012192-129012193 on Build GRCh38
rs142602092 Chr.9: 129012292-129012292 on Build GRCh38
rs372519135 Chr.9: 129012218-129012218 on Build GRCh38
rs759110861 Chr.9: 129012338-129012339 on Build GRCh38
rs200547620 Chr.9: 129012237-129012237 on Build GRCh38
rs140847824 Chr.9: 129012217-129012218 on Build GRCh38
rs566744997 Chr.9: 129012547-129012547 on Build GRCh38
rs917663245 Chr.9: 129012545-129012545 on Build GRCh38
rs768073271 Chr.9: 129012600-129012600 on Build GRCh38
rs377705830 Chr.9: 129012477-129012477 on Build GRCh38
rs148878350 Chr.9: 129012258-129012258 on Build GRCh38
rs954138368 Chr.9: 129012300-129012300 on Build GRCh38
rs761409857 Chr.9: 129012251-129012251 on Build GRCh38
rs1008042347 Chr.9: 129012612-129012612 on Build GRCh38
rs748642839 Chr.9: 129012187-129012187 on Build GRCh38
rs1049436178 Chr.9: 129012452-129012452 on Build GRCh38
rs1001510641 Chr.9: 129012354-129012354 on Build GRCh38
rs748162107 Chr.9: 129012268-129012268 on Build GRCh38
rs771222722 Chr.9: 129012228-129012228 on Build GRCh38
rs541932525 Chr.9: 129012345-129012345 on Build GRCh38
rs949377297 Chr.9: 129012587-129012587 on Build GRCh38
rs551023750 Chr.9: 129012423-129012423 on Build GRCh38
rs1001164057 Chr.9: 129012591-129012591 on Build GRCh38
rs540029552 Chr.9: 129012173-129012173 on Build GRCh38
rs912950450 Chr.9: 129012418-129012418 on Build GRCh38
rs776915482 Chr.9: 129012306-129012306 on Build GRCh38
rs1020144712 Chr.9: 129012617-129012617 on Build GRCh38
rs905174415 Chr.9: 129012589-129012589 on Build GRCh38
rs938853191 Chr.9: 129012466-129012466 on Build GRCh38
rs779122765 Chr.9: 129012267-129012267 on Build GRCh38
rs972745946 Chr.9: 129012406-129012406 on Build GRCh38
rs562229250 Chr.9: 129012383-129012383 on Build GRCh38
rs754088561 Chr.9: 129012259-129012259 on Build GRCh38

View Full Product Details