Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129061092-129061624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIGA2
Entrez Gene ID
84895
Gene Name
mitoguardin 2
Gene Aliases
C9orf54, FAM73B
UniGene
Hs.632693
Gene Chromosome Location
Chr.9: 129036605-129072082 on Build GRCh38
Transcripts
NM_032809.2 XM_005252282.1 XM_011519117.1 XM_011519118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4513435 c.938C>T p.S313F Chr.9: 129061274-129061274 on Build GRCh38
1206447 c.916G>T p.G306* Chr.9: 129061252-129061252 on Build GRCh38
3382441 c.925C>G p.P309A Chr.9: 129061261-129061261 on Build GRCh38
1666413 c.942A>C p.R314S Chr.9: 129061278-129061278 on Build GRCh38
1105809 c.1010G>A p.R337K Chr.9: 129061346-129061346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374189232 Chr.9: 129061222-129061222 on Build GRCh38
rs141679421 Chr.9: 129061268-129061268 on Build GRCh38
rs762886496 Chr.9: 129061387-129061387 on Build GRCh38
rs369671866 Chr.9: 129061255-129061255 on Build GRCh38
rs1008181450 Chr.9: 129061500-129061500 on Build GRCh38
rs924882643 Chr.9: 129061459-129061459 on Build GRCh38
rs149794842 Chr.9: 129061118-129061118 on Build GRCh38
rs551935398 Chr.9: 129061441-129061441 on Build GRCh38
rs745352542 Chr.9: 129061366-129061366 on Build GRCh38
rs1024770414 Chr.9: 129061431-129061431 on Build GRCh38
rs762628738 Chr.9: 129061282-129061282 on Build GRCh38
rs200956473 Chr.9: 129061219-129061219 on Build GRCh38
rs201910125 Chr.9: 129061281-129061281 on Build GRCh38
rs188731796 Chr.9: 129061155-129061155 on Build GRCh38
rs560721763 Chr.9: 129061277-129061277 on Build GRCh38
rs955300876 Chr.9: 129061291-129061291 on Build GRCh38
rs1052054381 Chr.9: 129061490-129061490 on Build GRCh38
rs556772940 Chr.9: 129061150-129061150 on Build GRCh38
rs368058014 Chr.9: 129061218-129061218 on Build GRCh38
rs764204014 Chr.9: 129061244-129061244 on Build GRCh38
rs922318020 Chr.9: 129061498-129061498 on Build GRCh38
rs764004271 Chr.9: 129061284-129061284 on Build GRCh38
rs1047029242 Chr.9: 129061343-129061343 on Build GRCh38
rs928882188 Chr.9: 129061187-129061187 on Build GRCh38
rs775762007 Chr.9: 129061205-129061205 on Build GRCh38
rs75367930 Chr.9: 129061273-129061273 on Build GRCh38
rs542656714 Chr.9: 129061149-129061149 on Build GRCh38
rs372660490 Chr.9: 129061260-129061260 on Build GRCh38
rs1025530369 Chr.9: 129061141-129061141 on Build GRCh38
rs958778184 Chr.9: 129061420-129061420 on Build GRCh38
rs770296291 Chr.9: 129061202-129061202 on Build GRCh38
rs761953850 Chr.9: 129061309-129061309 on Build GRCh38
rs35584792 Chr.9: 129061241-129061241 on Build GRCh38
rs758239685 Chr.9: 129061243-129061243 on Build GRCh38
rs943495769 Chr.9: 129061476-129061476 on Build GRCh38
rs751871282 Chr.9: 129061246-129061246 on Build GRCh38
rs746428792 Chr.9: 129061189-129061189 on Build GRCh38
rs750452261 Chr.9: 129061322-129061322 on Build GRCh38
rs746733250 Chr.9: 129061269-129061269 on Build GRCh38
rs776215710 Chr.9: 129061276-129061276 on Build GRCh38
rs770669910 Chr.9: 129061357-129061357 on Build GRCh38
rs532296060 Chr.9: 129061427-129061427 on Build GRCh38
rs1018148606 Chr.9: 129061312-129061312 on Build GRCh38
rs752397380 Chr.9: 129061242-129061242 on Build GRCh38
rs1008524166 Chr.9: 129061299-129061299 on Build GRCh38
rs761669500 Chr.9: 129061358-129061359 on Build GRCh38
rs986665785 Chr.9: 129061140-129061140 on Build GRCh38
rs779280362 Chr.9: 129061353-129061353 on Build GRCh38
rs957639573 Chr.9: 129061461-129061461 on Build GRCh38
rs781188681 Chr.9: 129061251-129061251 on Build GRCh38
rs771618727 Chr.9: 129061408-129061408 on Build GRCh38
rs910762484 Chr.9: 129061475-129061475 on Build GRCh38
rs903625909 Chr.9: 129061477-129061477 on Build GRCh38
rs192848874 Chr.9: 129061176-129061176 on Build GRCh38
rs756572664 Chr.9: 129061259-129061259 on Build GRCh38
rs755285604 Chr.9: 129061334-129061334 on Build GRCh38
rs938984042 Chr.9: 129061426-129061426 on Build GRCh38
rs759490976 Chr.9: 129061234-129061234 on Build GRCh38
rs916109378 Chr.9: 129061428-129061428 on Build GRCh38
rs772736343 Chr.9: 129061265-129061265 on Build GRCh38
rs774300653 Chr.9: 129061359-129061362 on Build GRCh38
rs541164161 Chr.9: 129061220-129061220 on Build GRCh38
rs760802619 Chr.9: 129061173-129061173 on Build GRCh38
rs990347367 Chr.9: 129061462-129061462 on Build GRCh38
rs756233621 Chr.9: 129061330-129061330 on Build GRCh38
rs781017120 Chr.9: 129061360-129061360 on Build GRCh38
rs964164881 Chr.9: 129061142-129061142 on Build GRCh38
rs529645386 Chr.9: 129061321-129061321 on Build GRCh38
rs776698562 Chr.9: 129061229-129061229 on Build GRCh38
rs369192111 Chr.9: 129061487-129061487 on Build GRCh38
rs769808240 Chr.9: 129061278-129061278 on Build GRCh38
rs17508349 Chr.9: 129061209-129061209 on Build GRCh38
rs749592844 Chr.9: 129061262-129061262 on Build GRCh38
rs377537395 Chr.9: 129061367-129061367 on Build GRCh38
rs748467606 Chr.9: 129061356-129061356 on Build GRCh38
rs1016806914 Chr.9: 129061418-129061418 on Build GRCh38
rs184997028 Chr.9: 129061415-129061415 on Build GRCh38
rs765273969 Chr.9: 129061241-129061241 on Build GRCh38
rs12685995 Chr.9: 129061332-129061332 on Build GRCh38
rs775272445 Chr.9: 129061372-129061372 on Build GRCh38
rs370744025 Chr.9: 129061180-129061180 on Build GRCh38
rs977363874 Chr.9: 129061458-129061458 on Build GRCh38
rs964215099 Chr.9: 129061313-129061313 on Build GRCh38
rs991970649 Chr.9: 129061175-129061175 on Build GRCh38
rs769104486 Chr.9: 129061263-129061263 on Build GRCh38
rs974217091 Chr.9: 129061328-129061328 on Build GRCh38
rs545588962 Chr.9: 129061172-129061172 on Build GRCh38
rs919960186 Chr.9: 129061158-129061158 on Build GRCh38
rs1056770413 Chr.9: 129061541-129061541 on Build GRCh38
rs112458013 Chr.9: 129061119-129061119 on Build GRCh38
rs1039465549 Chr.9: 129061483-129061483 on Build GRCh38
rs751321289 Chr.9: 129061289-129061289 on Build GRCh38
rs148435936 Chr.9: 129061250-129061250 on Build GRCh38

View Full Product Details