Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 8696291-8696791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST5
Entrez Gene ID
6764
Gene Name
suppression of tumorigenicity 5
Gene Aliases
DENND2B, HTS1, p126
UniGene
Hs.117715
Gene Chromosome Location
Chr.11: 8693352-8910951 on Build GRCh38
Transcripts
NM_005418.3 NM_139157.2 NM_213618.1 XM_005253077.1 XM_005253083.2 XM_011520309.1 XM_011520310.1 XM_011520311.2 XM_011520312.1 XM_011520313.1 XM_011520314.1 XM_011520315.1 XM_011520316.1 XM_011520317.1 XM_011520318.1 XM_011520319.2 XM_011520323.2 XM_011520324.2 XM_011520325.1 XM_011520326.1 XM_011520327.1 XM_011520328.1 XM_011520329.1 XM_017018182.1 XM_017018183.1 XM_017018184.1 XM_017018185.1 XM_017018186.1 XM_017018187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5016344 c.3053-13_3066del27 p.? Chr.11: 8696653-8696679 on Build GRCh38
4541991 c.3106G>A p.E1036K Chr.11: 8696613-8696613 on Build GRCh38
5999935 c.3291G>A p.K1097K Chr.11: 8696428-8696428 on Build GRCh38
4835608 c.3108G>A p.E1036E Chr.11: 8696611-8696611 on Build GRCh38
6518694 c.3082G>T p.E1028* Chr.11: 8696637-8696637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567812901 Chr.11: 8696436-8696436 on Build GRCh38
rs932947725 Chr.11: 8696693-8696693 on Build GRCh38
rs148715601 Chr.11: 8696656-8696656 on Build GRCh38
rs1026016259 Chr.11: 8696408-8696408 on Build GRCh38
rs763633987 Chr.11: 8696575-8696575 on Build GRCh38
rs762547818 Chr.11: 8696560-8696560 on Build GRCh38
rs1051140130 Chr.11: 8696397-8696397 on Build GRCh38
rs778673206 Chr.11: 8696587-8696587 on Build GRCh38
rs759156688 Chr.11: 8696441-8696441 on Build GRCh38
rs773785306 Chr.11: 8696639-8696639 on Build GRCh38
rs564987849 Chr.11: 8696327-8696327 on Build GRCh38
rs368933981 Chr.11: 8696515-8696515 on Build GRCh38
rs1017125755 Chr.11: 8696710-8696710 on Build GRCh38
rs774975391 Chr.11: 8696664-8696664 on Build GRCh38
rs747128246 Chr.11: 8696686-8696687 on Build GRCh38
rs376195907 Chr.11: 8696506-8696506 on Build GRCh38
rs981410400 Chr.11: 8696534-8696534 on Build GRCh38
rs768821454 Chr.11: 8696623-8696623 on Build GRCh38
rs201604689 Chr.11: 8696392-8696392 on Build GRCh38
rs748649616 Chr.11: 8696624-8696624 on Build GRCh38
rs767052109 Chr.11: 8696445-8696445 on Build GRCh38
rs754270551 Chr.11: 8696398-8696399 on Build GRCh38
rs147500806 Chr.11: 8696471-8696471 on Build GRCh38
rs886157616 Chr.11: 8696701-8696701 on Build GRCh38
rs535482058 Chr.11: 8696720-8696720 on Build GRCh38
rs768876591 Chr.11: 8696525-8696525 on Build GRCh38
rs1002492840 Chr.11: 8696723-8696723 on Build GRCh38
rs753804784 Chr.11: 8696461-8696461 on Build GRCh38
rs778730422 Chr.11: 8696482-8696482 on Build GRCh38
rs770361207 Chr.11: 8696638-8696638 on Build GRCh38
rs772248609 Chr.11: 8696510-8696510 on Build GRCh38
rs761238624 Chr.11: 8696409-8696409 on Build GRCh38
rs761315771 Chr.11: 8696689-8696689 on Build GRCh38
rs147922058 Chr.11: 8696673-8696673 on Build GRCh38
rs932114726 Chr.11: 8696382-8696382 on Build GRCh38
rs752084533 Chr.11: 8696446-8696446 on Build GRCh38
rs375990683 Chr.11: 8696511-8696511 on Build GRCh38
rs774516655 Chr.11: 8696390-8696390 on Build GRCh38
rs767023929 Chr.11: 8696557-8696557 on Build GRCh38
rs758085735 Chr.11: 8696708-8696708 on Build GRCh38
rs891590697 Chr.11: 8696490-8696490 on Build GRCh38
rs765025927 Chr.11: 8696698-8696698 on Build GRCh38
rs763582695 Chr.11: 8696686-8696686 on Build GRCh38
rs141148522 Chr.11: 8696371-8696371 on Build GRCh38
rs932166616 Chr.11: 8696733-8696733 on Build GRCh38
rs943325526 Chr.11: 8696669-8696669 on Build GRCh38
rs750395906 Chr.11: 8696700-8696700 on Build GRCh38
rs376097962 Chr.11: 8696687-8696687 on Build GRCh38
rs140057590 Chr.11: 8696767-8696767 on Build GRCh38
rs997251503 Chr.11: 8696406-8696406 on Build GRCh38
rs763383775 Chr.11: 8696556-8696556 on Build GRCh38
rs144416409 Chr.11: 8696607-8696607 on Build GRCh38
rs770735756 Chr.11: 8696387-8696387 on Build GRCh38
rs143547948 Chr.11: 8696449-8696449 on Build GRCh38
rs772171480 Chr.11: 8696395-8696395 on Build GRCh38
rs770258833 Chr.11: 8696537-8696537 on Build GRCh38
rs550462448 Chr.11: 8696531-8696531 on Build GRCh38
rs752420477 Chr.11: 8696559-8696559 on Build GRCh38
rs777639826 Chr.11: 8696455-8696455 on Build GRCh38
rs779410240 Chr.11: 8696670-8696671 on Build GRCh38
rs150693370 Chr.11: 8696440-8696440 on Build GRCh38
rs960985154 Chr.11: 8696545-8696545 on Build GRCh38
rs750116478 Chr.11: 8696602-8696602 on Build GRCh38
rs757244722 Chr.11: 8696582-8696582 on Build GRCh38
rs764628411 Chr.11: 8696428-8696428 on Build GRCh38
rs753755540 Chr.11: 8696579-8696579 on Build GRCh38
rs372611807 Chr.11: 8696569-8696569 on Build GRCh38
rs137996775 Chr.11: 8696435-8696435 on Build GRCh38
rs775489030 Chr.11: 8696398-8696398 on Build GRCh38
rs1050604543 Chr.11: 8696755-8696755 on Build GRCh38
rs1004356369 Chr.11: 8696709-8696709 on Build GRCh38
rs368966752 Chr.11: 8696316-8696317 on Build GRCh38
rs139942657 Chr.11: 8696533-8696533 on Build GRCh38
rs766232298 Chr.11: 8696713-8696713 on Build GRCh38
rs774097846 Chr.11: 8696725-8696725 on Build GRCh38
rs747283241 Chr.11: 8696608-8696608 on Build GRCh38
rs370181975 Chr.11: 8696703-8696703 on Build GRCh38
rs902030019 Chr.11: 8696378-8696378 on Build GRCh38
rs979233189 Chr.11: 8696632-8696632 on Build GRCh38
rs940774385 Chr.11: 8696339-8696339 on Build GRCh38
rs144049692 Chr.11: 8696599-8696599 on Build GRCh38
rs755559515 Chr.11: 8696463-8696464 on Build GRCh38
rs1804570 Chr.11: 8696590-8696590 on Build GRCh38
rs149828734 Chr.11: 8696649-8696649 on Build GRCh38
rs904077084 Chr.11: 8696420-8696420 on Build GRCh38
rs773519203 Chr.11: 8696538-8696538 on Build GRCh38

View Full Product Details