Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43638627-43639173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER2
Entrez Gene ID
117155
Gene Name
cation channel sperm associated 2
Gene Aliases
-
UniGene
Hs.662284
Gene Chromosome Location
Chr.15: 43630562-43648845 on Build GRCh38
Transcripts
NM_001282309.1 NM_001282310.1 NM_054020.3 NM_172095.2 NR_110319.1 XM_017021904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
962045 c.837C>A p.F279L Chr.15: 43638909-43638909 on Build GRCh38
5594356 c.811C>T p.R271C Chr.15: 43638935-43638935 on Build GRCh38
5726570 c.735G>A p.L245L Chr.15: 43639011-43639011 on Build GRCh38
6311162 c.791C>A p.S264* Chr.15: 43638955-43638955 on Build GRCh38
962044 c.842+1G>C p.? Chr.15: 43638903-43638903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764458768 Chr.15: 43639016-43639016 on Build GRCh38
rs780874595 Chr.15: 43638976-43638976 on Build GRCh38
rs1028858192 Chr.15: 43638810-43638810 on Build GRCh38
rs754746013 Chr.15: 43638967-43638967 on Build GRCh38
rs987794768 Chr.15: 43638824-43638824 on Build GRCh38
rs899286943 Chr.15: 43638971-43638971 on Build GRCh38
rs994656633 Chr.15: 43638788-43638788 on Build GRCh38
rs543629175 Chr.15: 43638706-43638706 on Build GRCh38
rs772431410 Chr.15: 43639008-43639008 on Build GRCh38
rs371464646 Chr.15: 43639034-43639034 on Build GRCh38
rs906534368 Chr.15: 43638680-43638680 on Build GRCh38
rs771418615 Chr.15: 43638990-43638990 on Build GRCh38
rs144487721 Chr.15: 43638815-43638815 on Build GRCh38
rs767315535 Chr.15: 43639044-43639044 on Build GRCh38
rs774680387 Chr.15: 43638935-43638935 on Build GRCh38
rs916720743 Chr.15: 43639133-43639133 on Build GRCh38
rs534595496 Chr.15: 43639132-43639132 on Build GRCh38
rs539776990 Chr.15: 43638922-43638922 on Build GRCh38
rs761918326 Chr.15: 43639020-43639020 on Build GRCh38
rs775746705 Chr.15: 43639010-43639010 on Build GRCh38
rs776848098 Chr.15: 43639017-43639017 on Build GRCh38
rs998956134 Chr.15: 43639051-43639051 on Build GRCh38
rs750335864 Chr.15: 43638885-43638885 on Build GRCh38
rs769247393 Chr.15: 43638916-43638916 on Build GRCh38
rs1015440302 Chr.15: 43638745-43638745 on Build GRCh38
rs372281377 Chr.15: 43638855-43638855 on Build GRCh38
rs948028069 Chr.15: 43638662-43638662 on Build GRCh38
rs755754604 Chr.15: 43639049-43639049 on Build GRCh38
rs753490249 Chr.15: 43639068-43639068 on Build GRCh38
rs140907870 Chr.15: 43638963-43638963 on Build GRCh38
rs1004940038 Chr.15: 43638687-43638687 on Build GRCh38
rs760920752 Chr.15: 43639013-43639013 on Build GRCh38
rs780615430 Chr.15: 43638938-43638938 on Build GRCh38
rs1018581280 Chr.15: 43638832-43638832 on Build GRCh38
rs2447194 Chr.15: 43639153-43639153 on Build GRCh38
rs368716853 Chr.15: 43638882-43638882 on Build GRCh38
rs763850019 Chr.15: 43638915-43638916 on Build GRCh38
rs752712375 Chr.15: 43638879-43638879 on Build GRCh38
rs778347224 Chr.15: 43638868-43638868 on Build GRCh38
rs558511342 Chr.15: 43639094-43639094 on Build GRCh38
rs994625588 Chr.15: 43638977-43638977 on Build GRCh38
rs754906468 Chr.15: 43639035-43639035 on Build GRCh38
rs749767558 Chr.15: 43638934-43638934 on Build GRCh38
rs576972008 Chr.15: 43639099-43639099 on Build GRCh38
rs991239942 Chr.15: 43639100-43639100 on Build GRCh38
rs767775803 Chr.15: 43638943-43638943 on Build GRCh38
rs376418301 Chr.15: 43638894-43638894 on Build GRCh38
rs1047098503 Chr.15: 43638955-43638955 on Build GRCh38
rs370694104 Chr.15: 43638880-43638880 on Build GRCh38
rs555127142 Chr.15: 43639047-43639047 on Build GRCh38
rs777246065 Chr.15: 43638985-43638985 on Build GRCh38
rs185553210 Chr.15: 43639144-43639144 on Build GRCh38
rs764229568 Chr.15: 43638951-43638951 on Build GRCh38
rs758398486 Chr.15: 43638960-43638960 on Build GRCh38
rs760827905 Chr.15: 43638946-43638946 on Build GRCh38
rs368268477 Chr.15: 43638962-43638962 on Build GRCh38
rs374608515 Chr.15: 43638752-43638752 on Build GRCh38
rs572108001 Chr.15: 43639113-43639113 on Build GRCh38
rs373885397 Chr.15: 43638992-43638992 on Build GRCh38
rs746880228 Chr.15: 43638898-43638898 on Build GRCh38
rs140331423 Chr.15: 43639022-43639022 on Build GRCh38
rs746372122 Chr.15: 43638998-43638998 on Build GRCh38
rs967463292 Chr.15: 43638842-43638842 on Build GRCh38
rs777466043 Chr.15: 43639052-43639052 on Build GRCh38
rs775041018 Chr.15: 43638857-43638857 on Build GRCh38
rs181613755 Chr.15: 43638858-43638858 on Build GRCh38
rs753114067 Chr.15: 43638835-43638835 on Build GRCh38
rs748851111 Chr.15: 43638986-43638986 on Build GRCh38
rs755808045 Chr.15: 43638979-43638979 on Build GRCh38
rs113881140 Chr.15: 43638807-43638807 on Build GRCh38
rs753753401 Chr.15: 43638989-43638991 on Build GRCh38
rs747799489 Chr.15: 43638978-43638978 on Build GRCh38
rs868402632 Chr.15: 43638846-43638846 on Build GRCh38
rs767833332 Chr.15: 43638874-43638874 on Build GRCh38
rs563367483 Chr.15: 43638744-43638744 on Build GRCh38
rs1046431901 Chr.15: 43638674-43638674 on Build GRCh38
rs199516208 Chr.15: 43638903-43638903 on Build GRCh38
rs754009898 Chr.15: 43638957-43638957 on Build GRCh38
rs142276830 Chr.15: 43638786-43638786 on Build GRCh38
rs150929073 Chr.15: 43638944-43638944 on Build GRCh38
rs112488180 Chr.15: 43638904-43638904 on Build GRCh38
rs191847990 Chr.15: 43638704-43638704 on Build GRCh38
rs751964673 Chr.15: 43638870-43638870 on Build GRCh38
rs760511322 Chr.15: 43638912-43638912 on Build GRCh38

View Full Product Details