Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32177894-32178403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT1
Entrez Gene ID
10554
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases
1-AGPAT1, G15, LPAAT-alpha, LPAATA
UniGene
Hs.409230
Gene Chromosome Location
Chr.6: 32168206-32178111 on Build GRCh38
Transcripts
NM_032741.4 NM_006913.3

Gene Symbol
RNF5
Entrez Gene ID
6048
Gene Name
ring finger protein 5
Gene Aliases
RING5, RMA1
UniGene
Hs.731774
Gene Chromosome Location
Chr.6: 32178385-32180793 on Build GRCh38
Transcripts
NM_032741.4 NM_006913.3

SNP Detail

SNP ID Chromosome Location
rs564964384 Chr.6: 32178160-32178160 on Build GRCh38
rs141920214 Chr.6: 32178088-32178088 on Build GRCh38
rs3134946 Chr.6: 32178216-32178216 on Build GRCh38
rs59564381 Chr.6: 32178089-32178089 on Build GRCh38
rs3096689 Chr.6: 32178222-32178222 on Build GRCh38
rs1032759829 Chr.6: 32178183-32178183 on Build GRCh38
rs750251880 Chr.6: 32177974-32177974 on Build GRCh38
rs987241890 Chr.6: 32177945-32177945 on Build GRCh38
rs908483726 Chr.6: 32178071-32178071 on Build GRCh38
rs1008194132 Chr.6: 32178236-32178236 on Build GRCh38
rs1013946386 Chr.6: 32178115-32178115 on Build GRCh38
rs542758753 Chr.6: 32178001-32178001 on Build GRCh38
rs570137390 Chr.6: 32178361-32178361 on Build GRCh38
rs543495442 Chr.6: 32178117-32178117 on Build GRCh38
rs867828708 Chr.6: 32178016-32178016 on Build GRCh38
rs931201678 Chr.6: 32178048-32178048 on Build GRCh38
rs531796614 Chr.6: 32178103-32178103 on Build GRCh38
rs895696774 Chr.6: 32178091-32178091 on Build GRCh38
rs975211786 Chr.6: 32178009-32178009 on Build GRCh38
rs912651479 Chr.6: 32177960-32177960 on Build GRCh38
rs554099349 Chr.6: 32177923-32177923 on Build GRCh38
rs982761892 Chr.6: 32178333-32178333 on Build GRCh38
rs767410119 Chr.6: 32178259-32178259 on Build GRCh38
rs1046957074 Chr.6: 32178058-32178058 on Build GRCh38
rs1055859174 Chr.6: 32178136-32178136 on Build GRCh38
rs184315351 Chr.6: 32178077-32178077 on Build GRCh38
rs949073025 Chr.6: 32178074-32178074 on Build GRCh38
rs967058334 Chr.6: 32178150-32178150 on Build GRCh38
rs2269423 Chr.6: 32177930-32177930 on Build GRCh38
rs1012180166 Chr.6: 32178200-32178200 on Build GRCh38
rs530254587 Chr.6: 32178344-32178344 on Build GRCh38
rs74395371 Chr.6: 32178356-32178356 on Build GRCh38
rs1019636511 Chr.6: 32178298-32178298 on Build GRCh38
rs753569466 Chr.6: 32178032-32178033 on Build GRCh38
rs752690289 Chr.6: 32178263-32178263 on Build GRCh38
rs187465580 Chr.6: 32178323-32178323 on Build GRCh38

View Full Product Details