Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 132644653-132645219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR39
Entrez Gene ID
2863
Gene Name
G protein-coupled receptor 39
Gene Aliases
-
UniGene
Hs.432395
Gene Chromosome Location
Chr.2: 132415972-132646596 on Build GRCh38
Transcripts
NM_001508.2 NM_001077427.3 NM_001321234.1 NM_001321235.1 NM_144586.6 XM_011510546.2

Gene Symbol
LYPD1
Entrez Gene ID
116372
Gene Name
LY6/PLAUR domain containing 1
Gene Aliases
LYPDC1, PHTS
UniGene
Hs.432395
Gene Chromosome Location
Chr.2: 132643280-132671579 on Build GRCh38
Transcripts
NM_001508.2 NM_001077427.3 NM_001321234.1 NM_001321235.1 NM_144586.6 XM_011510546.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
244775 c.880G>A p.V294I Chr.2: 132645124-132645124 on Build GRCh38
4688159 c.960G>T p.R320R Chr.2: 132645204-132645204 on Build GRCh38
6316168 c.919G>T p.A307S Chr.2: 132645163-132645163 on Build GRCh38
1006607 c.904C>T p.R302W Chr.2: 132645148-132645148 on Build GRCh38
5045561 c.962C>T p.A321V Chr.2: 132645206-132645206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372232546 Chr.2: 132645089-132645089 on Build GRCh38
rs534408545 Chr.2: 132645149-132645149 on Build GRCh38
rs549778402 Chr.2: 132644823-132644823 on Build GRCh38
rs771562914 Chr.2: 132645091-132645091 on Build GRCh38
rs752437139 Chr.2: 132645058-132645059 on Build GRCh38
rs143160922 Chr.2: 132645145-132645145 on Build GRCh38
rs151118262 Chr.2: 132645123-132645123 on Build GRCh38
rs765706388 Chr.2: 132644770-132644771 on Build GRCh38
rs775441952 Chr.2: 132645180-132645180 on Build GRCh38
rs755631699 Chr.2: 132644899-132644899 on Build GRCh38
rs929616785 Chr.2: 132644824-132644824 on Build GRCh38
rs749641131 Chr.2: 132645164-132645164 on Build GRCh38
rs751474067 Chr.2: 132645192-132645192 on Build GRCh38
rs766108578 Chr.2: 132645189-132645189 on Build GRCh38
rs978085205 Chr.2: 132644758-132644758 on Build GRCh38
rs982307046 Chr.2: 132645043-132645043 on Build GRCh38
rs929729654 Chr.2: 132644743-132644744 on Build GRCh38
rs376874164 Chr.2: 132645050-132645050 on Build GRCh38
rs3043654 Chr.2: 132644690-132644691 on Build GRCh38
rs1046155536 Chr.2: 132644816-132644816 on Build GRCh38
rs772128534 Chr.2: 132645178-132645178 on Build GRCh38
rs924181147 Chr.2: 132644793-132644793 on Build GRCh38
rs779934537 Chr.2: 132645126-132645126 on Build GRCh38
rs978054043 Chr.2: 132644903-132644903 on Build GRCh38
rs1013633218 Chr.2: 132644831-132644831 on Build GRCh38
rs751499179 Chr.2: 132645138-132645138 on Build GRCh38
rs899633111 Chr.2: 132644872-132644872 on Build GRCh38
rs1131671 Chr.2: 132645034-132645034 on Build GRCh38
rs141367491 Chr.2: 132644852-132644852 on Build GRCh38
rs267598880 Chr.2: 132645181-132645181 on Build GRCh38
rs537012526 Chr.2: 132645096-132645096 on Build GRCh38
rs183865697 Chr.2: 132644801-132644801 on Build GRCh38
rs892596892 Chr.2: 132644816-132644817 on Build GRCh38
rs916695578 Chr.2: 132644808-132644808 on Build GRCh38
rs777339841 Chr.2: 132645103-132645104 on Build GRCh38
rs756848610 Chr.2: 132645138-132645139 on Build GRCh38
rs951157474 Chr.2: 132644999-132644999 on Build GRCh38
rs777637145 Chr.2: 132645155-132645155 on Build GRCh38
rs752927626 Chr.2: 132645117-132645117 on Build GRCh38
rs538487466 Chr.2: 132644911-132644911 on Build GRCh38
rs141510593 Chr.2: 132644906-132644906 on Build GRCh38
rs948261088 Chr.2: 132644815-132644815 on Build GRCh38
rs948202472 Chr.2: 132645041-132645041 on Build GRCh38
rs1005844130 Chr.2: 132644857-132644857 on Build GRCh38
rs767550440 Chr.2: 132645199-132645199 on Build GRCh38
rs774875035 Chr.2: 132645166-132645166 on Build GRCh38
rs778485409 Chr.2: 132645122-132645122 on Build GRCh38
rs763051509 Chr.2: 132645188-132645188 on Build GRCh38
rs35301078 Chr.2: 132645016-132645017 on Build GRCh38
rs769557799 Chr.2: 132645150-132645150 on Build GRCh38
rs1030353499 Chr.2: 132644893-132644893 on Build GRCh38
rs147418150 Chr.2: 132645023-132645023 on Build GRCh38
rs1131667 Chr.2: 132645056-132645056 on Build GRCh38
rs990110249 Chr.2: 132645026-132645026 on Build GRCh38
rs1053053840 Chr.2: 132644809-132644809 on Build GRCh38
rs762604633 Chr.2: 132644877-132644877 on Build GRCh38
rs547906872 Chr.2: 132644943-132644943 on Build GRCh38
rs71929358 Chr.2: 132644795-132644798 on Build GRCh38
rs1046799630 Chr.2: 132644841-132644841 on Build GRCh38
rs148565069 Chr.2: 132645118-132645118 on Build GRCh38
rs1031459447 Chr.2: 132645012-132645012 on Build GRCh38
rs200065072 Chr.2: 132644683-132644683 on Build GRCh38
rs759995225 Chr.2: 132645176-132645176 on Build GRCh38
rs746438443 Chr.2: 132645171-132645172 on Build GRCh38
rs566135107 Chr.2: 132644972-132644972 on Build GRCh38
rs1018200779 Chr.2: 132644868-132644868 on Build GRCh38
rs780598440 Chr.2: 132645148-132645148 on Build GRCh38
rs375942168 Chr.2: 132645184-132645184 on Build GRCh38
rs541754640 Chr.2: 132644752-132644752 on Build GRCh38
rs869261035 Chr.2: 132644691-132644692 on Build GRCh38
rs768089283 Chr.2: 132645053-132645053 on Build GRCh38
rs918396574 Chr.2: 132644742-132644743 on Build GRCh38
rs368291385 Chr.2: 132645086-132645086 on Build GRCh38
rs370197937 Chr.2: 132644681-132644681 on Build GRCh38
rs892670592 Chr.2: 132645116-132645116 on Build GRCh38
rs1025900904 Chr.2: 132644949-132644949 on Build GRCh38
rs67974310 Chr.2: 132644684-132644685 on Build GRCh38
rs997991777 Chr.2: 132644889-132644889 on Build GRCh38
rs755789707 Chr.2: 132645073-132645074 on Build GRCh38
rs150565296 Chr.2: 132644683-132644684 on Build GRCh38
rs748759867 Chr.2: 132645132-132645143 on Build GRCh38
rs767520443 Chr.2: 132645107-132645107 on Build GRCh38
rs375272701 Chr.2: 132645074-132645074 on Build GRCh38
rs912310246 Chr.2: 132645029-132645029 on Build GRCh38
rs758254362 Chr.2: 132645124-132645124 on Build GRCh38
rs372259135 Chr.2: 132645088-132645088 on Build GRCh38
rs754885823 Chr.2: 132645193-132645193 on Build GRCh38
rs1002597026 Chr.2: 132644865-132644865 on Build GRCh38
rs975499105 Chr.2: 132645049-132645049 on Build GRCh38
rs771473286 Chr.2: 132645165-132645165 on Build GRCh38
rs759163909 Chr.2: 132644743-132644746 on Build GRCh38
rs767345408 Chr.2: 132645053-132645054 on Build GRCh38
rs563244879 Chr.2: 132644794-132644794 on Build GRCh38
rs1030384627 Chr.2: 132644716-132644716 on Build GRCh38
rs59022117 Chr.2: 132644801-132644804 on Build GRCh38
rs778258456 Chr.2: 132645156-132645157 on Build GRCh38
rs759642837 Chr.2: 132645100-132645100 on Build GRCh38

View Full Product Details