Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205088201-205088776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP5
Entrez Gene ID
5929
Gene Name
RB binding protein 5, histone lysine methyltransferase complex subunit
Gene Aliases
RBQ3, SWD1
UniGene
Hs.519230
Gene Chromosome Location
Chr.1: 205086142-205122022 on Build GRCh38
Transcripts
NM_001193272.1 NM_001193273.1 NM_005057.3 XM_011509833.2

SNP Detail

SNP ID Chromosome Location
rs563961093 Chr.1: 205088482-205088483 on Build GRCh38
rs958519171 Chr.1: 205088543-205088543 on Build GRCh38
rs907026906 Chr.1: 205088323-205088323 on Build GRCh38
rs1055018828 Chr.1: 205088272-205088272 on Build GRCh38
rs1037262377 Chr.1: 205088735-205088741 on Build GRCh38
rs762173105 Chr.1: 205088489-205088489 on Build GRCh38
rs138053747 Chr.1: 205088478-205088478 on Build GRCh38
rs555076570 Chr.1: 205088402-205088402 on Build GRCh38
rs957652124 Chr.1: 205088441-205088441 on Build GRCh38
rs779661123 Chr.1: 205088407-205088407 on Build GRCh38
rs1033220859 Chr.1: 205088419-205088419 on Build GRCh38
rs971101042 Chr.1: 205088553-205088553 on Build GRCh38
rs1040472101 Chr.1: 205088394-205088394 on Build GRCh38
rs771737258 Chr.1: 205088747-205088747 on Build GRCh38
rs757695936 Chr.1: 205088243-205088243 on Build GRCh38
rs978641259 Chr.1: 205088533-205088533 on Build GRCh38
rs925487001 Chr.1: 205088227-205088227 on Build GRCh38
rs375511841 Chr.1: 205088455-205088455 on Build GRCh38
rs11240354 Chr.1: 205088354-205088354 on Build GRCh38
rs765245138 Chr.1: 205088618-205088618 on Build GRCh38
rs577613393 Chr.1: 205088639-205088639 on Build GRCh38
rs1004417683 Chr.1: 205088329-205088329 on Build GRCh38
rs1038656914 Chr.1: 205088275-205088275 on Build GRCh38
rs754833130 Chr.1: 205088406-205088406 on Build GRCh38
rs1027976130 Chr.1: 205088443-205088443 on Build GRCh38
rs953731772 Chr.1: 205088449-205088449 on Build GRCh38
rs1047787134 Chr.1: 205088305-205088305 on Build GRCh38
rs777352467 Chr.1: 205088750-205088750 on Build GRCh38
rs116452997 Chr.1: 205088680-205088680 on Build GRCh38
rs958786325 Chr.1: 205088628-205088628 on Build GRCh38
rs76361181 Chr.1: 205088398-205088398 on Build GRCh38
rs751539913 Chr.1: 205088731-205088731 on Build GRCh38
rs925485842 Chr.1: 205088539-205088539 on Build GRCh38
rs535182260 Chr.1: 205088570-205088570 on Build GRCh38
rs182208082 Chr.1: 205088517-205088517 on Build GRCh38
rs940801324 Chr.1: 205088727-205088727 on Build GRCh38
rs376720708 Chr.1: 205088610-205088610 on Build GRCh38
rs16855072 Chr.1: 205088277-205088277 on Build GRCh38
rs12145807 Chr.1: 205088292-205088292 on Build GRCh38
rs886502022 Chr.1: 205088351-205088351 on Build GRCh38
rs929898531 Chr.1: 205088742-205088742 on Build GRCh38
rs369357886 Chr.1: 205088689-205088689 on Build GRCh38
rs189203708 Chr.1: 205088276-205088276 on Build GRCh38
rs140376352 Chr.1: 205088248-205088248 on Build GRCh38
rs973969987 Chr.1: 205088670-205088670 on Build GRCh38
rs550598127 Chr.1: 205088232-205088232 on Build GRCh38
rs878946602 Chr.1: 205088622-205088622 on Build GRCh38
rs1048948 Chr.1: 205088596-205088596 on Build GRCh38
rs981565658 Chr.1: 205088609-205088609 on Build GRCh38
rs1046502183 Chr.1: 205088307-205088309 on Build GRCh38
rs907930355 Chr.1: 205088725-205088725 on Build GRCh38
rs936956965 Chr.1: 205088237-205088237 on Build GRCh38
rs778686359 Chr.1: 205088525-205088525 on Build GRCh38

View Full Product Details