Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87846172-87846753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEPE
Entrez Gene ID
56955
Gene Name
matrix extracellular phosphoglycoprotein
Gene Aliases
OF45
UniGene
Hs.676357
Gene Chromosome Location
Chr.4: 87821398-87846817 on Build GRCh38
Transcripts
NM_001184694.2 NM_001184695.1 NM_001184696.1 NM_001184697.1 NM_001291183.1 NM_020203.3 XM_006714278.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6496818 c.1308G>T p.L436L Chr.4: 87846176-87846176 on Build GRCh38
3918314 c.1348G>A p.E450K Chr.4: 87846216-87846216 on Build GRCh38
1319302 c.1367G>A p.G456D Chr.4: 87846235-87846235 on Build GRCh38
6711492 c.1392A>G p.T464T Chr.4: 87846260-87846260 on Build GRCh38
2957259 c.1311C>T p.P437P Chr.4: 87846179-87846179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771776733 Chr.4: 87846434-87846434 on Build GRCh38
rs575199484 Chr.4: 87846331-87846331 on Build GRCh38
rs748314336 Chr.4: 87846433-87846433 on Build GRCh38
rs952550039 Chr.4: 87846241-87846241 on Build GRCh38
rs372163675 Chr.4: 87846451-87846451 on Build GRCh38
rs758762875 Chr.4: 87846203-87846203 on Build GRCh38
rs995669808 Chr.4: 87846654-87846655 on Build GRCh38
rs143167488 Chr.4: 87846376-87846376 on Build GRCh38
rs763209961 Chr.4: 87846253-87846253 on Build GRCh38
rs554226584 Chr.4: 87846528-87846528 on Build GRCh38
rs149174816 Chr.4: 87846280-87846280 on Build GRCh38
rs779077375 Chr.4: 87846488-87846488 on Build GRCh38
rs150153522 Chr.4: 87846215-87846215 on Build GRCh38
rs561124973 Chr.4: 87846523-87846523 on Build GRCh38
rs774083608 Chr.4: 87846246-87846246 on Build GRCh38
rs749358196 Chr.4: 87846402-87846402 on Build GRCh38
rs138152179 Chr.4: 87846310-87846310 on Build GRCh38
rs527970865 Chr.4: 87846423-87846423 on Build GRCh38
rs984068372 Chr.4: 87846245-87846245 on Build GRCh38
rs61731017 Chr.4: 87846214-87846214 on Build GRCh38
rs574032295 Chr.4: 87846701-87846701 on Build GRCh38
rs927449629 Chr.4: 87846499-87846499 on Build GRCh38
rs938808161 Chr.4: 87846502-87846502 on Build GRCh38
rs774334422 Chr.4: 87846341-87846341 on Build GRCh38
rs761045015 Chr.4: 87846465-87846466 on Build GRCh38
rs960039415 Chr.4: 87846536-87846536 on Build GRCh38
rs753441970 Chr.4: 87846292-87846292 on Build GRCh38
rs200838653 Chr.4: 87846395-87846395 on Build GRCh38
rs755112401 Chr.4: 87846487-87846487 on Build GRCh38
rs751991145 Chr.4: 87846382-87846382 on Build GRCh38
rs924743675 Chr.4: 87846626-87846626 on Build GRCh38
rs962075221 Chr.4: 87846252-87846252 on Build GRCh38
rs1003908927 Chr.4: 87846551-87846551 on Build GRCh38
rs368881949 Chr.4: 87846465-87846465 on Build GRCh38
rs775140234 Chr.4: 87846458-87846458 on Build GRCh38
rs112631801 Chr.4: 87846492-87846492 on Build GRCh38
rs368990363 Chr.4: 87846466-87846466 on Build GRCh38
rs371234606 Chr.4: 87846221-87846221 on Build GRCh38
rs61731015 Chr.4: 87846309-87846309 on Build GRCh38
rs75514522 Chr.4: 87846250-87846250 on Build GRCh38
rs760599019 Chr.4: 87846362-87846362 on Build GRCh38
rs151093256 Chr.4: 87846209-87846209 on Build GRCh38
rs761883793 Chr.4: 87846274-87846274 on Build GRCh38
rs770123763 Chr.4: 87846249-87846249 on Build GRCh38
rs757821304 Chr.4: 87846320-87846320 on Build GRCh38
rs961289274 Chr.4: 87846468-87846468 on Build GRCh38
rs367970153 Chr.4: 87846351-87846351 on Build GRCh38
rs781549826 Chr.4: 87846322-87846322 on Build GRCh38
rs989205126 Chr.4: 87846467-87846467 on Build GRCh38
rs1015374647 Chr.4: 87846356-87846356 on Build GRCh38
rs188533121 Chr.4: 87846618-87846618 on Build GRCh38
rs761606055 Chr.4: 87846346-87846346 on Build GRCh38
rs779271558 Chr.4: 87846426-87846426 on Build GRCh38
rs746948427 Chr.4: 87846439-87846439 on Build GRCh38
rs757627001 Chr.4: 87846387-87846387 on Build GRCh38
rs868133447 Chr.4: 87846216-87846216 on Build GRCh38
rs549816204 Chr.4: 87846702-87846702 on Build GRCh38
rs770867648 Chr.4: 87846448-87846448 on Build GRCh38
rs370126410 Chr.4: 87846603-87846603 on Build GRCh38
rs992011363 Chr.4: 87846547-87846547 on Build GRCh38
rs767934775 Chr.4: 87846431-87846432 on Build GRCh38
rs760099024 Chr.4: 87846282-87846282 on Build GRCh38
rs61731016 Chr.4: 87846329-87846329 on Build GRCh38
rs932318762 Chr.4: 87846704-87846704 on Build GRCh38
rs764664410 Chr.4: 87846457-87846457 on Build GRCh38
rs201756883 Chr.4: 87846365-87846365 on Build GRCh38
rs886680159 Chr.4: 87846549-87846549 on Build GRCh38
rs750916260 Chr.4: 87846393-87846393 on Build GRCh38
rs759045131 Chr.4: 87846454-87846454 on Build GRCh38
rs577928169 Chr.4: 87846337-87846337 on Build GRCh38
rs774250152 Chr.4: 87846271-87846271 on Build GRCh38
rs754036412 Chr.4: 87846484-87846484 on Build GRCh38
rs201208969 Chr.4: 87846381-87846381 on Build GRCh38
rs746322441 Chr.4: 87846326-87846326 on Build GRCh38
rs367826576 Chr.4: 87846435-87846435 on Build GRCh38
rs144955894 Chr.4: 87846535-87846535 on Build GRCh38
rs768970787 Chr.4: 87846338-87846338 on Build GRCh38
rs1049981075 Chr.4: 87846707-87846707 on Build GRCh38
rs571197309 Chr.4: 87846200-87846200 on Build GRCh38
rs754572700 Chr.4: 87846305-87846305 on Build GRCh38
rs61731018 Chr.4: 87846261-87846261 on Build GRCh38
rs776066868 Chr.4: 87846466-87846466 on Build GRCh38
rs371913298 Chr.4: 87846482-87846482 on Build GRCh38
rs947590658 Chr.4: 87846592-87846592 on Build GRCh38
rs574076245 Chr.4: 87846287-87846287 on Build GRCh38
rs761685582 Chr.4: 87846345-87846345 on Build GRCh38
rs767869973 Chr.4: 87846389-87846389 on Build GRCh38
rs4314251 Chr.4: 87846610-87846610 on Build GRCh38
rs934935162 Chr.4: 87846641-87846641 on Build GRCh38
rs747318796 Chr.4: 87846567-87846567 on Build GRCh38
rs1037104156 Chr.4: 87846576-87846576 on Build GRCh38
rs934273768 Chr.4: 87846719-87846719 on Build GRCh38
rs200104318 Chr.4: 87846483-87846483 on Build GRCh38
rs11732238 Chr.4: 87846564-87846564 on Build GRCh38
rs780040273 Chr.4: 87846396-87846396 on Build GRCh38
rs150220687 Chr.4: 87846225-87846225 on Build GRCh38
rs750770333 Chr.4: 87846469-87846469 on Build GRCh38

View Full Product Details