Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 52800519-52801098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA1
Entrez Gene ID
3672
Gene Name
integrin subunit alpha 1
Gene Aliases
CD49a, VLA1
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52788302-52953655 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Gene Symbol
PELO
Entrez Gene ID
53918
Gene Name
pelota homolog (Drosophila)
Gene Aliases
CGI-17, PRO1770
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52787940-52802617 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1186763 c.269A>C p.E90A Chr.5: 52800663-52800663 on Build GRCh38
6868346 c.696G>A p.L232L Chr.5: 52801090-52801090 on Build GRCh38
4347979 c.371G>C p.R124P Chr.5: 52800765-52800765 on Build GRCh38
1132152 c.622G>T p.A208S Chr.5: 52801016-52801016 on Build GRCh38
4141990 c.235T>A p.C79S Chr.5: 52800629-52800629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751137927 Chr.5: 52800592-52800592 on Build GRCh38
rs749483513 Chr.5: 52800633-52800633 on Build GRCh38
rs374003501 Chr.5: 52800716-52800716 on Build GRCh38
rs548469886 Chr.5: 52800752-52800752 on Build GRCh38
rs367564121 Chr.5: 52800637-52800637 on Build GRCh38
rs141211888 Chr.5: 52800652-52800652 on Build GRCh38
rs139161716 Chr.5: 52800769-52800769 on Build GRCh38
rs776377670 Chr.5: 52800995-52800995 on Build GRCh38
rs966918814 Chr.5: 52800670-52800670 on Build GRCh38
rs991725772 Chr.5: 52800807-52800807 on Build GRCh38
rs139977042 Chr.5: 52801050-52801050 on Build GRCh38
rs752511091 Chr.5: 52801049-52801049 on Build GRCh38
rs549701701 Chr.5: 52800918-52800918 on Build GRCh38
rs747488089 Chr.5: 52800678-52800678 on Build GRCh38
rs984090337 Chr.5: 52800978-52800978 on Build GRCh38
rs749484314 Chr.5: 52800543-52800543 on Build GRCh38
rs750442388 Chr.5: 52800610-52800610 on Build GRCh38
rs1054497597 Chr.5: 52800620-52800620 on Build GRCh38
rs779152067 Chr.5: 52800642-52800642 on Build GRCh38
rs771824545 Chr.5: 52800971-52800971 on Build GRCh38
rs777861421 Chr.5: 52800902-52800902 on Build GRCh38
rs951778516 Chr.5: 52800968-52800968 on Build GRCh38
rs753877060 Chr.5: 52800878-52800878 on Build GRCh38
rs201700908 Chr.5: 52800936-52800936 on Build GRCh38
rs750307879 Chr.5: 52800763-52800763 on Build GRCh38
rs771667455 Chr.5: 52800836-52800836 on Build GRCh38
rs971203469 Chr.5: 52800866-52800866 on Build GRCh38
rs773698128 Chr.5: 52800672-52800672 on Build GRCh38
rs762254838 Chr.5: 52800557-52800557 on Build GRCh38
rs559863446 Chr.5: 52800696-52800696 on Build GRCh38
rs762618064 Chr.5: 52800718-52800718 on Build GRCh38
rs150767441 Chr.5: 52800730-52800730 on Build GRCh38
rs761392908 Chr.5: 52800749-52800749 on Build GRCh38
rs755880743 Chr.5: 52800627-52800627 on Build GRCh38
rs144148189 Chr.5: 52800785-52800785 on Build GRCh38
rs747792288 Chr.5: 52801021-52801022 on Build GRCh38
rs752854138 Chr.5: 52800896-52800896 on Build GRCh38
rs754805559 Chr.5: 52800774-52800774 on Build GRCh38
rs758654730 Chr.5: 52800900-52800900 on Build GRCh38
rs781748090 Chr.5: 52800805-52800805 on Build GRCh38
rs772209348 Chr.5: 52800658-52800658 on Build GRCh38
rs770216010 Chr.5: 52800951-52800951 on Build GRCh38
rs775154862 Chr.5: 52800573-52800573 on Build GRCh38
rs200600121 Chr.5: 52800962-52800962 on Build GRCh38
rs776373330 Chr.5: 52800787-52800788 on Build GRCh38
rs774047496 Chr.5: 52800739-52800739 on Build GRCh38
rs368443275 Chr.5: 52800789-52800789 on Build GRCh38
rs766144124 Chr.5: 52800770-52800770 on Build GRCh38
rs753550497 Chr.5: 52800772-52800772 on Build GRCh38
rs907628725 Chr.5: 52801013-52801013 on Build GRCh38
rs764192082 Chr.5: 52801052-52801052 on Build GRCh38
rs751570929 Chr.5: 52800915-52800915 on Build GRCh38
rs765033407 Chr.5: 52800892-52800892 on Build GRCh38
rs760277990 Chr.5: 52800852-52800852 on Build GRCh38
rs747511590 Chr.5: 52800791-52800791 on Build GRCh38
rs756878110 Chr.5: 52800593-52800593 on Build GRCh38
rs760096987 Chr.5: 52800972-52800972 on Build GRCh38
rs571174898 Chr.5: 52800944-52800944 on Build GRCh38
rs983191126 Chr.5: 52801009-52801009 on Build GRCh38
rs766192469 Chr.5: 52800874-52800874 on Build GRCh38
rs771365583 Chr.5: 52800793-52800793 on Build GRCh38
rs746188540 Chr.5: 52800812-52800812 on Build GRCh38
rs770530262 Chr.5: 52800991-52800991 on Build GRCh38
rs1024183492 Chr.5: 52800833-52800833 on Build GRCh38
rs763658783 Chr.5: 52800581-52800581 on Build GRCh38
rs747754030 Chr.5: 52800965-52800965 on Build GRCh38
rs779717661 Chr.5: 52800631-52800631 on Build GRCh38
rs201423354 Chr.5: 52800950-52800950 on Build GRCh38
rs772615662 Chr.5: 52800839-52800839 on Build GRCh38
rs762516997 Chr.5: 52800578-52800578 on Build GRCh38
rs761560784 Chr.5: 52800824-52800824 on Build GRCh38
rs770338598 Chr.5: 52800813-52800813 on Build GRCh38
rs773751186 Chr.5: 52800567-52800567 on Build GRCh38
rs780308633 Chr.5: 52800541-52800541 on Build GRCh38
rs565843408 Chr.5: 52800996-52800996 on Build GRCh38
rs200636575 Chr.5: 52800779-52800779 on Build GRCh38
rs146042794 Chr.5: 52800957-52800957 on Build GRCh38
rs768714228 Chr.5: 52800545-52800545 on Build GRCh38
rs774188716 Chr.5: 52800732-52800732 on Build GRCh38
rs371706485 Chr.5: 52800697-52800697 on Build GRCh38
rs958659616 Chr.5: 52800773-52800773 on Build GRCh38
rs774441545 Chr.5: 52800556-52800556 on Build GRCh38
rs769542363 Chr.5: 52800539-52800539 on Build GRCh38
rs774088225 Chr.5: 52800818-52800818 on Build GRCh38
rs143871783 Chr.5: 52800811-52800812 on Build GRCh38
rs202063626 Chr.5: 52800788-52800788 on Build GRCh38
rs541189319 Chr.5: 52800564-52800564 on Build GRCh38
rs779106178 Chr.5: 52800778-52800778 on Build GRCh38
rs1499280 Chr.5: 52801055-52801055 on Build GRCh38
rs767556330 Chr.5: 52800607-52800607 on Build GRCh38
rs373725475 Chr.5: 52800570-52800570 on Build GRCh38
rs35160817 Chr.5: 52800841-52800841 on Build GRCh38
rs35749418 Chr.5: 52800798-52800798 on Build GRCh38
rs759316671 Chr.5: 52800881-52800881 on Build GRCh38
rs750569887 Chr.5: 52800827-52800827 on Build GRCh38

View Full Product Details