Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 11260389-11260971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRH1
Entrez Gene ID
3269
Gene Name
histamine receptor H1
Gene Aliases
H1-R, H1R, HH1R, hisH1
UniGene
Hs.1570
Gene Chromosome Location
Chr.3: 11137093-11263253 on Build GRCh38
Transcripts
NM_000861.3 NM_001098211.1 NM_001098212.1 NM_001098213.1 XM_011533652.1 XM_011533653.2 XM_017006283.1 XM_017006284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3914574 c.1364G>A p.W455* Chr.3: 11260401-11260401 on Build GRCh38
4112279 c.1359C>T p.T453T Chr.3: 11260396-11260396 on Build GRCh38
5708421 c.1399A>T p.I467F Chr.3: 11260436-11260436 on Build GRCh38
4602141 c.1460C>G p.S487C Chr.3: 11260497-11260497 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773496652 Chr.3: 11260818-11260818 on Build GRCh38
rs774068490 Chr.3: 11260464-11260464 on Build GRCh38
rs918538360 Chr.3: 11260824-11260824 on Build GRCh38
rs887900662 Chr.3: 11260564-11260564 on Build GRCh38
rs867306560 Chr.3: 11260636-11260636 on Build GRCh38
rs79733405 Chr.3: 11260899-11260899 on Build GRCh38
rs1018219554 Chr.3: 11260580-11260580 on Build GRCh38
rs988508372 Chr.3: 11260714-11260714 on Build GRCh38
rs546954000 Chr.3: 11260507-11260507 on Build GRCh38
rs749997385 Chr.3: 11260493-11260493 on Build GRCh38
rs929308157 Chr.3: 11260832-11260832 on Build GRCh38
rs906488844 Chr.3: 11260870-11260870 on Build GRCh38
rs945198556 Chr.3: 11260780-11260780 on Build GRCh38
rs767043272 Chr.3: 11260443-11260443 on Build GRCh38
rs201724478 Chr.3: 11260688-11260688 on Build GRCh38
rs558952658 Chr.3: 11260519-11260519 on Build GRCh38
rs751961466 Chr.3: 11260459-11260459 on Build GRCh38
rs1017531999 Chr.3: 11260666-11260666 on Build GRCh38
rs201054505 Chr.3: 11260794-11260794 on Build GRCh38
rs1005622610 Chr.3: 11260644-11260644 on Build GRCh38
rs201236680 Chr.3: 11260777-11260777 on Build GRCh38
rs200848971 Chr.3: 11260660-11260660 on Build GRCh38
rs570709265 Chr.3: 11260682-11260682 on Build GRCh38
rs202100549 Chr.3: 11260416-11260416 on Build GRCh38
rs200699665 Chr.3: 11260606-11260606 on Build GRCh38
rs199749810 Chr.3: 11260568-11260568 on Build GRCh38
rs202143972 Chr.3: 11260784-11260784 on Build GRCh38
rs200595957 Chr.3: 11260786-11260786 on Build GRCh38
rs111560166 Chr.3: 11260887-11260887 on Build GRCh38
rs763526390 Chr.3: 11260439-11260439 on Build GRCh38
rs756750763 Chr.3: 11260473-11260473 on Build GRCh38
rs992711920 Chr.3: 11260817-11260817 on Build GRCh38
rs201219261 Chr.3: 11260771-11260771 on Build GRCh38
rs200699265 Chr.3: 11260946-11260946 on Build GRCh38
rs201791741 Chr.3: 11260803-11260803 on Build GRCh38
rs200338344 Chr.3: 11260548-11260548 on Build GRCh38
rs572504650 Chr.3: 11260835-11260835 on Build GRCh38
rs773732133 Chr.3: 11260423-11260423 on Build GRCh38
rs78611023 Chr.3: 11260775-11260775 on Build GRCh38
rs201170181 Chr.3: 11260563-11260563 on Build GRCh38
rs758315820 Chr.3: 11260494-11260494 on Build GRCh38
rs1030578293 Chr.3: 11260901-11260901 on Build GRCh38
rs772449726 Chr.3: 11260723-11260723 on Build GRCh38
rs186758589 Chr.3: 11260837-11260837 on Build GRCh38
rs867224636 Chr.3: 11260604-11260604 on Build GRCh38
rs772839430 Chr.3: 11260499-11260499 on Build GRCh38
rs202180123 Chr.3: 11260774-11260774 on Build GRCh38
rs938563352 Chr.3: 11260785-11260788 on Build GRCh38
rs200664625 Chr.3: 11260885-11260885 on Build GRCh38
rs200067259 Chr.3: 11260691-11260691 on Build GRCh38
rs568627633 Chr.3: 11260752-11260752 on Build GRCh38
rs778254410 Chr.3: 11260492-11260492 on Build GRCh38
rs368468363 Chr.3: 11260794-11260795 on Build GRCh38
rs753454323 Chr.3: 11260471-11260471 on Build GRCh38
rs371113844 Chr.3: 11260645-11260645 on Build GRCh38
rs201060044 Chr.3: 11260639-11260639 on Build GRCh38
rs199865702 Chr.3: 11260676-11260676 on Build GRCh38
rs973397922 Chr.3: 11260677-11260677 on Build GRCh38
rs141806016 Chr.3: 11260785-11260786 on Build GRCh38
rs959745243 Chr.3: 11260617-11260617 on Build GRCh38
rs887856197 Chr.3: 11260861-11260861 on Build GRCh38
rs910199557 Chr.3: 11260738-11260738 on Build GRCh38
rs1021370937 Chr.3: 11260731-11260731 on Build GRCh38
rs892092279 Chr.3: 11260921-11260921 on Build GRCh38
rs770097130 Chr.3: 11260533-11260533 on Build GRCh38
rs748145095 Chr.3: 11260704-11260704 on Build GRCh38
rs201224954 Chr.3: 11260852-11260852 on Build GRCh38
rs199671400 Chr.3: 11260914-11260914 on Build GRCh38
rs532848763 Chr.3: 11260530-11260530 on Build GRCh38
rs1003579702 Chr.3: 11260556-11260556 on Build GRCh38
rs763185123 Chr.3: 11260553-11260553 on Build GRCh38
rs781430466 Chr.3: 11260522-11260522 on Build GRCh38
rs200608315 Chr.3: 11260683-11260683 on Build GRCh38
rs201855298 Chr.3: 11260672-11260672 on Build GRCh38
rs755461695 Chr.3: 11260461-11260461 on Build GRCh38
rs199509386 Chr.3: 11260869-11260869 on Build GRCh38
rs765258942 Chr.3: 11260411-11260411 on Build GRCh38
rs780030834 Chr.3: 11260503-11260503 on Build GRCh38
rs200339731 Chr.3: 11260826-11260826 on Build GRCh38
rs199642041 Chr.3: 11260651-11260651 on Build GRCh38
rs867228115 Chr.3: 11260496-11260496 on Build GRCh38
rs200819849 Chr.3: 11260900-11260900 on Build GRCh38
rs906542807 Chr.3: 11260524-11260524 on Build GRCh38
rs763773398 Chr.3: 11260467-11260467 on Build GRCh38
rs201664311 Chr.3: 11260909-11260909 on Build GRCh38
rs548266241 Chr.3: 11260541-11260541 on Build GRCh38
rs1009917677 Chr.3: 11260429-11260429 on Build GRCh38

View Full Product Details