Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143222454-143223022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAS2R40
Entrez Gene ID
259286
Gene Name
taste 2 receptor member 40
Gene Aliases
GPR60, T2R40, T2R58
UniGene
Hs.352241
Gene Chromosome Location
Chr.7: 143222079-143223050 on Build GRCh38
Transcripts
NM_176882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5062266 c.454G>A p.V152I Chr.7: 143222532-143222532 on Build GRCh38
3257959 c.749G>T p.S250I Chr.7: 143222827-143222827 on Build GRCh38
86806 c.543G>A p.T181T Chr.7: 143222621-143222621 on Build GRCh38
3257951 c.542C>A p.T181K Chr.7: 143222620-143222620 on Build GRCh38
745196 c.807C>G p.N269K Chr.7: 143222885-143222885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375632365 Chr.7: 143222964-143222964 on Build GRCh38
rs555941214 Chr.7: 143222965-143222965 on Build GRCh38
rs556788116 Chr.7: 143222529-143222529 on Build GRCh38
rs200388030 Chr.7: 143222902-143222902 on Build GRCh38
rs757878368 Chr.7: 143222774-143222774 on Build GRCh38
rs771250675 Chr.7: 143222678-143222678 on Build GRCh38
rs758507415 Chr.7: 143222876-143222876 on Build GRCh38
rs542480400 Chr.7: 143222550-143222550 on Build GRCh38
rs754034479 Chr.7: 143222579-143222579 on Build GRCh38
rs777249856 Chr.7: 143222880-143222880 on Build GRCh38
rs149217752 Chr.7: 143222949-143222949 on Build GRCh38
rs371379062 Chr.7: 143222489-143222489 on Build GRCh38
rs780208038 Chr.7: 143222933-143222933 on Build GRCh38
rs768942312 Chr.7: 143222941-143222941 on Build GRCh38
rs866076173 Chr.7: 143222994-143222994 on Build GRCh38
rs751419234 Chr.7: 143222518-143222518 on Build GRCh38
rs561103554 Chr.7: 143222554-143222554 on Build GRCh38
rs766645659 Chr.7: 143222919-143222919 on Build GRCh38
rs765172871 Chr.7: 143222824-143222824 on Build GRCh38
rs755951183 Chr.7: 143222621-143222621 on Build GRCh38
rs867642647 Chr.7: 143222914-143222914 on Build GRCh38
rs754960504 Chr.7: 143222483-143222483 on Build GRCh38
rs765436585 Chr.7: 143222688-143222688 on Build GRCh38
rs759661695 Chr.7: 143222684-143222684 on Build GRCh38
rs538618791 Chr.7: 143222522-143222522 on Build GRCh38
rs747626076 Chr.7: 143222650-143222650 on Build GRCh38
rs776867997 Chr.7: 143222793-143222793 on Build GRCh38
rs766352119 Chr.7: 143222858-143222858 on Build GRCh38
rs763043636 Chr.7: 143222700-143222700 on Build GRCh38
rs754589612 Chr.7: 143222767-143222767 on Build GRCh38
rs753609251 Chr.7: 143222758-143222758 on Build GRCh38
rs754576235 Chr.7: 143222636-143222636 on Build GRCh38
rs536576907 Chr.7: 143222563-143222563 on Build GRCh38
rs188336135 Chr.7: 143222895-143222895 on Build GRCh38
rs781240146 Chr.7: 143222604-143222604 on Build GRCh38
rs771340498 Chr.7: 143222783-143222783 on Build GRCh38
rs747344312 Chr.7: 143222948-143222948 on Build GRCh38
rs750268819 Chr.7: 143222620-143222620 on Build GRCh38
rs751095334 Chr.7: 143222978-143222978 on Build GRCh38
rs759699946 Chr.7: 143222582-143222582 on Build GRCh38
rs767613389 Chr.7: 143222709-143222709 on Build GRCh38
rs762212261 Chr.7: 143222559-143222559 on Build GRCh38
rs766264628 Chr.7: 143222747-143222747 on Build GRCh38
rs746205821 Chr.7: 143222802-143222802 on Build GRCh38
rs973319650 Chr.7: 143222698-143222698 on Build GRCh38
rs991254020 Chr.7: 143222954-143222954 on Build GRCh38
rs762983530 Chr.7: 143222812-143222812 on Build GRCh38
rs145138257 Chr.7: 143222961-143222961 on Build GRCh38
rs772598755 Chr.7: 143222657-143222657 on Build GRCh38
rs747508076 Chr.7: 143222778-143222778 on Build GRCh38
rs757139550 Chr.7: 143222520-143222520 on Build GRCh38
rs747419695 Chr.7: 143222671-143222671 on Build GRCh38
rs778588146 Chr.7: 143222769-143222769 on Build GRCh38
rs762604993 Chr.7: 143222966-143222966 on Build GRCh38
rs748890475 Chr.7: 143222627-143222627 on Build GRCh38
rs551713450 Chr.7: 143222943-143222943 on Build GRCh38
rs752767895 Chr.7: 143222601-143222601 on Build GRCh38
rs765224218 Chr.7: 143222590-143222590 on Build GRCh38
rs1040216068 Chr.7: 143222768-143222768 on Build GRCh38
rs1008617834 Chr.7: 143222485-143222485 on Build GRCh38
rs763699274 Chr.7: 143222976-143222976 on Build GRCh38
rs866945851 Chr.7: 143222484-143222484 on Build GRCh38
rs776236021 Chr.7: 143222680-143222681 on Build GRCh38
rs866765172 Chr.7: 143222909-143222909 on Build GRCh38
rs549191879 Chr.7: 143222962-143222962 on Build GRCh38
rs765772486 Chr.7: 143222978-143222978 on Build GRCh38
rs754753968 Chr.7: 143222866-143222866 on Build GRCh38
rs757148442 Chr.7: 143222602-143222602 on Build GRCh38
rs199846375 Chr.7: 143222827-143222827 on Build GRCh38
rs776608483 Chr.7: 143222958-143222958 on Build GRCh38
rs750354646 Chr.7: 143222719-143222719 on Build GRCh38
rs918945318 Chr.7: 143222982-143222982 on Build GRCh38
rs747633325 Chr.7: 143222770-143222770 on Build GRCh38
rs773659284 Chr.7: 143222658-143222658 on Build GRCh38
rs929188241 Chr.7: 143222717-143222717 on Build GRCh38
rs971506780 Chr.7: 143222599-143222599 on Build GRCh38
rs10260248 Chr.7: 143222638-143222638 on Build GRCh38
rs746272026 Chr.7: 143222913-143222913 on Build GRCh38
rs372647466 Chr.7: 143222776-143222776 on Build GRCh38
rs377107150 Chr.7: 143222813-143222813 on Build GRCh38
rs543619768 Chr.7: 143222847-143222847 on Build GRCh38
rs533197142 Chr.7: 143222939-143222939 on Build GRCh38
rs757966360 Chr.7: 143222879-143222879 on Build GRCh38
rs769936298 Chr.7: 143222810-143222810 on Build GRCh38
rs937834298 Chr.7: 143222695-143222695 on Build GRCh38
rs780417600 Chr.7: 143222878-143222878 on Build GRCh38
rs960171298 Chr.7: 143222951-143222951 on Build GRCh38
rs553641532 Chr.7: 143222927-143222927 on Build GRCh38
rs781594773 Chr.7: 143222983-143222983 on Build GRCh38
rs199540322 Chr.7: 143222546-143222546 on Build GRCh38
rs779804599 Chr.7: 143222542-143222542 on Build GRCh38
rs772571667 Chr.7: 143222560-143222560 on Build GRCh38
rs930215695 Chr.7: 143222575-143222575 on Build GRCh38
rs766607335 Chr.7: 143222565-143222565 on Build GRCh38
rs775466420 Chr.7: 143222696-143222696 on Build GRCh38
rs757343694 Chr.7: 143222687-143222687 on Build GRCh38
rs565742335 Chr.7: 143222623-143222625 on Build GRCh38
rs760133711 Chr.7: 143222898-143222899 on Build GRCh38
rs920543274 Chr.7: 143222713-143222713 on Build GRCh38
rs201465162 Chr.7: 143222561-143222561 on Build GRCh38
rs200620382 Chr.7: 143222984-143222984 on Build GRCh38
rs779892503 Chr.7: 143222622-143222622 on Build GRCh38
rs753604159 Chr.7: 143222862-143222862 on Build GRCh38
rs200656848 Chr.7: 143222979-143222979 on Build GRCh38
rs770164488 Chr.7: 143222932-143222932 on Build GRCh38
rs576272436 Chr.7: 143222680-143222680 on Build GRCh38
rs991658545 Chr.7: 143222536-143222536 on Build GRCh38
rs756042719 Chr.7: 143222722-143222722 on Build GRCh38

View Full Product Details