Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112588731-112589276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHCHD5
Entrez Gene ID
84269
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 5
Gene Aliases
C2orf9, MIC14
UniGene
Hs.375707
Gene Chromosome Location
Chr.2: 112583606-112589040 on Build GRCh38
Transcripts
NM_001304353.1 NM_001304354.1 NM_032309.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
295981 c.316C>T p.P106S Chr.2: 112588872-112588872 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771667091 Chr.2: 112588936-112588936 on Build GRCh38
rs747099296 Chr.2: 112588825-112588825 on Build GRCh38
rs115052296 Chr.2: 112589033-112589033 on Build GRCh38
rs1021588349 Chr.2: 112589158-112589158 on Build GRCh38
rs202075189 Chr.2: 112588892-112588892 on Build GRCh38
rs202174405 Chr.2: 112588935-112588935 on Build GRCh38
rs1055993972 Chr.2: 112589118-112589118 on Build GRCh38
rs558190195 Chr.2: 112588809-112588809 on Build GRCh38
rs765150422 Chr.2: 112588878-112588878 on Build GRCh38
rs3748024 Chr.2: 112588836-112588836 on Build GRCh38
rs41278942 Chr.2: 112588903-112588903 on Build GRCh38
rs374566266 Chr.2: 112588762-112588762 on Build GRCh38
rs767255769 Chr.2: 112588854-112588854 on Build GRCh38
rs941682258 Chr.2: 112588971-112588971 on Build GRCh38
rs969647337 Chr.2: 112589249-112589249 on Build GRCh38
rs571972235 Chr.2: 112589211-112589211 on Build GRCh38
rs918923749 Chr.2: 112589057-112589057 on Build GRCh38
rs750498889 Chr.2: 112588884-112588884 on Build GRCh38
rs369665974 Chr.2: 112588862-112588862 on Build GRCh38
rs775233413 Chr.2: 112588858-112588858 on Build GRCh38
rs892215043 Chr.2: 112589160-112589160 on Build GRCh38
rs1038488322 Chr.2: 112589063-112589063 on Build GRCh38
rs762637880 Chr.2: 112589016-112589016 on Build GRCh38
rs541992125 Chr.2: 112588752-112588752 on Build GRCh38
rs1022120879 Chr.2: 112589228-112589228 on Build GRCh38
rs759283730 Chr.2: 112588872-112588872 on Build GRCh38
rs1038542412 Chr.2: 112588923-112588923 on Build GRCh38
rs907483107 Chr.2: 112589096-112589096 on Build GRCh38
rs1047652 Chr.2: 112588935-112588935 on Build GRCh38
rs1003026306 Chr.2: 112589105-112589105 on Build GRCh38
rs894535968 Chr.2: 112589129-112589129 on Build GRCh38
rs542082686 Chr.2: 112589000-112589000 on Build GRCh38
rs1008980556 Chr.2: 112589119-112589119 on Build GRCh38
rs556810453 Chr.2: 112589068-112589068 on Build GRCh38
rs776478251 Chr.2: 112588941-112588941 on Build GRCh38
rs768246878 Chr.2: 112588940-112588940 on Build GRCh38
rs757202674 Chr.2: 112588869-112588869 on Build GRCh38
rs774372080 Chr.2: 112588883-112588892 on Build GRCh38
rs748528676 Chr.2: 112588916-112588916 on Build GRCh38
rs1051795114 Chr.2: 112589101-112589101 on Build GRCh38
rs375117258 Chr.2: 112588840-112588840 on Build GRCh38
rs759651882 Chr.2: 112588929-112588930 on Build GRCh38
rs1028638017 Chr.2: 112588933-112588933 on Build GRCh38
rs956489608 Chr.2: 112588983-112588983 on Build GRCh38
rs773908034 Chr.2: 112588841-112588841 on Build GRCh38
rs998992841 Chr.2: 112589097-112589097 on Build GRCh38
rs1011676433 Chr.2: 112589149-112589149 on Build GRCh38
rs369148167 Chr.2: 112588835-112588835 on Build GRCh38
rs770303560 Chr.2: 112588919-112588919 on Build GRCh38
rs758426876 Chr.2: 112588891-112588891 on Build GRCh38
rs534936292 Chr.2: 112588989-112588989 on Build GRCh38
rs969320689 Chr.2: 112589001-112589001 on Build GRCh38
rs768971304 Chr.2: 112588834-112588834 on Build GRCh38
rs770263831 Chr.2: 112588837-112588837 on Build GRCh38
rs910237031 Chr.2: 112588962-112588962 on Build GRCh38
rs930202229 Chr.2: 112589071-112589071 on Build GRCh38
rs530873620 Chr.2: 112588853-112588853 on Build GRCh38
rs35327779 Chr.2: 112589171-112589171 on Build GRCh38
rs371511842 Chr.2: 112588895-112588895 on Build GRCh38
rs576760342 Chr.2: 112589156-112589156 on Build GRCh38
rs201419705 Chr.2: 112588931-112588931 on Build GRCh38
rs766609621 Chr.2: 112588876-112588877 on Build GRCh38
rs765920790 Chr.2: 112589087-112589087 on Build GRCh38
rs1025931603 Chr.2: 112588796-112588796 on Build GRCh38
rs1047757103 Chr.2: 112589072-112589072 on Build GRCh38
rs776061121 Chr.2: 112589164-112589164 on Build GRCh38
rs558387720 Chr.2: 112588860-112588860 on Build GRCh38
rs75129286 Chr.2: 112588868-112588868 on Build GRCh38
rs375867530 Chr.2: 112588832-112588832 on Build GRCh38
rs760343828 Chr.2: 112588937-112588937 on Build GRCh38
rs1021704354 Chr.2: 112588930-112588930 on Build GRCh38
rs1010691675 Chr.2: 112589194-112589194 on Build GRCh38
rs981120868 Chr.2: 112589009-112589009 on Build GRCh38
rs1030715428 Chr.2: 112589121-112589121 on Build GRCh38
rs941482473 Chr.2: 112589059-112589059 on Build GRCh38
rs200858904 Chr.2: 112588902-112588902 on Build GRCh38
rs537150271 Chr.2: 112589058-112589058 on Build GRCh38
rs548356363 Chr.2: 112588990-112588990 on Build GRCh38

View Full Product Details