Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7729753-7730227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4G
Entrez Gene ID
339390
Gene Name
C-type lectin domain family 4 member G
Gene Aliases
DTTR431, LP2698, LSECtin, UNQ431
UniGene
Hs.220649
Gene Chromosome Location
Chr.19: 7728957-7732171 on Build GRCh38
Transcripts
NM_001244856.1 NM_198492.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1201361 c.679G>A p.A227T Chr.19: 7729885-7729885 on Build GRCh38
4749521 c.517G>A p.E173K Chr.19: 7730129-7730129 on Build GRCh38
6630203 c.497C>T p.P166L Chr.19: 7730149-7730149 on Build GRCh38
1248426 c.735C>T p.L245L Chr.19: 7729829-7729829 on Build GRCh38
4836263 c.636C>T p.L212L Chr.19: 7729928-7729928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777198188 Chr.19: 7729947-7729947 on Build GRCh38
rs368297239 Chr.19: 7730091-7730091 on Build GRCh38
rs149297990 Chr.19: 7730172-7730172 on Build GRCh38
rs781657449 Chr.19: 7730109-7730109 on Build GRCh38
rs746155614 Chr.19: 7729914-7729914 on Build GRCh38
rs761300902 Chr.19: 7729820-7729820 on Build GRCh38
rs375391912 Chr.19: 7729858-7729858 on Build GRCh38
rs962514316 Chr.19: 7729811-7729811 on Build GRCh38
rs760711933 Chr.19: 7730079-7730079 on Build GRCh38
rs756487039 Chr.19: 7730173-7730173 on Build GRCh38
rs759383324 Chr.19: 7729971-7729971 on Build GRCh38
rs775952840 Chr.19: 7729841-7729841 on Build GRCh38
rs752741607 Chr.19: 7729985-7729985 on Build GRCh38
rs778243626 Chr.19: 7730183-7730183 on Build GRCh38
rs866056649 Chr.19: 7729840-7729840 on Build GRCh38
rs368273649 Chr.19: 7729894-7729894 on Build GRCh38
rs772662136 Chr.19: 7730040-7730040 on Build GRCh38
rs1014143982 Chr.19: 7730175-7730175 on Build GRCh38
rs769076170 Chr.19: 7730015-7730015 on Build GRCh38
rs746180361 Chr.19: 7729809-7729809 on Build GRCh38
rs748756421 Chr.19: 7730035-7730035 on Build GRCh38
rs773992274 Chr.19: 7730143-7730143 on Build GRCh38
rs750090061 Chr.19: 7729870-7729870 on Build GRCh38
rs138871018 Chr.19: 7729955-7729955 on Build GRCh38
rs555610194 Chr.19: 7730106-7730106 on Build GRCh38
rs1001253846 Chr.19: 7729973-7729973 on Build GRCh38
rs1054195473 Chr.19: 7729978-7729978 on Build GRCh38
rs776721083 Chr.19: 7730157-7730157 on Build GRCh38
rs375205366 Chr.19: 7729953-7729953 on Build GRCh38
rs770985333 Chr.19: 7729915-7729915 on Build GRCh38
rs200560949 Chr.19: 7730209-7730209 on Build GRCh38
rs370327157 Chr.19: 7730002-7730002 on Build GRCh38
rs750648603 Chr.19: 7729995-7729995 on Build GRCh38
rs373307374 Chr.19: 7730009-7730009 on Build GRCh38
rs766631531 Chr.19: 7730160-7730160 on Build GRCh38
rs764728666 Chr.19: 7729864-7729864 on Build GRCh38
rs769164537 Chr.19: 7730038-7730038 on Build GRCh38
rs756574920 Chr.19: 7730126-7730126 on Build GRCh38
rs566389972 Chr.19: 7730145-7730145 on Build GRCh38
rs752542530 Chr.19: 7730064-7730064 on Build GRCh38
rs748483954 Chr.19: 7730125-7730125 on Build GRCh38
rs770488586 Chr.19: 7730139-7730139 on Build GRCh38
rs767302020 Chr.19: 7729977-7729977 on Build GRCh38
rs775302612 Chr.19: 7730151-7730151 on Build GRCh38
rs750487345 Chr.19: 7730086-7730086 on Build GRCh38
rs1010346889 Chr.19: 7730185-7730185 on Build GRCh38
rs966080755 Chr.19: 7729780-7729780 on Build GRCh38
rs757376436 Chr.19: 7729909-7729909 on Build GRCh38
rs780612239 Chr.19: 7729935-7729935 on Build GRCh38
rs761440960 Chr.19: 7729947-7729947 on Build GRCh38
rs749262496 Chr.19: 7729907-7729907 on Build GRCh38
rs751865095 Chr.19: 7730104-7730104 on Build GRCh38
rs772821573 Chr.19: 7729960-7729960 on Build GRCh38
rs767665694 Chr.19: 7730166-7730166 on Build GRCh38
rs758642828 Chr.19: 7729997-7729997 on Build GRCh38
rs778258404 Chr.19: 7730132-7730132 on Build GRCh38
rs749863098 Chr.19: 7730191-7730191 on Build GRCh38
rs779171175 Chr.19: 7729991-7729991 on Build GRCh38
rs756733681 Chr.19: 7730195-7730195 on Build GRCh38
rs755178489 Chr.19: 7730164-7730164 on Build GRCh38
rs771421057 Chr.19: 7730007-7730007 on Build GRCh38
rs757283246 Chr.19: 7729989-7729989 on Build GRCh38
rs781537650 Chr.19: 7730030-7730030 on Build GRCh38
rs760415857 Chr.19: 7730154-7730154 on Build GRCh38
rs763109759 Chr.19: 7730159-7730159 on Build GRCh38
rs73493927 Chr.19: 7729945-7729945 on Build GRCh38
rs758185253 Chr.19: 7729873-7729873 on Build GRCh38
rs370720246 Chr.19: 7730063-7730063 on Build GRCh38
rs891996050 Chr.19: 7730176-7730176 on Build GRCh38
rs565957284 Chr.19: 7730137-7730137 on Build GRCh38
rs1020240141 Chr.19: 7730026-7730026 on Build GRCh38
rs201996252 Chr.19: 7730083-7730083 on Build GRCh38
rs1010535611 Chr.19: 7730004-7730004 on Build GRCh38
rs879152094 Chr.19: 7729990-7729990 on Build GRCh38
rs745578277 Chr.19: 7730148-7730148 on Build GRCh38
rs762483347 Chr.19: 7730042-7730042 on Build GRCh38
rs150770740 Chr.19: 7729861-7729861 on Build GRCh38
rs199503373 Chr.19: 7730056-7730056 on Build GRCh38
rs766044275 Chr.19: 7729965-7729965 on Build GRCh38
rs139161592 Chr.19: 7729880-7729880 on Build GRCh38
rs771022273 Chr.19: 7729786-7729786 on Build GRCh38
rs762643925 Chr.19: 7729964-7729964 on Build GRCh38
rs771737816 Chr.19: 7730149-7730149 on Build GRCh38
rs772952991 Chr.19: 7730187-7730188 on Build GRCh38
rs763635364 Chr.19: 7729821-7729821 on Build GRCh38
rs751477877 Chr.19: 7729892-7729892 on Build GRCh38
rs747396861 Chr.19: 7729942-7729942 on Build GRCh38
rs376581873 Chr.19: 7730052-7730052 on Build GRCh38
rs774639838 Chr.19: 7729787-7729787 on Build GRCh38
rs369759693 Chr.19: 7729968-7729968 on Build GRCh38
rs778992697 Chr.19: 7729910-7729910 on Build GRCh38
rs747269516 Chr.19: 7730014-7730014 on Build GRCh38

View Full Product Details