Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115806297-115806702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDO1
Entrez Gene ID
1036
Gene Name
cysteine dioxygenase type 1
Gene Aliases
CDO-I
UniGene
Hs.442378
Gene Chromosome Location
Chr.5: 115804733-115816708 on Build GRCh38
Transcripts
NM_001323565.1 NM_001323566.1 NM_001323567.1 NM_001801.2 NR_136618.1 NR_136619.1 NR_136620.1 NR_136621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6851878 c.517C>T p.H173Y Chr.5: 115806405-115806405 on Build GRCh38
5599282 c.479C>T p.P160L Chr.5: 115806443-115806443 on Build GRCh38
32394 c.427G>C p.E143Q Chr.5: 115806495-115806495 on Build GRCh38
3607713 c.445G>A p.E149K Chr.5: 115806477-115806477 on Build GRCh38
1696062 c.569C>T p.P190L Chr.5: 115806353-115806353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201520905 Chr.5: 115806534-115806534 on Build GRCh38
rs916666511 Chr.5: 115806610-115806610 on Build GRCh38
rs1052124630 Chr.5: 115806321-115806321 on Build GRCh38
rs757126802 Chr.5: 115806425-115806425 on Build GRCh38
rs764849319 Chr.5: 115806471-115806471 on Build GRCh38
rs764049604 Chr.5: 115806421-115806421 on Build GRCh38
rs866899471 Chr.5: 115806663-115806663 on Build GRCh38
rs752381626 Chr.5: 115806473-115806473 on Build GRCh38
rs776810485 Chr.5: 115806464-115806464 on Build GRCh38
rs17852213 Chr.5: 115806513-115806513 on Build GRCh38
rs772121757 Chr.5: 115806443-115806443 on Build GRCh38
rs536229174 Chr.5: 115806479-115806479 on Build GRCh38
rs113616899 Chr.5: 115806358-115806358 on Build GRCh38
rs927860445 Chr.5: 115806655-115806655 on Build GRCh38
rs369839442 Chr.5: 115806531-115806531 on Build GRCh38
rs777175829 Chr.5: 115806547-115806547 on Build GRCh38
rs749865216 Chr.5: 115806469-115806469 on Build GRCh38
rs1007073801 Chr.5: 115806458-115806458 on Build GRCh38
rs567449443 Chr.5: 115806450-115806450 on Build GRCh38
rs754717913 Chr.5: 115806543-115806546 on Build GRCh38
rs766725666 Chr.5: 115806331-115806331 on Build GRCh38
rs373296506 Chr.5: 115806532-115806532 on Build GRCh38
rs746276329 Chr.5: 115806557-115806557 on Build GRCh38
rs186288688 Chr.5: 115806335-115806335 on Build GRCh38
rs748221907 Chr.5: 115806434-115806434 on Build GRCh38
rs981923828 Chr.5: 115806654-115806654 on Build GRCh38
rs753528832 Chr.5: 115806433-115806433 on Build GRCh38
rs969159971 Chr.5: 115806625-115806625 on Build GRCh38
rs1024229129 Chr.5: 115806634-115806634 on Build GRCh38
rs1025674550 Chr.5: 115806521-115806521 on Build GRCh38
rs1010683107 Chr.5: 115806404-115806404 on Build GRCh38
rs143367368 Chr.5: 115806428-115806428 on Build GRCh38
rs372817136 Chr.5: 115806576-115806576 on Build GRCh38
rs747535035 Chr.5: 115806543-115806543 on Build GRCh38
rs781078499 Chr.5: 115806426-115806426 on Build GRCh38
rs751558879 Chr.5: 115806486-115806486 on Build GRCh38
rs376126377 Chr.5: 115806346-115806346 on Build GRCh38
rs200372326 Chr.5: 115806512-115806512 on Build GRCh38
rs375188135 Chr.5: 115806384-115806384 on Build GRCh38
rs200935045 Chr.5: 115806452-115806452 on Build GRCh38
rs146759585 Chr.5: 115806456-115806456 on Build GRCh38
rs762619116 Chr.5: 115806478-115806478 on Build GRCh38
rs371359579 Chr.5: 115806412-115806412 on Build GRCh38
rs143396814 Chr.5: 115806398-115806398 on Build GRCh38
rs771469569 Chr.5: 115806545-115806545 on Build GRCh38
rs764835868 Chr.5: 115806377-115806377 on Build GRCh38
rs957104850 Chr.5: 115806661-115806661 on Build GRCh38
rs753492426 Chr.5: 115806332-115806332 on Build GRCh38
rs756802735 Chr.5: 115806439-115806439 on Build GRCh38
rs375963286 Chr.5: 115806500-115806500 on Build GRCh38
rs781161372 Chr.5: 115806501-115806501 on Build GRCh38
rs977242936 Chr.5: 115806638-115806638 on Build GRCh38

View Full Product Details