Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44813733-44814132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD19
Entrez Gene ID
149478
Gene Name
BTB domain containing 19
Gene Aliases
-
UniGene
Hs.632400
Gene Chromosome Location
Chr.1: 44807442-44814199 on Build GRCh38
Transcripts
NM_001136537.1 XM_006710386.3 XM_011540803.2 XM_017000447.1 XM_017000448.1 XM_017000449.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7200437 c.840G>A p.R280R Chr.1: 44813736-44813736 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776944820 Chr.1: 44813769-44813769 on Build GRCh38
rs11584031 Chr.1: 44814044-44814044 on Build GRCh38
rs998838187 Chr.1: 44814001-44814001 on Build GRCh38
rs988450326 Chr.1: 44813898-44813898 on Build GRCh38
rs539782439 Chr.1: 44813971-44813971 on Build GRCh38
rs1034498225 Chr.1: 44814108-44814108 on Build GRCh38
rs913108930 Chr.1: 44813913-44813913 on Build GRCh38
rs541530291 Chr.1: 44814031-44814031 on Build GRCh38
rs981415842 Chr.1: 44813931-44813931 on Build GRCh38
rs1030243297 Chr.1: 44813850-44813850 on Build GRCh38
rs941306501 Chr.1: 44813972-44813972 on Build GRCh38
rs1028199152 Chr.1: 44813839-44813839 on Build GRCh38
rs552343595 Chr.1: 44813782-44813782 on Build GRCh38
rs773033431 Chr.1: 44813917-44813917 on Build GRCh38
rs897195993 Chr.1: 44813988-44813988 on Build GRCh38
rs980261304 Chr.1: 44813939-44813939 on Build GRCh38
rs889934163 Chr.1: 44814032-44814032 on Build GRCh38
rs1033081259 Chr.1: 44813860-44813860 on Build GRCh38
rs1037256238 Chr.1: 44813795-44813795 on Build GRCh38
rs117344385 Chr.1: 44813976-44813976 on Build GRCh38
rs370243473 Chr.1: 44813995-44813995 on Build GRCh38
rs907884294 Chr.1: 44813982-44813983 on Build GRCh38
rs370918173 Chr.1: 44813907-44813907 on Build GRCh38
rs532714371 Chr.1: 44813777-44813777 on Build GRCh38
rs888429648 Chr.1: 44813775-44813775 on Build GRCh38
rs1000505513 Chr.1: 44814020-44814020 on Build GRCh38
rs1005572989 Chr.1: 44813780-44813780 on Build GRCh38
rs1051381842 Chr.1: 44814012-44814012 on Build GRCh38
rs568331873 Chr.1: 44813848-44813848 on Build GRCh38
rs971718150 Chr.1: 44813925-44813925 on Build GRCh38
rs544173081 Chr.1: 44814102-44814102 on Build GRCh38
rs373795892 Chr.1: 44813844-44813844 on Build GRCh38
rs954272840 Chr.1: 44813854-44813854 on Build GRCh38
rs1040434822 Chr.1: 44813784-44813784 on Build GRCh38
rs368986942 Chr.1: 44813861-44813861 on Build GRCh38
rs1049543415 Chr.1: 44814013-44814013 on Build GRCh38
rs868350119 Chr.1: 44813805-44813805 on Build GRCh38
rs1032695034 Chr.1: 44814061-44814061 on Build GRCh38
rs988987630 Chr.1: 44813885-44813885 on Build GRCh38
rs565942881 Chr.1: 44813821-44813821 on Build GRCh38
rs1024099486 Chr.1: 44814078-44814078 on Build GRCh38
rs889731632 Chr.1: 44814013-44814014 on Build GRCh38
rs901027146 Chr.1: 44813789-44813789 on Build GRCh38
rs371648956 Chr.1: 44814081-44814085 on Build GRCh38
rs1032747726 Chr.1: 44813868-44813868 on Build GRCh38
rs1055189677 Chr.1: 44813962-44813962 on Build GRCh38
rs927105760 Chr.1: 44813936-44813936 on Build GRCh38
rs537463533 Chr.1: 44813944-44813944 on Build GRCh38
rs201181299 Chr.1: 44814078-44814082 on Build GRCh38
rs901897460 Chr.1: 44814000-44814000 on Build GRCh38
rs575278064 Chr.1: 44814087-44814087 on Build GRCh38
rs762061356 Chr.1: 44813783-44813783 on Build GRCh38
rs940672280 Chr.1: 44813987-44813987 on Build GRCh38
rs570753256 Chr.1: 44813947-44813947 on Build GRCh38
rs554796209 Chr.1: 44813847-44813847 on Build GRCh38
rs930868548 Chr.1: 44813972-44813972 on Build GRCh38
rs573076170 Chr.1: 44813994-44813994 on Build GRCh38
rs957012516 Chr.1: 44813897-44813897 on Build GRCh38

View Full Product Details