Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3629418-3629917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPRG1L
Entrez Gene ID
127262
Gene Name
tumor protein p63 regulated 1-like
Gene Aliases
FAM79A, h-mover, mover
UniGene
Hs.20529
Gene Chromosome Location
Chr.1: 3624992-3630131 on Build GRCh38
Transcripts
NM_182752.3

SNP Detail

SNP ID Chromosome Location
rs397862385 Chr.1: 3629704-3629704 on Build GRCh38
rs748470496 Chr.1: 3629702-3629705 on Build GRCh38
rs887750425 Chr.1: 3629804-3629804 on Build GRCh38
rs903418222 Chr.1: 3629505-3629505 on Build GRCh38
rs28712325 Chr.1: 3629655-3629655 on Build GRCh38
rs1006636678 Chr.1: 3629819-3629819 on Build GRCh38
rs527263177 Chr.1: 3629517-3629517 on Build GRCh38
rs990699730 Chr.1: 3629504-3629504 on Build GRCh38
rs554284659 Chr.1: 3629821-3629821 on Build GRCh38
rs923641758 Chr.1: 3629861-3629861 on Build GRCh38
rs55905548 Chr.1: 3629703-3629706 on Build GRCh38
rs959308922 Chr.1: 3629495-3629495 on Build GRCh38
rs924947051 Chr.1: 3629688-3629688 on Build GRCh38
rs397861350 Chr.1: 3629703-3629703 on Build GRCh38
rs894771713 Chr.1: 3629708-3629708 on Build GRCh38
rs549706617 Chr.1: 3629637-3629637 on Build GRCh38
rs184699239 Chr.1: 3629684-3629684 on Build GRCh38
rs966549977 Chr.1: 3629889-3629889 on Build GRCh38
rs975898940 Chr.1: 3629854-3629854 on Build GRCh38
rs769899053 Chr.1: 3629767-3629767 on Build GRCh38
rs35021450 Chr.1: 3629702-3629702 on Build GRCh38
rs971490038 Chr.1: 3629585-3629585 on Build GRCh38
rs570410709 Chr.1: 3629524-3629524 on Build GRCh38
rs182198072 Chr.1: 3629516-3629516 on Build GRCh38
rs964656764 Chr.1: 3629832-3629832 on Build GRCh38
rs1026338459 Chr.1: 3629632-3629632 on Build GRCh38
rs923703206 Chr.1: 3629528-3629528 on Build GRCh38
rs927824553 Chr.1: 3629443-3629443 on Build GRCh38
rs1024343005 Chr.1: 3629554-3629554 on Build GRCh38
rs1022194600 Chr.1: 3629770-3629770 on Build GRCh38
rs144580928 Chr.1: 3629865-3629865 on Build GRCh38
rs993447490 Chr.1: 3629586-3629586 on Build GRCh38
rs386627766 Chr.1: 3629683-3629684 on Build GRCh38
rs1045906047 Chr.1: 3629741-3629741 on Build GRCh38
rs71813640 Chr.1: 3629564-3629567 on Build GRCh38
rs746647584 Chr.1: 3629700-3629701 on Build GRCh38
rs201769672 Chr.1: 3629561-3629564 on Build GRCh38
rs563976292 Chr.1: 3629793-3629793 on Build GRCh38
rs137965653 Chr.1: 3629701-3629704 on Build GRCh38
rs769006842 Chr.1: 3629890-3629890 on Build GRCh38
rs530112481 Chr.1: 3629669-3629669 on Build GRCh38
rs948883384 Chr.1: 3629735-3629735 on Build GRCh38
rs1004826460 Chr.1: 3629671-3629671 on Build GRCh38
rs192366199 Chr.1: 3629683-3629683 on Build GRCh38
rs984695427 Chr.1: 3629638-3629638 on Build GRCh38
rs1017517923 Chr.1: 3629826-3629826 on Build GRCh38
rs138757642 Chr.1: 3629562-3629565 on Build GRCh38
rs936344635 Chr.1: 3629691-3629691 on Build GRCh38
rs768220367 Chr.1: 3629561-3629562 on Build GRCh38
rs575669666 Chr.1: 3629820-3629820 on Build GRCh38
rs900703183 Chr.1: 3629558-3629558 on Build GRCh38
rs976203494 Chr.1: 3629887-3629887 on Build GRCh38
rs1054677074 Chr.1: 3629701-3629701 on Build GRCh38
rs202040049 Chr.1: 3629703-3629703 on Build GRCh38
rs923006619 Chr.1: 3629681-3629681 on Build GRCh38
rs907279857 Chr.1: 3629742-3629742 on Build GRCh38
rs959426169 Chr.1: 3629714-3629714 on Build GRCh38
rs201107120 Chr.1: 3629564-3629564 on Build GRCh38
rs547471002 Chr.1: 3629518-3629518 on Build GRCh38
rs533902494 Chr.1: 3629773-3629773 on Build GRCh38
rs186800166 Chr.1: 3629670-3629670 on Build GRCh38
rs34755492 Chr.1: 3629701-3629701 on Build GRCh38
rs539658574 Chr.1: 3629553-3629553 on Build GRCh38
rs138172462 Chr.1: 3629461-3629461 on Build GRCh38
rs199881245 Chr.1: 3629704-3629704 on Build GRCh38

View Full Product Details