Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 30968051-30968557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUM1
Entrez Gene ID
9698
Gene Name
pumilio RNA binding family member 1
Gene Aliases
HSPUM, PUMH, PUMH1, PUML1
UniGene
Hs.281707
Gene Chromosome Location
Chr.1: 30931506-31065717 on Build GRCh38
Transcripts
NM_001020658.1 NM_014676.2 NR_003066.1

Gene Symbol
SNORD103C
Entrez Gene ID
692200
Gene Name
small nucleolar RNA, C/D box 103C
Gene Aliases
HBII-251, SNORD85
UniGene
Gene Chromosome Location
Chr.1: 30968163-30968237 on Build GRCh38
Transcripts
NM_001020658.1 NM_014676.2 NR_003066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2077028 c.1529A>G p.Q510R Chr.1: 30968470-30968470 on Build GRCh38
3865279 c.1576C>T p.P526S Chr.1: 30968423-30968423 on Build GRCh38
5502198 c.1561G>A p.G521R Chr.1: 30968438-30968438 on Build GRCh38
6760321 c.1513C>A p.R505S Chr.1: 30968486-30968486 on Build GRCh38
2077027 c.1543C>G p.P515A Chr.1: 30968456-30968456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763482843 Chr.1: 30968190-30968190 on Build GRCh38
rs756762964 Chr.1: 30968350-30968350 on Build GRCh38
rs757134251 Chr.1: 30968457-30968457 on Build GRCh38
rs966895496 Chr.1: 30968393-30968393 on Build GRCh38
rs761820434 Chr.1: 30968308-30968308 on Build GRCh38
rs143359453 Chr.1: 30968428-30968428 on Build GRCh38
rs762800421 Chr.1: 30968118-30968118 on Build GRCh38
rs753360228 Chr.1: 30968334-30968334 on Build GRCh38
rs375871818 Chr.1: 30968179-30968179 on Build GRCh38
rs1037178676 Chr.1: 30968277-30968277 on Build GRCh38
rs377130586 Chr.1: 30968276-30968276 on Build GRCh38
rs776727881 Chr.1: 30968116-30968116 on Build GRCh38
rs755725967 Chr.1: 30968188-30968188 on Build GRCh38
rs560030696 Chr.1: 30968247-30968247 on Build GRCh38
rs139411220 Chr.1: 30968439-30968439 on Build GRCh38
rs201060525 Chr.1: 30968259-30968259 on Build GRCh38
rs758932012 Chr.1: 30968379-30968379 on Build GRCh38
rs766281229 Chr.1: 30968119-30968119 on Build GRCh38
rs1033688164 Chr.1: 30968324-30968324 on Build GRCh38
rs1003580208 Chr.1: 30968298-30968298 on Build GRCh38
rs747316928 Chr.1: 30968402-30968402 on Build GRCh38
rs1022441224 Chr.1: 30968335-30968335 on Build GRCh38
rs767196030 Chr.1: 30968181-30968181 on Build GRCh38
rs779340599 Chr.1: 30968364-30968364 on Build GRCh38
rs202191174 Chr.1: 30968469-30968469 on Build GRCh38
rs1011241061 Chr.1: 30968451-30968451 on Build GRCh38
rs112422525 Chr.1: 30968223-30968223 on Build GRCh38
rs895191600 Chr.1: 30968326-30968326 on Build GRCh38
rs753509889 Chr.1: 30968499-30968499 on Build GRCh38
rs775945456 Chr.1: 30968224-30968224 on Build GRCh38
rs182081080 Chr.1: 30968527-30968527 on Build GRCh38
rs776608978 Chr.1: 30968196-30968197 on Build GRCh38
rs918755289 Chr.1: 30968282-30968282 on Build GRCh38
rs369003742 Chr.1: 30968284-30968284 on Build GRCh38
rs4317854 Chr.1: 30968074-30968074 on Build GRCh38
rs906524615 Chr.1: 30968281-30968282 on Build GRCh38
rs769056688 Chr.1: 30968423-30968423 on Build GRCh38
rs149265040 Chr.1: 30968367-30968367 on Build GRCh38
rs755777323 Chr.1: 30968316-30968316 on Build GRCh38
rs200399390 Chr.1: 30968168-30968168 on Build GRCh38
rs200987656 Chr.1: 30968465-30968465 on Build GRCh38
rs184759162 Chr.1: 30968242-30968242 on Build GRCh38
rs752422974 Chr.1: 30968304-30968304 on Build GRCh38
rs188871885 Chr.1: 30968509-30968509 on Build GRCh38
rs779473203 Chr.1: 30968213-30968213 on Build GRCh38
rs369295704 Chr.1: 30968275-30968275 on Build GRCh38
rs374392777 Chr.1: 30968198-30968198 on Build GRCh38
rs752369977 Chr.1: 30968187-30968187 on Build GRCh38
rs757766804 Chr.1: 30968202-30968202 on Build GRCh38
rs551662997 Chr.1: 30968485-30968485 on Build GRCh38
rs1001516832 Chr.1: 30968392-30968392 on Build GRCh38
rs768929685 Chr.1: 30968240-30968240 on Build GRCh38
rs910238500 Chr.1: 30968514-30968518 on Build GRCh38
rs529357214 Chr.1: 30968344-30968344 on Build GRCh38
rs572824735 Chr.1: 30968081-30968081 on Build GRCh38
rs372534831 Chr.1: 30968228-30968228 on Build GRCh38
rs987117858 Chr.1: 30968470-30968470 on Build GRCh38
rs759476398 Chr.1: 30968440-30968440 on Build GRCh38
rs527309319 Chr.1: 30968155-30968155 on Build GRCh38
rs541335871 Chr.1: 30968096-30968096 on Build GRCh38
rs780567159 Chr.1: 30968388-30968388 on Build GRCh38
rs1011012489 Chr.1: 30968327-30968327 on Build GRCh38
rs771939648 Chr.1: 30968449-30968449 on Build GRCh38
rs1045829009 Chr.1: 30968366-30968366 on Build GRCh38
rs781471570 Chr.1: 30968427-30968427 on Build GRCh38
rs763792726 Chr.1: 30968324-30968324 on Build GRCh38
rs769894521 Chr.1: 30968436-30968436 on Build GRCh38
rs369974872 Chr.1: 30968218-30968218 on Build GRCh38

View Full Product Details