Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33077462-33077957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00294
Entrez Gene ID
283267
Gene Name
long intergenic non-protein coding RNA 294
Gene Aliases
NCRNA00294
UniGene
Hs.533701
Gene Chromosome Location
Chr.11: 33076150-33079454 on Build GRCh38
Transcripts
NR_015451.1 XM_011520204.2 XM_011520205.2 XM_011520206.2 XM_011520207.2 XM_017017989.1 XM_017017990.1 XM_017017991.1 XM_017017992.1

Gene Symbol
TCP11L1
Entrez Gene ID
55346
Gene Name
t-complex 11 like 1
Gene Aliases
dJ85M6.3
UniGene
Hs.655341
Gene Chromosome Location
Chr.11: 33039417-33079454 on Build GRCh38
Transcripts
NR_015451.1 XM_011520204.2 XM_011520205.2 XM_011520206.2 XM_011520207.2 XM_017017989.1 XM_017017990.1 XM_017017991.1 XM_017017992.1

SNP Detail

SNP ID Chromosome Location
rs921064949 Chr.11: 33077585-33077585 on Build GRCh38
rs4756561 Chr.11: 33077909-33077909 on Build GRCh38
rs1048665326 Chr.11: 33077910-33077910 on Build GRCh38
rs185407650 Chr.11: 33077713-33077713 on Build GRCh38
rs969322949 Chr.11: 33077830-33077830 on Build GRCh38
rs913597286 Chr.11: 33077599-33077599 on Build GRCh38
rs937724020 Chr.11: 33077782-33077782 on Build GRCh38
rs879768248 Chr.11: 33077716-33077716 on Build GRCh38
rs1051968623 Chr.11: 33077625-33077625 on Build GRCh38
rs538418891 Chr.11: 33077546-33077546 on Build GRCh38
rs560027126 Chr.11: 33077862-33077862 on Build GRCh38
rs192748892 Chr.11: 33077606-33077606 on Build GRCh38
rs546394989 Chr.11: 33077848-33077848 on Build GRCh38
rs549426313 Chr.11: 33077495-33077495 on Build GRCh38
rs1023298409 Chr.11: 33077812-33077812 on Build GRCh38
rs189037597 Chr.11: 33077792-33077792 on Build GRCh38
rs559120585 Chr.11: 33077545-33077545 on Build GRCh38
rs146137140 Chr.11: 33077711-33077711 on Build GRCh38
rs577532843 Chr.11: 33077833-33077833 on Build GRCh38
rs112460229 Chr.11: 33077636-33077636 on Build GRCh38
rs552081880 Chr.11: 33077595-33077595 on Build GRCh38
rs1034919996 Chr.11: 33077871-33077871 on Build GRCh38
rs987958437 Chr.11: 33077885-33077885 on Build GRCh38
rs965430589 Chr.11: 33077544-33077544 on Build GRCh38
rs879823506 Chr.11: 33077906-33077906 on Build GRCh38
rs1005021813 Chr.11: 33077656-33077656 on Build GRCh38
rs947398750 Chr.11: 33077494-33077494 on Build GRCh38
rs148122063 Chr.11: 33077541-33077541 on Build GRCh38
rs554661564 Chr.11: 33077702-33077702 on Build GRCh38
rs956760248 Chr.11: 33077883-33077883 on Build GRCh38
rs958016932 Chr.11: 33077588-33077588 on Build GRCh38
rs148480497 Chr.11: 33077635-33077635 on Build GRCh38
rs142934292 Chr.11: 33077697-33077697 on Build GRCh38
rs74234036 Chr.11: 33077935-33077935 on Build GRCh38
rs557455014 Chr.11: 33077783-33077783 on Build GRCh38
rs945055220 Chr.11: 33077698-33077698 on Build GRCh38

View Full Product Details