Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51815536-51816042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00282
Entrez Gene ID
283521
Gene Name
long intergenic non-protein coding RNA 282
Gene Aliases
NCRNA00282
UniGene
Hs.649896
Gene Chromosome Location
Chr.13: 51813347-51845150 on Build GRCh38
Transcripts
NR_027047.1 NR_027048.1

SNP Detail

SNP ID Chromosome Location
rs781343576 Chr.13: 51815670-51815671 on Build GRCh38
rs1038242726 Chr.13: 51815722-51815722 on Build GRCh38
rs9316557 Chr.13: 51815607-51815607 on Build GRCh38
rs898316773 Chr.13: 51815728-51815728 on Build GRCh38
rs867806148 Chr.13: 51815899-51815899 on Build GRCh38
rs747019766 Chr.13: 51815921-51815921 on Build GRCh38
rs149195981 Chr.13: 51815809-51815809 on Build GRCh38
rs558740778 Chr.13: 51815952-51815952 on Build GRCh38
rs547081528 Chr.13: 51815815-51815815 on Build GRCh38
rs1030011317 Chr.13: 51815608-51815608 on Build GRCh38
rs916848210 Chr.13: 51815698-51815698 on Build GRCh38
rs771920456 Chr.13: 51815752-51815752 on Build GRCh38
rs191559953 Chr.13: 51815869-51815869 on Build GRCh38
rs942120132 Chr.13: 51815909-51815909 on Build GRCh38
rs182852384 Chr.13: 51815948-51815948 on Build GRCh38
rs74538934 Chr.13: 51815632-51815632 on Build GRCh38
rs114298305 Chr.13: 51815572-51815572 on Build GRCh38
rs979572439 Chr.13: 51815562-51815562 on Build GRCh38
rs997411573 Chr.13: 51815894-51815894 on Build GRCh38
rs1047438823 Chr.13: 51815750-51815750 on Build GRCh38
rs1039332341 Chr.13: 51815819-51815819 on Build GRCh38
rs926604943 Chr.13: 51815877-51815877 on Build GRCh38
rs573890499 Chr.13: 51815862-51815862 on Build GRCh38
rs918152693 Chr.13: 51815593-51815593 on Build GRCh38
rs988299673 Chr.13: 51815839-51815839 on Build GRCh38
rs958414939 Chr.13: 51815988-51815988 on Build GRCh38
rs143283314 Chr.13: 51815671-51815671 on Build GRCh38
rs916795141 Chr.13: 51816011-51816011 on Build GRCh38
rs746095588 Chr.13: 51815608-51815609 on Build GRCh38
rs956981980 Chr.13: 51815589-51815589 on Build GRCh38
rs551266660 Chr.13: 51815606-51815606 on Build GRCh38
rs745451225 Chr.13: 51815641-51815641 on Build GRCh38
rs1006546005 Chr.13: 51815584-51815584 on Build GRCh38
rs988780174 Chr.13: 51815590-51815590 on Build GRCh38
rs999227331 Chr.13: 51815969-51815969 on Build GRCh38
rs572270229 Chr.13: 51816001-51816001 on Build GRCh38
rs981221957 Chr.13: 51815872-51815872 on Build GRCh38
rs993309539 Chr.13: 51815563-51815563 on Build GRCh38
rs1048545623 Chr.13: 51815780-51815780 on Build GRCh38
rs929808175 Chr.13: 51815977-51815977 on Build GRCh38
rs552703642 Chr.13: 51815703-51815703 on Build GRCh38
rs9563074 Chr.13: 51815758-51815758 on Build GRCh38
rs966381310 Chr.13: 51815913-51815913 on Build GRCh38
rs994106168 Chr.13: 51815747-51815747 on Build GRCh38
rs1046453038 Chr.13: 51815742-51815742 on Build GRCh38
rs949384877 Chr.13: 51815718-51815718 on Build GRCh38
rs930034167 Chr.13: 51815768-51815768 on Build GRCh38
rs1032095902 Chr.13: 51815976-51815976 on Build GRCh38
rs1017898899 Chr.13: 51815778-51815778 on Build GRCh38
rs146463112 Chr.13: 51815940-51815940 on Build GRCh38
rs567057555 Chr.13: 51815868-51815868 on Build GRCh38
rs1000558730 Chr.13: 51815791-51815791 on Build GRCh38
rs958268831 Chr.13: 51815681-51815681 on Build GRCh38
rs145330218 Chr.13: 51815912-51815912 on Build GRCh38
rs530463601 Chr.13: 51815624-51815624 on Build GRCh38
rs963801343 Chr.13: 51815790-51815790 on Build GRCh38
rs1025658579 Chr.13: 51815841-51815841 on Build GRCh38
rs1047578875 Chr.13: 51815565-51815565 on Build GRCh38
rs1046859715 Chr.13: 51816004-51816004 on Build GRCh38
rs1032497938 Chr.13: 51815806-51815806 on Build GRCh38
rs977839992 Chr.13: 51815663-51815663 on Build GRCh38
rs891038096 Chr.13: 51816017-51816017 on Build GRCh38
rs970987025 Chr.13: 51816020-51816020 on Build GRCh38
rs991441245 Chr.13: 51815683-51815683 on Build GRCh38

View Full Product Details