Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31705251-31705746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLUHP3
Entrez Gene ID
100132341
Gene Name
clustered mitochondria homolog pseudogene 3
Gene Aliases
C16orf67, KIAA0664L3, KIAA0664P3, URLC3
UniGene
Hs.715792
Gene Chromosome Location
Chr.16: 31700613-31707437 on Build GRCh38
Transcripts
NR_024034.1

SNP Detail

SNP ID Chromosome Location
rs892138934 Chr.16: 31705583-31705583 on Build GRCh38
rs959072127 Chr.16: 31705395-31705395 on Build GRCh38
rs550138361 Chr.16: 31705556-31705556 on Build GRCh38
rs1009157860 Chr.16: 31705626-31705626 on Build GRCh38
rs138320027 Chr.16: 31705352-31705352 on Build GRCh38
rs767430064 Chr.16: 31705397-31705397 on Build GRCh38
rs974146375 Chr.16: 31705404-31705404 on Build GRCh38
rs537995883 Chr.16: 31705484-31705484 on Build GRCh38
rs997130513 Chr.16: 31705524-31705524 on Build GRCh38
rs867989363 Chr.16: 31705696-31705696 on Build GRCh38
rs150222252 Chr.16: 31705694-31705694 on Build GRCh38
rs535470598 Chr.16: 31705582-31705582 on Build GRCh38
rs1040307484 Chr.16: 31705543-31705543 on Build GRCh38
rs935887271 Chr.16: 31705411-31705411 on Build GRCh38
rs949893732 Chr.16: 31705704-31705704 on Build GRCh38
rs9938037 Chr.16: 31705280-31705280 on Build GRCh38
rs925637222 Chr.16: 31705687-31705687 on Build GRCh38
rs908644924 Chr.16: 31705709-31705709 on Build GRCh38
rs915820896 Chr.16: 31705703-31705703 on Build GRCh38
rs879197631 Chr.16: 31705448-31705448 on Build GRCh38
rs911628529 Chr.16: 31705372-31705372 on Build GRCh38
rs898761884 Chr.16: 31705504-31705504 on Build GRCh38
rs61744981 Chr.16: 31705406-31705406 on Build GRCh38
rs774447971 Chr.16: 31705330-31705330 on Build GRCh38
rs576270344 Chr.16: 31705725-31705725 on Build GRCh38
rs963239930 Chr.16: 31705691-31705692 on Build GRCh38
rs1002420316 Chr.16: 31705387-31705387 on Build GRCh38
rs1033500384 Chr.16: 31705388-31705388 on Build GRCh38
rs557794640 Chr.16: 31705708-31705708 on Build GRCh38
rs551826045 Chr.16: 31705442-31705442 on Build GRCh38
rs1055624250 Chr.16: 31705415-31705415 on Build GRCh38
rs936465365 Chr.16: 31705558-31705558 on Build GRCh38
rs1024812504 Chr.16: 31705356-31705356 on Build GRCh38
rs887155605 Chr.16: 31705473-31705473 on Build GRCh38
rs547366889 Chr.16: 31705375-31705375 on Build GRCh38
rs970598325 Chr.16: 31705364-31705364 on Build GRCh38
rs113131983 Chr.16: 31705611-31705611 on Build GRCh38
rs955513807 Chr.16: 31705332-31705332 on Build GRCh38
rs1028563616 Chr.16: 31705530-31705530 on Build GRCh38
rs571972568 Chr.16: 31705683-31705683 on Build GRCh38
rs951379176 Chr.16: 31705707-31705707 on Build GRCh38
rs977125958 Chr.16: 31705676-31705676 on Build GRCh38
rs915765513 Chr.16: 31705428-31705428 on Build GRCh38
rs940072132 Chr.16: 31705713-31705713 on Build GRCh38
rs977459235 Chr.16: 31705403-31705403 on Build GRCh38
rs1046314865 Chr.16: 31705634-31705634 on Build GRCh38
rs906463958 Chr.16: 31705650-31705650 on Build GRCh38
rs955565743 Chr.16: 31705548-31705548 on Build GRCh38
rs1038754466 Chr.16: 31705489-31705489 on Build GRCh38
rs570220260 Chr.16: 31705447-31705447 on Build GRCh38
rs997037376 Chr.16: 31705310-31705310 on Build GRCh38
rs1008393787 Chr.16: 31705551-31705551 on Build GRCh38
rs181600203 Chr.16: 31705673-31705673 on Build GRCh38
rs533284268 Chr.16: 31705432-31705432 on Build GRCh38
rs113458074 Chr.16: 31705579-31705579 on Build GRCh38

View Full Product Details