Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178184248-178184810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMCL1P1
Entrez Gene ID
64396
Gene Name
germ cell-less, spermatogenesis associated 1 pseudogene 1
Gene Aliases
GCL, GMCL1L, GMCL2
UniGene
Hs.591738
Gene Chromosome Location
Chr.5: 178184510-178187432 on Build GRCh38
Transcripts
NR_003281.1

SNP Detail

SNP ID Chromosome Location
rs571535218 Chr.5: 178184564-178184564 on Build GRCh38
rs77654038 Chr.5: 178184383-178184384 on Build GRCh38
rs773699452 Chr.5: 178184433-178184433 on Build GRCh38
rs973914381 Chr.5: 178184465-178184465 on Build GRCh38
rs4976745 Chr.5: 178184675-178184675 on Build GRCh38
rs558761080 Chr.5: 178184778-178184778 on Build GRCh38
rs982666877 Chr.5: 178184736-178184736 on Build GRCh38
rs938794018 Chr.5: 178184508-178184508 on Build GRCh38
rs185611098 Chr.5: 178184506-178184506 on Build GRCh38
rs530973288 Chr.5: 178184485-178184485 on Build GRCh38
rs563527368 Chr.5: 178184444-178184444 on Build GRCh38
rs754556346 Chr.5: 178184725-178184725 on Build GRCh38
rs540564492 Chr.5: 178184765-178184765 on Build GRCh38
rs928523739 Chr.5: 178184741-178184741 on Build GRCh38
rs955130681 Chr.5: 178184322-178184322 on Build GRCh38
rs115931401 Chr.5: 178184617-178184617 on Build GRCh38
rs9647596 Chr.5: 178184666-178184666 on Build GRCh38
rs61599137 Chr.5: 178184377-178184377 on Build GRCh38
rs929870458 Chr.5: 178184487-178184487 on Build GRCh38
rs762146310 Chr.5: 178184318-178184318 on Build GRCh38
rs535678310 Chr.5: 178184627-178184627 on Build GRCh38
rs554539195 Chr.5: 178184719-178184719 on Build GRCh38
rs573189857 Chr.5: 178184732-178184732 on Build GRCh38
rs986929692 Chr.5: 178184362-178184362 on Build GRCh38
rs974345360 Chr.5: 178184685-178184685 on Build GRCh38
rs188958752 Chr.5: 178184551-178184551 on Build GRCh38
rs144292756 Chr.5: 178184438-178184439 on Build GRCh38
rs9329136 Chr.5: 178184647-178184647 on Build GRCh38
rs567643123 Chr.5: 178184517-178184517 on Build GRCh38
rs1030640055 Chr.5: 178184582-178184582 on Build GRCh38
rs919880585 Chr.5: 178184467-178184467 on Build GRCh38
rs1055797879 Chr.5: 178184749-178184749 on Build GRCh38
rs534858304 Chr.5: 178184518-178184518 on Build GRCh38
rs75950232 Chr.5: 178184384-178184384 on Build GRCh38
rs4976744 Chr.5: 178184406-178184406 on Build GRCh38
rs558143465 Chr.5: 178184733-178184733 on Build GRCh38
rs947461803 Chr.5: 178184779-178184779 on Build GRCh38
rs1012199738 Chr.5: 178184532-178184533 on Build GRCh38
rs1047102166 Chr.5: 178184494-178184497 on Build GRCh38
rs903455694 Chr.5: 178184544-178184544 on Build GRCh38
rs374564174 Chr.5: 178184642-178184642 on Build GRCh38
rs398000044 Chr.5: 178184384-178184384 on Build GRCh38
rs371387033 Chr.5: 178184591-178184591 on Build GRCh38
rs911069900 Chr.5: 178184388-178184388 on Build GRCh38
rs1021788582 Chr.5: 178184542-178184542 on Build GRCh38
rs938705145 Chr.5: 178184746-178184746 on Build GRCh38

View Full Product Details