Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178185047-178185607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMCL1P1
Entrez Gene ID
64396
Gene Name
germ cell-less, spermatogenesis associated 1 pseudogene 1
Gene Aliases
GCL, GMCL1L, GMCL2
UniGene
Hs.591738
Gene Chromosome Location
Chr.5: 178184510-178187432 on Build GRCh38
Transcripts
NR_003281.1

SNP Detail

SNP ID Chromosome Location
rs934851781 Chr.5: 178185280-178185280 on Build GRCh38
rs534073856 Chr.5: 178185581-178185581 on Build GRCh38
rs761163383 Chr.5: 178185547-178185547 on Build GRCh38
rs1039779454 Chr.5: 178185372-178185372 on Build GRCh38
rs1035496521 Chr.5: 178185574-178185574 on Build GRCh38
rs188826425 Chr.5: 178185434-178185434 on Build GRCh38
rs915876882 Chr.5: 178185132-178185132 on Build GRCh38
rs566498328 Chr.5: 178185564-178185564 on Build GRCh38
rs7719717 Chr.5: 178185292-178185292 on Build GRCh38
rs868290589 Chr.5: 178185277-178185277 on Build GRCh38
rs947478923 Chr.5: 178185161-178185161 on Build GRCh38
rs899474170 Chr.5: 178185472-178185472 on Build GRCh38
rs995157722 Chr.5: 178185499-178185499 on Build GRCh38
rs372650530 Chr.5: 178185193-178185193 on Build GRCh38
rs771308381 Chr.5: 178185554-178185554 on Build GRCh38
rs542833912 Chr.5: 178185211-178185211 on Build GRCh38
rs924683339 Chr.5: 178185254-178185254 on Build GRCh38
rs12657143 Chr.5: 178185356-178185356 on Build GRCh38
rs1026759057 Chr.5: 178185540-178185541 on Build GRCh38
rs768730474 Chr.5: 178185139-178185140 on Build GRCh38
rs7715462 Chr.5: 178185540-178185540 on Build GRCh38
rs890736078 Chr.5: 178185300-178185300 on Build GRCh38
rs991908488 Chr.5: 178185114-178185114 on Build GRCh38
rs772421053 Chr.5: 178185370-178185370 on Build GRCh38
rs551456171 Chr.5: 178185148-178185148 on Build GRCh38
rs554589592 Chr.5: 178185550-178185550 on Build GRCh38

View Full Product Details