Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 240177830-240178103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMN2
Entrez Gene ID
56776
Gene Name
formin 2
Gene Aliases
-
UniGene
Hs.24889
Gene Chromosome Location
Chr.1: 240013776-240475189 on Build GRCh38
Transcripts
NM_001305424.1 NM_020066.4 XM_011544237.2 XM_017001837.1 XM_017001838.1 XM_017001839.1 XM_017001840.1 XM_017001841.1 XM_017001842.1 XM_017001843.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4473972 c.2315C>T p.S772F Chr.1: 240178024-240178024 on Build GRCh38
311242 c.2291G>T p.R764L Chr.1: 240178000-240178000 on Build GRCh38
6192224 c.2269T>C p.S757P Chr.1: 240177978-240177978 on Build GRCh38
5964745 c.2343C>A p.L781L Chr.1: 240178052-240178052 on Build GRCh38
5380225 c.2297C>T p.P766L Chr.1: 240178006-240178006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750125669 Chr.1: 240177953-240177953 on Build GRCh38
rs372902195 Chr.1: 240178074-240178074 on Build GRCh38
rs770042272 Chr.1: 240177878-240177878 on Build GRCh38
rs753071977 Chr.1: 240177922-240177922 on Build GRCh38
rs543546993 Chr.1: 240178072-240178072 on Build GRCh38
rs777504752 Chr.1: 240178000-240178000 on Build GRCh38
rs781151128 Chr.1: 240177989-240177989 on Build GRCh38
rs895732310 Chr.1: 240177868-240177868 on Build GRCh38
rs1034244490 Chr.1: 240177867-240177867 on Build GRCh38
rs973160262 Chr.1: 240177965-240177965 on Build GRCh38
rs771085071 Chr.1: 240178029-240178029 on Build GRCh38
rs766122332 Chr.1: 240178060-240178060 on Build GRCh38
rs751067966 Chr.1: 240178067-240178067 on Build GRCh38
rs375865107 Chr.1: 240177941-240177941 on Build GRCh38
rs772498975 Chr.1: 240178035-240178035 on Build GRCh38
rs974691443 Chr.1: 240178011-240178011 on Build GRCh38
rs141897898 Chr.1: 240178034-240178034 on Build GRCh38
rs747941856 Chr.1: 240177993-240177993 on Build GRCh38
rs776046459 Chr.1: 240178043-240178043 on Build GRCh38
rs955282208 Chr.1: 240177928-240177928 on Build GRCh38
rs779720695 Chr.1: 240177960-240177960 on Build GRCh38
rs369120788 Chr.1: 240178022-240178022 on Build GRCh38
rs768057945 Chr.1: 240177911-240177911 on Build GRCh38
rs918023063 Chr.1: 240178075-240178075 on Build GRCh38
rs777007362 Chr.1: 240178053-240178053 on Build GRCh38
rs764589234 Chr.1: 240178049-240178049 on Build GRCh38
rs201141753 Chr.1: 240177888-240177888 on Build GRCh38
rs765034823 Chr.1: 240177952-240177952 on Build GRCh38
rs766476454 Chr.1: 240177890-240177890 on Build GRCh38
rs3765588 Chr.1: 240177968-240177968 on Build GRCh38
rs761219159 Chr.1: 240178046-240178046 on Build GRCh38
rs773399468 Chr.1: 240177886-240177886 on Build GRCh38
rs1014546542 Chr.1: 240177893-240177893 on Build GRCh38
rs758087582 Chr.1: 240177957-240177957 on Build GRCh38
rs372602602 Chr.1: 240177906-240177906 on Build GRCh38
rs147945249 Chr.1: 240178007-240178007 on Build GRCh38
rs3765589 Chr.1: 240177864-240177864 on Build GRCh38
rs199795761 Chr.1: 240177916-240177916 on Build GRCh38
rs762749579 Chr.1: 240178054-240178054 on Build GRCh38
rs769597954 Chr.1: 240177998-240177998 on Build GRCh38
rs200182734 Chr.1: 240177969-240177969 on Build GRCh38
rs756471884 Chr.1: 240177948-240177948 on Build GRCh38
rs774423627 Chr.1: 240177900-240177900 on Build GRCh38
rs953549682 Chr.1: 240177985-240177985 on Build GRCh38

View Full Product Details