Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41853864-41854101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL11
Entrez Gene ID
55175
Gene Name
kelch like family member 11
Gene Aliases
-
UniGene
Hs.13268
Gene Chromosome Location
Chr.17: 41848518-41865431 on Build GRCh38
Transcripts
NM_018143.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5852523 c.2002G>T p.G668C Chr.17: 41853865-41853865 on Build GRCh38
6839393 c.1843G>C p.G615R Chr.17: 41854024-41854024 on Build GRCh38
3187963 c.1907C>T p.A636V Chr.17: 41853960-41853960 on Build GRCh38
6696466 c.1832T>C p.V611A Chr.17: 41854035-41854035 on Build GRCh38
5386369 c.1797C>T p.I599I Chr.17: 41854070-41854070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781921338 Chr.17: 41853991-41853991 on Build GRCh38
rs562966381 Chr.17: 41854070-41854070 on Build GRCh38
rs782206413 Chr.17: 41853960-41853960 on Build GRCh38
rs782149069 Chr.17: 41853890-41853890 on Build GRCh38
rs782255335 Chr.17: 41853974-41853974 on Build GRCh38
rs781940858 Chr.17: 41853897-41853897 on Build GRCh38
rs782429391 Chr.17: 41853951-41853951 on Build GRCh38
rs782701237 Chr.17: 41853893-41853893 on Build GRCh38
rs566584745 Chr.17: 41853892-41853892 on Build GRCh38
rs781899221 Chr.17: 41853944-41853944 on Build GRCh38
rs148410954 Chr.17: 41854064-41854064 on Build GRCh38
rs782476795 Chr.17: 41853961-41853961 on Build GRCh38
rs781975879 Chr.17: 41854056-41854056 on Build GRCh38
rs146181967 Chr.17: 41853920-41853920 on Build GRCh38
rs782407439 Chr.17: 41853989-41853989 on Build GRCh38
rs782025209 Chr.17: 41854075-41854075 on Build GRCh38
rs782083927 Chr.17: 41853900-41853900 on Build GRCh38
rs142287839 Chr.17: 41853887-41853887 on Build GRCh38
rs782579158 Chr.17: 41853959-41853959 on Build GRCh38
rs782639299 Chr.17: 41853970-41853970 on Build GRCh38
rs782201695 Chr.17: 41854026-41854026 on Build GRCh38
rs201183215 Chr.17: 41854076-41854076 on Build GRCh38
rs782347713 Chr.17: 41854048-41854048 on Build GRCh38
rs782765270 Chr.17: 41853904-41853904 on Build GRCh38
rs782536248 Chr.17: 41853918-41853918 on Build GRCh38
rs897357584 Chr.17: 41853965-41853965 on Build GRCh38
rs142556293 Chr.17: 41854079-41854079 on Build GRCh38
rs201471518 Chr.17: 41853915-41853915 on Build GRCh38

View Full Product Details