Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45367026-45367265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYS1
Entrez Gene ID
90196
Gene Name
Sys1 golgi trafficking protein
Gene Aliases
C20orf169, dJ453C12.4, dJ453C12.4.1
UniGene
Hs.655055
Gene Chromosome Location
Chr.20: 45361937-45376802 on Build GRCh38
Transcripts
NM_001099791.2 NM_001197129.1 NM_033542.3 NR_003189.2

Gene Symbol
SYS1-DBNDD2
Entrez Gene ID
767557
Gene Name
SYS1-DBNDD2 readthrough (NMD candidate)
Gene Aliases
C20orf169-DBNDD2
UniGene
Hs.730643
Gene Chromosome Location
Chr.20: 45363169-45410610 on Build GRCh38
Transcripts
NM_001099791.2 NM_001197129.1 NM_033542.3 NR_003189.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1307453 c.454C>T p.P152S Chr.20: 45367098-45367098 on Build GRCh38
459741 c.387C>T p.L129L Chr.20: 45367031-45367031 on Build GRCh38
1228302 c.418C>T p.R140W Chr.20: 45367062-45367062 on Build GRCh38
3547010 c.461C>T p.S154F Chr.20: 45367105-45367105 on Build GRCh38
3293411 c.399C>T p.I133I Chr.20: 45367043-45367043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772730174 Chr.20: 45367146-45367146 on Build GRCh38
rs776461956 Chr.20: 45367063-45367063 on Build GRCh38
rs770901952 Chr.20: 45367113-45367113 on Build GRCh38
rs190104853 Chr.20: 45367151-45367151 on Build GRCh38
rs756373449 Chr.20: 45367107-45367107 on Build GRCh38
rs569244951 Chr.20: 45367144-45367144 on Build GRCh38
rs865906429 Chr.20: 45367106-45367106 on Build GRCh38
rs201471133 Chr.20: 45367066-45367066 on Build GRCh38
rs549101436 Chr.20: 45367112-45367112 on Build GRCh38
rs746344394 Chr.20: 45367120-45367120 on Build GRCh38
rs772663420 Chr.20: 45367123-45367123 on Build GRCh38
rs772367283 Chr.20: 45367086-45367086 on Build GRCh38
rs754809931 Chr.20: 45367090-45367090 on Build GRCh38
rs776016913 Chr.20: 45367130-45367130 on Build GRCh38
rs893269992 Chr.20: 45367201-45367201 on Build GRCh38
rs566745672 Chr.20: 45367062-45367062 on Build GRCh38
rs909007069 Chr.20: 45367195-45367195 on Build GRCh38
rs764745223 Chr.20: 45367067-45367067 on Build GRCh38
rs749958472 Chr.20: 45367068-45367068 on Build GRCh38
rs557871174 Chr.20: 45367152-45367152 on Build GRCh38
rs144570687 Chr.20: 45367091-45367091 on Build GRCh38
rs981539628 Chr.20: 45367145-45367145 on Build GRCh38
rs975510364 Chr.20: 45367089-45367089 on Build GRCh38
rs577930332 Chr.20: 45367232-45367232 on Build GRCh38
rs762299946 Chr.20: 45367072-45367072 on Build GRCh38
rs958648475 Chr.20: 45367170-45367170 on Build GRCh38
rs184455498 Chr.20: 45367108-45367108 on Build GRCh38
rs760887044 Chr.20: 45367143-45367143 on Build GRCh38
rs752443252 Chr.20: 45367094-45367094 on Build GRCh38
rs760035492 Chr.20: 45367057-45367057 on Build GRCh38
rs779611547 Chr.20: 45367119-45367119 on Build GRCh38
rs766401702 Chr.20: 45367080-45367080 on Build GRCh38
rs762577243 Chr.20: 45367148-45367148 on Build GRCh38
rs2245717 Chr.20: 45367240-45367240 on Build GRCh38
rs759492850 Chr.20: 45367159-45367159 on Build GRCh38
rs991318376 Chr.20: 45367179-45367179 on Build GRCh38

View Full Product Details