Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179114172-179114415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS2
Entrez Gene ID
9509
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 2
Gene Aliases
ADAM-TS2, ADAMTS-2, ADAMTS-3, NPI, PC I-NP, PCI-NP, PCINP, PCPNI, PNPI
UniGene
Hs.23871
Gene Chromosome Location
Chr.5: 179110851-179345430 on Build GRCh38
Transcripts
NM_014244.4 XM_017010063.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3614732 c.3220G>A p.E1074K Chr.5: 179114283-179114283 on Build GRCh38
3854169 c.3218T>C p.M1073T Chr.5: 179114285-179114285 on Build GRCh38
4772130 c.3326C>T p.P1109L Chr.5: 179114177-179114177 on Build GRCh38
3614733 c.3216G>A p.R1072R Chr.5: 179114287-179114287 on Build GRCh38
3614730 c.3275C>T p.S1092F Chr.5: 179114228-179114228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766731589 Chr.5: 179114200-179114200 on Build GRCh38
rs979699810 Chr.5: 179114207-179114207 on Build GRCh38
rs962934152 Chr.5: 179114353-179114353 on Build GRCh38
rs781661133 Chr.5: 179114351-179114351 on Build GRCh38
rs908848228 Chr.5: 179114352-179114352 on Build GRCh38
rs755578308 Chr.5: 179114271-179114271 on Build GRCh38
rs1023260232 Chr.5: 179114191-179114191 on Build GRCh38
rs912743717 Chr.5: 179114280-179114280 on Build GRCh38
rs899825126 Chr.5: 179114319-179114319 on Build GRCh38
rs753308886 Chr.5: 179114288-179114288 on Build GRCh38
rs756891532 Chr.5: 179114303-179114303 on Build GRCh38
rs1051658495 Chr.5: 179114390-179114390 on Build GRCh38
rs762648423 Chr.5: 179114199-179114199 on Build GRCh38
rs763179895 Chr.5: 179114196-179114196 on Build GRCh38
rs374842471 Chr.5: 179114261-179114261 on Build GRCh38
rs747348237 Chr.5: 179114310-179114310 on Build GRCh38
rs746853740 Chr.5: 179114258-179114258 on Build GRCh38
rs760653331 Chr.5: 179114209-179114211 on Build GRCh38
rs73806887 Chr.5: 179114224-179114224 on Build GRCh38
rs778572930 Chr.5: 179114308-179114308 on Build GRCh38
rs748619743 Chr.5: 179114357-179114357 on Build GRCh38
rs960542347 Chr.5: 179114241-179114241 on Build GRCh38
rs934159753 Chr.5: 179114387-179114387 on Build GRCh38
rs755393352 Chr.5: 179114348-179114348 on Build GRCh38
rs540171408 Chr.5: 179114363-179114363 on Build GRCh38
rs770564733 Chr.5: 179114380-179114380 on Build GRCh38
rs759444266 Chr.5: 179114234-179114234 on Build GRCh38
rs143326244 Chr.5: 179114284-179114284 on Build GRCh38
rs944065693 Chr.5: 179114296-179114296 on Build GRCh38
rs925554043 Chr.5: 179114217-179114217 on Build GRCh38
rs950427186 Chr.5: 179114317-179114317 on Build GRCh38
rs752742612 Chr.5: 179114270-179114270 on Build GRCh38
rs975666229 Chr.5: 179114378-179114378 on Build GRCh38

View Full Product Details