Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 150445448-150445703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAMLD1
Entrez Gene ID
10046
Gene Name
mastermind like domain containing 1
Gene Aliases
CG1, CXorf6, F18, HYSP2
UniGene
Hs.20136
Gene Chromosome Location
Chr.X: 150361422-150514178 on Build GRCh38
Transcripts
NM_001177465.2 NM_001177466.2 NM_005491.4 XM_006724802.3 XM_006724803.3 XM_011531092.2 XM_011531093.2 XM_011531095.2 XM_017029188.1 XM_017029189.1

SNP Detail

SNP ID Chromosome Location
rs370838699 Chr.X: 150445573-150445573 on Build GRCh38
rs371792879 Chr.X: 150445643-150445643 on Build GRCh38
rs367901550 Chr.X: 150445493-150445493 on Build GRCh38
rs782120018 Chr.X: 150445574-150445574 on Build GRCh38
rs201825641 Chr.X: 150445480-150445480 on Build GRCh38
rs962285386 Chr.X: 150445633-150445633 on Build GRCh38
rs781873091 Chr.X: 150445632-150445632 on Build GRCh38
rs781889915 Chr.X: 150445619-150445619 on Build GRCh38
rs782646291 Chr.X: 150445628-150445628 on Build GRCh38
rs781933957 Chr.X: 150445567-150445567 on Build GRCh38
rs782470142 Chr.X: 150445568-150445568 on Build GRCh38
rs782474376 Chr.X: 150445641-150445641 on Build GRCh38
rs200035082 Chr.X: 150445534-150445534 on Build GRCh38
rs781800896 Chr.X: 150445469-150445469 on Build GRCh38
rs782649876 Chr.X: 150445516-150445516 on Build GRCh38
rs368441164 Chr.X: 150445612-150445612 on Build GRCh38
rs782046657 Chr.X: 150445606-150445606 on Build GRCh38
rs782761871 Chr.X: 150445630-150445630 on Build GRCh38
rs373970502 Chr.X: 150445584-150445584 on Build GRCh38
rs782488152 Chr.X: 150445637-150445637 on Build GRCh38
rs781837754 Chr.X: 150445603-150445603 on Build GRCh38
rs376112194 Chr.X: 150445468-150445468 on Build GRCh38
rs199908749 Chr.X: 150445539-150445539 on Build GRCh38
rs782416424 Chr.X: 150445572-150445572 on Build GRCh38

View Full Product Details