Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45658867-45659130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SQRDL
Entrez Gene ID
58472
Gene Name
sulfide quinone reductase-like (yeast)
Gene Aliases
CGI-44, PRO1975, SQOR
UniGene
Hs.511251
Gene Chromosome Location
Chr.15: 45631148-45691294 on Build GRCh38
Transcripts
NM_001271213.1 NM_021199.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5549773 c.116C>T p.A39V Chr.15: 45659039-45659039 on Build GRCh38
7391943 c.50C>T p.A17V Chr.15: 45658973-45658973 on Build GRCh38
6274534 c.147G>T p.L49L Chr.15: 45659070-45659070 on Build GRCh38
4520051 c.103G>A p.G35R Chr.15: 45659026-45659026 on Build GRCh38
7391942 c.19G>C p.V7L Chr.15: 45658942-45658942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200511008 Chr.15: 45659007-45659007 on Build GRCh38
rs528515915 Chr.15: 45659029-45659029 on Build GRCh38
rs752283192 Chr.15: 45658889-45658889 on Build GRCh38
rs374284135 Chr.15: 45659086-45659086 on Build GRCh38
rs147877839 Chr.15: 45659059-45659059 on Build GRCh38
rs115371596 Chr.15: 45658944-45658944 on Build GRCh38
rs749115602 Chr.15: 45658917-45658917 on Build GRCh38
rs758231078 Chr.15: 45659078-45659078 on Build GRCh38
rs771327815 Chr.15: 45659092-45659092 on Build GRCh38
rs114299990 Chr.15: 45659090-45659090 on Build GRCh38
rs748292795 Chr.15: 45659025-45659025 on Build GRCh38
rs562295995 Chr.15: 45659039-45659039 on Build GRCh38
rs752299752 Chr.15: 45658989-45658989 on Build GRCh38
rs150789874 Chr.15: 45658973-45658973 on Build GRCh38
rs777406121 Chr.15: 45658911-45658911 on Build GRCh38
rs528993979 Chr.15: 45659045-45659045 on Build GRCh38
rs774896218 Chr.15: 45659040-45659040 on Build GRCh38
rs763564633 Chr.15: 45659106-45659106 on Build GRCh38
rs147700533 Chr.15: 45659010-45659010 on Build GRCh38
rs763412007 Chr.15: 45658963-45658963 on Build GRCh38
rs760186824 Chr.15: 45658978-45658978 on Build GRCh38
rs770790961 Chr.15: 45658919-45658919 on Build GRCh38
rs780043896 Chr.15: 45659079-45659079 on Build GRCh38
rs182682752 Chr.15: 45658910-45658910 on Build GRCh38
rs144678798 Chr.15: 45659051-45659051 on Build GRCh38
rs760247500 Chr.15: 45659056-45659056 on Build GRCh38
rs139154395 Chr.15: 45659004-45659004 on Build GRCh38
rs753484574 Chr.15: 45659067-45659067 on Build GRCh38
rs889980977 Chr.15: 45659094-45659094 on Build GRCh38
rs535949978 Chr.15: 45659105-45659105 on Build GRCh38
rs776170946 Chr.15: 45659107-45659107 on Build GRCh38
rs756201981 Chr.15: 45659085-45659085 on Build GRCh38
rs945342614 Chr.15: 45659064-45659064 on Build GRCh38
rs778061542 Chr.15: 45659087-45659087 on Build GRCh38
rs748115877 Chr.15: 45659083-45659083 on Build GRCh38
rs370293201 Chr.15: 45659074-45659074 on Build GRCh38
rs774806357 Chr.15: 45659098-45659098 on Build GRCh38
rs372958070 Chr.15: 45658987-45658987 on Build GRCh38
rs142428650 Chr.15: 45659026-45659026 on Build GRCh38
rs540812384 Chr.15: 45658993-45658993 on Build GRCh38
rs917119412 Chr.15: 45659002-45659002 on Build GRCh38
rs200945908 Chr.15: 45658953-45658953 on Build GRCh38
rs149926870 Chr.15: 45659005-45659005 on Build GRCh38
rs745777676 Chr.15: 45659009-45659009 on Build GRCh38
rs972438389 Chr.15: 45658997-45658997 on Build GRCh38
rs187332233 Chr.15: 45658958-45658958 on Build GRCh38
rs769395245 Chr.15: 45659057-45659058 on Build GRCh38
rs761922019 Chr.15: 45658957-45658957 on Build GRCh38
rs375963712 Chr.15: 45659041-45659041 on Build GRCh38
rs768614491 Chr.15: 45658952-45658952 on Build GRCh38
rs778838657 Chr.15: 45658925-45658925 on Build GRCh38
rs779128697 Chr.15: 45658927-45658927 on Build GRCh38
rs757034077 Chr.15: 45659006-45659006 on Build GRCh38
rs376077055 Chr.15: 45659104-45659104 on Build GRCh38
rs142221428 Chr.15: 45658965-45658965 on Build GRCh38
rs550473526 Chr.15: 45659076-45659076 on Build GRCh38
rs543144723 Chr.15: 45659008-45659008 on Build GRCh38
rs752082929 Chr.15: 45659054-45659054 on Build GRCh38
rs148921714 Chr.15: 45659077-45659077 on Build GRCh38
rs773669761 Chr.15: 45658959-45658959 on Build GRCh38
rs551364879 Chr.15: 45659038-45659038 on Build GRCh38

View Full Product Details