Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3639482-3639747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART5
Entrez Gene ID
116969
Gene Name
ADP-ribosyltransferase 5
Gene Aliases
ARTC5
UniGene
Hs.125680
Gene Chromosome Location
Chr.11: 3638503-3642392 on Build GRCh38
Transcripts
NM_001079536.1 NM_001297668.1 NM_053017.4 NR_123732.1 XM_005252779.3 XM_006718136.3 XM_011519879.2 XM_017017171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3447073 c.719C>T p.A240V Chr.11: 3639710-3639710 on Build GRCh38
687407 c.740G>T p.W247L Chr.11: 3639689-3639689 on Build GRCh38
7127524 c.745T>C p.Y249H Chr.11: 3639684-3639684 on Build GRCh38
7425968 c.688T>A p.F230I Chr.11: 3639741-3639741 on Build GRCh38
6265477 c.787+6C>A p.? Chr.11: 3639636-3639636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002004094 Chr.11: 3639608-3639608 on Build GRCh38
rs200339999 Chr.11: 3639654-3639654 on Build GRCh38
rs760651964 Chr.11: 3639629-3639629 on Build GRCh38
rs770881980 Chr.11: 3639700-3639700 on Build GRCh38
rs754205045 Chr.11: 3639676-3639676 on Build GRCh38
rs762486736 Chr.11: 3639650-3639650 on Build GRCh38
rs368897365 Chr.11: 3639640-3639640 on Build GRCh38
rs376801553 Chr.11: 3639716-3639716 on Build GRCh38
rs905094882 Chr.11: 3639586-3639586 on Build GRCh38
rs933420214 Chr.11: 3639569-3639569 on Build GRCh38
rs755213415 Chr.11: 3639678-3639678 on Build GRCh38
rs752286751 Chr.11: 3639645-3639645 on Build GRCh38
rs776313535 Chr.11: 3639709-3639709 on Build GRCh38
rs143414771 Chr.11: 3639713-3639713 on Build GRCh38
rs750852006 Chr.11: 3639567-3639567 on Build GRCh38
rs980666775 Chr.11: 3639553-3639553 on Build GRCh38
rs139576225 Chr.11: 3639690-3639690 on Build GRCh38
rs764936751 Chr.11: 3639642-3639642 on Build GRCh38
rs779063382 Chr.11: 3639682-3639682 on Build GRCh38
rs768726199 Chr.11: 3639642-3639643 on Build GRCh38
rs780452120 Chr.11: 3639604-3639604 on Build GRCh38
rs988124956 Chr.11: 3639558-3639558 on Build GRCh38
rs768834172 Chr.11: 3639609-3639609 on Build GRCh38
rs1032238602 Chr.11: 3639555-3639555 on Build GRCh38
rs748174487 Chr.11: 3639616-3639616 on Build GRCh38
rs201775531 Chr.11: 3639632-3639632 on Build GRCh38
rs2271589 Chr.11: 3639684-3639684 on Build GRCh38
rs931291548 Chr.11: 3639557-3639557 on Build GRCh38
rs750949438 Chr.11: 3639655-3639655 on Build GRCh38
rs772023577 Chr.11: 3639626-3639626 on Build GRCh38
rs200399189 Chr.11: 3639567-3639567 on Build GRCh38
rs749537477 Chr.11: 3639605-3639605 on Build GRCh38
rs969321556 Chr.11: 3639515-3639515 on Build GRCh38
rs374818380 Chr.11: 3639630-3639630 on Build GRCh38
rs142871932 Chr.11: 3639683-3639683 on Build GRCh38
rs753617164 Chr.11: 3639699-3639699 on Build GRCh38
rs759313228 Chr.11: 3639639-3639639 on Build GRCh38
rs371743932 Chr.11: 3639601-3639601 on Build GRCh38
rs536417141 Chr.11: 3639566-3639566 on Build GRCh38
rs777909444 Chr.11: 3639691-3639691 on Build GRCh38
rs756637453 Chr.11: 3639667-3639667 on Build GRCh38
rs780320762 Chr.11: 3639674-3639674 on Build GRCh38
rs750397186 Chr.11: 3639563-3639563 on Build GRCh38

View Full Product Details