Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196722490-196722715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGX
Entrez Gene ID
54965
Gene Name
phosphatidylinositol glycan anchor biosynthesis class X
Gene Aliases
PIG-X
UniGene
Hs.223296
Gene Chromosome Location
Chr.3: 196712374-196736007 on Build GRCh38
Transcripts
NM_001166304.1 NM_017861.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5978788 c.140G>A p.G47E Chr.3: 196722501-196722501 on Build GRCh38
4879952 c.193G>T p.E65* Chr.3: 196722554-196722554 on Build GRCh38
6319465 c.166G>T p.A56S Chr.3: 196722527-196722527 on Build GRCh38
3380503 c.299G>A p.R100Q Chr.3: 196722537-196722537 on Build GRCh38
5978787 c.263G>A p.G88E Chr.3: 196722501-196722501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774321275 Chr.3: 196722573-196722574 on Build GRCh38
rs1039253216 Chr.3: 196722548-196722548 on Build GRCh38
rs752310032 Chr.3: 196722517-196722517 on Build GRCh38
rs1017601509 Chr.3: 196722676-196722676 on Build GRCh38
rs755004221 Chr.3: 196722514-196722514 on Build GRCh38
rs112322208 Chr.3: 196722661-196722661 on Build GRCh38
rs200546966 Chr.3: 196722572-196722572 on Build GRCh38
rs149218490 Chr.3: 196722688-196722688 on Build GRCh38
rs747489665 Chr.3: 196722586-196722586 on Build GRCh38
rs993690419 Chr.3: 196722538-196722538 on Build GRCh38
rs777489645 Chr.3: 196722554-196722554 on Build GRCh38
rs776565142 Chr.3: 196722601-196722601 on Build GRCh38
rs758787442 Chr.3: 196722569-196722569 on Build GRCh38
rs530669068 Chr.3: 196722638-196722638 on Build GRCh38
rs964763849 Chr.3: 196722680-196722680 on Build GRCh38
rs767114998 Chr.3: 196722516-196722516 on Build GRCh38
rs952869102 Chr.3: 196722649-196722649 on Build GRCh38
rs748260101 Chr.3: 196722607-196722607 on Build GRCh38
rs199645579 Chr.3: 196722537-196722537 on Build GRCh38
rs1008789352 Chr.3: 196722573-196722573 on Build GRCh38
rs538942057 Chr.3: 196722643-196722643 on Build GRCh38
rs964520153 Chr.3: 196722626-196722626 on Build GRCh38
rs746164540 Chr.3: 196722557-196722557 on Build GRCh38

View Full Product Details