Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129022107-129022380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3GLB2
Entrez Gene ID
56904
Gene Name
SH3 domain containing GRB2 like endophilin B2
Gene Aliases
PP6569, PP9455, RRIG1
UniGene
Hs.460238
Gene Chromosome Location
Chr.9: 129007792-129028353 on Build GRCh38
Transcripts
NM_001287045.1 NM_001287046.1 NM_020145.3 XM_005252098.2 XM_005252100.2 XM_005252101.3 XM_006717188.2 XM_006717189.2 XM_006717190.1 XM_006717191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1105804 c.134G>A p.R45Q Chr.9: 129022353-129022353 on Build GRCh38
4971673 c.144C>T p.S48S Chr.9: 129022343-129022343 on Build GRCh38
3847811 c.150G>T p.K50N Chr.9: 129022337-129022337 on Build GRCh38
4971674 c.144C>T p.S48S Chr.9: 129022343-129022343 on Build GRCh38
3847810 c.150G>T p.K50N Chr.9: 129022337-129022337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762172912 Chr.9: 129022259-129022259 on Build GRCh38
rs774874088 Chr.9: 129022354-129022354 on Build GRCh38
rs369693171 Chr.9: 129022180-129022180 on Build GRCh38
rs989161014 Chr.9: 129022194-129022194 on Build GRCh38
rs138289691 Chr.9: 129022353-129022353 on Build GRCh38
rs117054692 Chr.9: 129022244-129022244 on Build GRCh38
rs1018159688 Chr.9: 129022156-129022156 on Build GRCh38
rs1021511434 Chr.9: 129022201-129022201 on Build GRCh38
rs142474605 Chr.9: 129022357-129022357 on Build GRCh38
rs1054698545 Chr.9: 129022175-129022175 on Build GRCh38
rs1002570119 Chr.9: 129022138-129022138 on Build GRCh38
rs974161153 Chr.9: 129022192-129022192 on Build GRCh38
rs752676035 Chr.9: 129022273-129022273 on Build GRCh38
rs757138953 Chr.9: 129022285-129022285 on Build GRCh38
rs748249433 Chr.9: 129022239-129022240 on Build GRCh38
rs749381145 Chr.9: 129022352-129022352 on Build GRCh38
rs138489520 Chr.9: 129022260-129022260 on Build GRCh38
rs559976678 Chr.9: 129022254-129022254 on Build GRCh38
rs377137977 Chr.9: 129022274-129022274 on Build GRCh38
rs570033543 Chr.9: 129022267-129022267 on Build GRCh38
rs780050940 Chr.9: 129022340-129022340 on Build GRCh38
rs773967089 Chr.9: 129022310-129022310 on Build GRCh38
rs111947151 Chr.9: 129022136-129022136 on Build GRCh38
rs776030306 Chr.9: 129022243-129022243 on Build GRCh38
rs746115370 Chr.9: 129022308-129022308 on Build GRCh38
rs940688769 Chr.9: 129022300-129022300 on Build GRCh38
rs372967504 Chr.9: 129022268-129022268 on Build GRCh38
rs201485191 Chr.9: 129022266-129022266 on Build GRCh38
rs891071226 Chr.9: 129022130-129022130 on Build GRCh38
rs772777458 Chr.9: 129022256-129022260 on Build GRCh38
rs547472467 Chr.9: 129022191-129022191 on Build GRCh38
rs528306350 Chr.9: 129022179-129022179 on Build GRCh38
rs769813169 Chr.9: 129022240-129022240 on Build GRCh38
rs762119648 Chr.9: 129022355-129022355 on Build GRCh38
rs774637179 Chr.9: 129022245-129022245 on Build GRCh38
rs559254961 Chr.9: 129022135-129022135 on Build GRCh38
rs35770147 Chr.9: 129022214-129022214 on Build GRCh38
rs369733753 Chr.9: 129022286-129022286 on Build GRCh38

View Full Product Details