Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 52800345-52800598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA1
Entrez Gene ID
3672
Gene Name
integrin subunit alpha 1
Gene Aliases
CD49a, VLA1
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52788302-52953655 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Gene Symbol
PELO
Entrez Gene ID
53918
Gene Name
pelota homolog (Drosophila)
Gene Aliases
CGI-17, PRO1770
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52787940-52802617 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5521342 c.195C>T p.L65L Chr.5: 52800589-52800589 on Build GRCh38
3769962 c.159G>A p.T53T Chr.5: 52800553-52800553 on Build GRCh38
3343378 c.103G>A p.V35M Chr.5: 52800497-52800497 on Build GRCh38
7383589 c.138G>T p.K46N Chr.5: 52800532-52800532 on Build GRCh38
117995 c.81G>A p.W27* Chr.5: 52800475-52800475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757916583 Chr.5: 52800480-52800480 on Build GRCh38
rs950314995 Chr.5: 52800526-52800526 on Build GRCh38
rs554934628 Chr.5: 52800432-52800432 on Build GRCh38
rs781139548 Chr.5: 52800433-52800433 on Build GRCh38
rs199820732 Chr.5: 52800445-52800445 on Build GRCh38
rs767881270 Chr.5: 52800382-52800382 on Build GRCh38
rs762516997 Chr.5: 52800578-52800578 on Build GRCh38
rs752362594 Chr.5: 52800472-52800472 on Build GRCh38
rs753418857 Chr.5: 52800384-52800384 on Build GRCh38
rs974381808 Chr.5: 52800435-52800435 on Build GRCh38
rs773751186 Chr.5: 52800567-52800567 on Build GRCh38
rs780308633 Chr.5: 52800541-52800541 on Build GRCh38
rs749484314 Chr.5: 52800543-52800543 on Build GRCh38
rs750806545 Chr.5: 52800461-52800461 on Build GRCh38
rs745637916 Chr.5: 52800520-52800520 on Build GRCh38
rs553902408 Chr.5: 52800407-52800407 on Build GRCh38
rs768714228 Chr.5: 52800545-52800545 on Build GRCh38
rs532592445 Chr.5: 52800515-52800515 on Build GRCh38
rs542925164 Chr.5: 52800429-52800429 on Build GRCh38
rs148212245 Chr.5: 52800466-52800466 on Build GRCh38
rs11547950 Chr.5: 52800422-52800422 on Build GRCh38
rs538357669 Chr.5: 52800403-52800403 on Build GRCh38
rs763082521 Chr.5: 52800450-52800450 on Build GRCh38
rs774441545 Chr.5: 52800556-52800556 on Build GRCh38
rs201194553 Chr.5: 52800495-52800495 on Build GRCh38
rs762254838 Chr.5: 52800557-52800557 on Build GRCh38
rs565147402 Chr.5: 52800502-52800502 on Build GRCh38
rs769542363 Chr.5: 52800539-52800539 on Build GRCh38
rs11547951 Chr.5: 52800451-52800451 on Build GRCh38
rs541189319 Chr.5: 52800564-52800564 on Build GRCh38
rs775613671 Chr.5: 52800449-52800449 on Build GRCh38
rs757292312 Chr.5: 52800509-52800509 on Build GRCh38
rs775154862 Chr.5: 52800573-52800573 on Build GRCh38
rs754654424 Chr.5: 52800395-52800395 on Build GRCh38
rs768283647 Chr.5: 52800535-52800538 on Build GRCh38
rs930216052 Chr.5: 52800427-52800427 on Build GRCh38
rs746048979 Chr.5: 52800442-52800442 on Build GRCh38
rs746821651 Chr.5: 52800479-52800482 on Build GRCh38
rs1046374330 Chr.5: 52800538-52800538 on Build GRCh38
rs779902266 Chr.5: 52800381-52800381 on Build GRCh38
rs762152336 Chr.5: 52800377-52800377 on Build GRCh38
rs767825694 Chr.5: 52800456-52800456 on Build GRCh38
rs572160494 Chr.5: 52800411-52800411 on Build GRCh38
rs373725475 Chr.5: 52800570-52800570 on Build GRCh38
rs777164866 Chr.5: 52800498-52800498 on Build GRCh38
rs777765641 Chr.5: 52800421-52800421 on Build GRCh38
rs774782233 Chr.5: 52800455-52800455 on Build GRCh38
rs900915886 Chr.5: 52800470-52800470 on Build GRCh38

View Full Product Details