Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13935667-13935940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PODNL1
Entrez Gene ID
79883
Gene Name
podocan like 1
Gene Aliases
SLRR5B
UniGene
Hs.448497
Gene Chromosome Location
Chr.19: 13931187-13953392 on Build GRCh38
Transcripts
NM_001146254.1 NM_001146255.1 NM_024825.3 XM_006722901.1 XM_006722902.1 XM_006722903.1 XM_011528308.1 XM_011528309.2 XM_011528310.2 XM_011528311.1 XM_011528313.1 XM_011528314.1 XM_011528316.1 XM_017027313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1711893 c.460G>A p.A154T Chr.19: 13935776-13935776 on Build GRCh38
1722530 c.437C>T p.S146F Chr.19: 13935799-13935799 on Build GRCh38
2148921 c.433C>T p.R145W Chr.19: 13935803-13935803 on Build GRCh38
1162793 c.429G>A p.L143L Chr.19: 13935807-13935807 on Build GRCh38
2150726 c.449C>T p.A150V Chr.19: 13935787-13935787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779971244 Chr.19: 13935751-13935751 on Build GRCh38
rs557005438 Chr.19: 13935870-13935870 on Build GRCh38
rs377072451 Chr.19: 13935835-13935835 on Build GRCh38
rs1033581393 Chr.19: 13935796-13935796 on Build GRCh38
rs938553347 Chr.19: 13935736-13935737 on Build GRCh38
rs778090413 Chr.19: 13935845-13935845 on Build GRCh38
rs371954013 Chr.19: 13935727-13935727 on Build GRCh38
rs774486136 Chr.19: 13935763-13935763 on Build GRCh38
rs535858344 Chr.19: 13935869-13935869 on Build GRCh38
rs568396399 Chr.19: 13935850-13935850 on Build GRCh38
rs150464916 Chr.19: 13935762-13935762 on Build GRCh38
rs779982571 Chr.19: 13935844-13935844 on Build GRCh38
rs767871297 Chr.19: 13935692-13935692 on Build GRCh38
rs763519569 Chr.19: 13935791-13935791 on Build GRCh38
rs757033145 Chr.19: 13935794-13935794 on Build GRCh38
rs749335730 Chr.19: 13935752-13935752 on Build GRCh38
rs142458800 Chr.19: 13935720-13935720 on Build GRCh38
rs750178163 Chr.19: 13935722-13935722 on Build GRCh38
rs761138263 Chr.19: 13935778-13935778 on Build GRCh38
rs761530024 Chr.19: 13935882-13935882 on Build GRCh38
rs766831138 Chr.19: 13935735-13935736 on Build GRCh38
rs779999172 Chr.19: 13935806-13935806 on Build GRCh38
rs752293561 Chr.19: 13935709-13935709 on Build GRCh38
rs925376434 Chr.19: 13935889-13935889 on Build GRCh38
rs748381095 Chr.19: 13935766-13935766 on Build GRCh38
rs754137982 Chr.19: 13935785-13935785 on Build GRCh38
rs369092727 Chr.19: 13935706-13935706 on Build GRCh38
rs773651048 Chr.19: 13935693-13935693 on Build GRCh38
rs142083249 Chr.19: 13935803-13935803 on Build GRCh38
rs140598542 Chr.19: 13935759-13935759 on Build GRCh38
rs370226496 Chr.19: 13935730-13935730 on Build GRCh38
rs545553915 Chr.19: 13935892-13935892 on Build GRCh38
rs202144687 Chr.19: 13935744-13935744 on Build GRCh38
rs151337979 Chr.19: 13935749-13935749 on Build GRCh38
rs772411067 Chr.19: 13935773-13935773 on Build GRCh38
rs774076038 Chr.19: 13935874-13935874 on Build GRCh38
rs367865065 Chr.19: 13935786-13935786 on Build GRCh38
rs764647690 Chr.19: 13935708-13935708 on Build GRCh38
rs748364837 Chr.19: 13935839-13935839 on Build GRCh38
rs762487027 Chr.19: 13935789-13935789 on Build GRCh38
rs200371573 Chr.19: 13935729-13935729 on Build GRCh38
rs868069592 Chr.19: 13935738-13935738 on Build GRCh38
rs375193197 Chr.19: 13935777-13935777 on Build GRCh38
rs145828342 Chr.19: 13935818-13935818 on Build GRCh38
rs1050457849 Chr.19: 13935758-13935758 on Build GRCh38
rs755880296 Chr.19: 13935805-13935805 on Build GRCh38
rs1043903854 Chr.19: 13935725-13935725 on Build GRCh38
rs535050778 Chr.19: 13935802-13935802 on Build GRCh38
rs749184667 Chr.19: 13935807-13935807 on Build GRCh38
rs761226189 Chr.19: 13935698-13935698 on Build GRCh38
rs969846082 Chr.19: 13935829-13935829 on Build GRCh38
rs147712582 Chr.19: 13935788-13935788 on Build GRCh38
rs568991640 Chr.19: 13935691-13935691 on Build GRCh38
rs897588875 Chr.19: 13935783-13935783 on Build GRCh38
rs747350670 Chr.19: 13935853-13935853 on Build GRCh38
rs571113982 Chr.19: 13935793-13935793 on Build GRCh38
rs771232909 Chr.19: 13935854-13935854 on Build GRCh38
rs575336476 Chr.19: 13935883-13935883 on Build GRCh38
rs745791303 Chr.19: 13935735-13935735 on Build GRCh38
rs772319918 Chr.19: 13935840-13935840 on Build GRCh38
rs868432124 Chr.19: 13935785-13935785 on Build GRCh38

View Full Product Details