Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106321479-106321738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCP11L2
Entrez Gene ID
255394
Gene Name
t-complex 11 like 2
Gene Aliases
-
UniGene
Hs.696047
Gene Chromosome Location
Chr.12: 106302026-106347455 on Build GRCh38
Transcripts
NM_001286262.1 NM_152772.2 XM_005268767.4 XM_005268768.4 XM_011538129.2 XM_011538130.2 XM_011538131.2 XM_011538132.2 XM_017019127.1 XM_017019128.1 XM_017019129.1 XM_017019130.1 XM_017019131.1 XM_017019132.1 XM_017019133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1944943 c.597A>T p.R199S Chr.12: 106321668-106321668 on Build GRCh38
1944941 c.454C>T p.R152C Chr.12: 106321525-106321525 on Build GRCh38
4038263 c.576C>T p.P192P Chr.12: 106321647-106321647 on Build GRCh38
7089066 c.576C>A p.P192P Chr.12: 106321647-106321647 on Build GRCh38
200382 c.580C>T p.R194* Chr.12: 106321651-106321651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765135038 Chr.12: 106321538-106321538 on Build GRCh38
rs747804995 Chr.12: 106321637-106321637 on Build GRCh38
rs780578715 Chr.12: 106321609-106321609 on Build GRCh38
rs758784304 Chr.12: 106321676-106321676 on Build GRCh38
rs374633600 Chr.12: 106321607-106321607 on Build GRCh38
rs768266247 Chr.12: 106321618-106321618 on Build GRCh38
rs772989988 Chr.12: 106321543-106321543 on Build GRCh38
rs1027995493 Chr.12: 106321661-106321661 on Build GRCh38
rs774335516 Chr.12: 106321652-106321652 on Build GRCh38
rs750751883 Chr.12: 106321597-106321597 on Build GRCh38
rs746564653 Chr.12: 106321617-106321617 on Build GRCh38
rs769616906 Chr.12: 106321695-106321695 on Build GRCh38
rs142176314 Chr.12: 106321554-106321554 on Build GRCh38
rs755466014 Chr.12: 106321692-106321692 on Build GRCh38
rs1001486661 Chr.12: 106321636-106321636 on Build GRCh38
rs750791537 Chr.12: 106321672-106321672 on Build GRCh38
rs772901806 Chr.12: 106321640-106321640 on Build GRCh38
rs763394230 Chr.12: 106321588-106321588 on Build GRCh38
rs529050823 Chr.12: 106321643-106321643 on Build GRCh38
rs752763419 Chr.12: 106321557-106321557 on Build GRCh38
rs368190626 Chr.12: 106321647-106321647 on Build GRCh38
rs138066544 Chr.12: 106321520-106321520 on Build GRCh38
rs769526802 Chr.12: 106321638-106321638 on Build GRCh38
rs764165615 Chr.12: 106321657-106321657 on Build GRCh38
rs761756927 Chr.12: 106321529-106321529 on Build GRCh38
rs891553515 Chr.12: 106321546-106321546 on Build GRCh38
rs760808655 Chr.12: 106321571-106321571 on Build GRCh38
rs372929065 Chr.12: 106321521-106321521 on Build GRCh38
rs766287437 Chr.12: 106321555-106321555 on Build GRCh38
rs552042741 Chr.12: 106321589-106321589 on Build GRCh38
rs768464120 Chr.12: 106321525-106321525 on Build GRCh38
rs201595663 Chr.12: 106321594-106321594 on Build GRCh38
rs138884019 Chr.12: 106321526-106321526 on Build GRCh38
rs951917388 Chr.12: 106321678-106321678 on Build GRCh38
rs779166835 Chr.12: 106321537-106321538 on Build GRCh38
rs900802762 Chr.12: 106321604-106321604 on Build GRCh38
rs919210927 Chr.12: 106321518-106321518 on Build GRCh38
rs759414837 Chr.12: 106321654-106321654 on Build GRCh38
rs79646734 Chr.12: 106321510-106321510 on Build GRCh38
rs757608534 Chr.12: 106321514-106321514 on Build GRCh38
rs762045600 Chr.12: 106321669-106321669 on Build GRCh38
rs754073751 Chr.12: 106321581-106321581 on Build GRCh38
rs533971528 Chr.12: 106321517-106321517 on Build GRCh38
rs904276062 Chr.12: 106321577-106321577 on Build GRCh38
rs138672494 Chr.12: 106321690-106321690 on Build GRCh38
rs765423965 Chr.12: 106321671-106321671 on Build GRCh38
rs749722416 Chr.12: 106321513-106321513 on Build GRCh38
rs867950257 Chr.12: 106321598-106321598 on Build GRCh38
rs922087384 Chr.12: 106321511-106321511 on Build GRCh38
rs756343446 Chr.12: 106321509-106321509 on Build GRCh38
rs763998268 Chr.12: 106321645-106321645 on Build GRCh38
rs150164085 Chr.12: 106321664-106321664 on Build GRCh38
rs764360195 Chr.12: 106321572-106321572 on Build GRCh38
rs752119908 Chr.12: 106321691-106321691 on Build GRCh38
rs777479623 Chr.12: 106321704-106321704 on Build GRCh38
rs1024181670 Chr.12: 106321613-106321613 on Build GRCh38
rs780788108 Chr.12: 106321625-106321625 on Build GRCh38
rs772592153 Chr.12: 106321519-106321519 on Build GRCh38
rs758876968 Chr.12: 106321599-106321599 on Build GRCh38
rs770844609 Chr.12: 106321648-106321648 on Build GRCh38
rs77806322 Chr.12: 106321693-106321693 on Build GRCh38

View Full Product Details